Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TACI Antibodies

          Invitrogen

          CD267 (TACI) Polyclonal Antibody, PeproTech®

          View all (24) TACI antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD267 (TACI) Polyclonal Antibody, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          CD267 (TACI) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CD267 (TACI) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD267 (TACI) Antibody (500-P166G-50UG) in WB

          Western Blot: To detect Human TACI by Western Blot analysis CD267 (TACI) Polyclonal Antibody (Product # 500-P166G-1MG) can be used at a concentration of 0.1- 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human TACI is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD267 (TACI) Antibody in Western Blot (WB)
          CD267 (TACI) Antibody in Western Blot (WB)
          CD267 (TACI) Antibody in ELISA (ELISA)
          CD267 (TACI) Polyclonal Antibody, PeproTech®

          Product Details

          500-P166G-50UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          ELISA (ELISA)

          0.5-2.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Goat

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived Recombinant Human TACI
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2929590

          Product Specific Information

          AA Sequence of recombinant protein: SGLGRSRRGG RSRVDQEERF PQGLWTGVAM RSCPEEQYWD PLLGTCMSCK TICNHQSQRT CAAFCRSLSC RKEQGKFYDH LLRDCISCAS ICGQHPKQCA YFCENKLRSP VNLPPELRRQ RSGEVENNSD NSGRYQGLEH RGSEASPALP GLKLSADQV.

          Preparation: Produced from sera of goats immunized with highly pure Recombinant Human TACI. Anti-Human TACI-specific antibody was purified by affinity chromatography employing an immobilized Human TACI matrix.

          Sandwich ELISA: To detect Human TACI by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5-2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TACI (500-P166GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TACI.

          Western Blot: To detect Human TACI by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human TACI is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          Target Information

          Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD267; Transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; Tumor necrosis factor receptor superfamily member 13B; unnamed protein product

          View more View less

          Gene Aliases: CD267; CVID; CVID2; IGAD2; RYZN; TACI; TNFRSF13B; TNFRSF14B

          View more View less

          UniProt ID: (Human) O14836

          View more View less

          Entrez Gene ID: (Human) 23495

          View more View less

          Function(s)
          protein binding signaling receptor activity
          Process(es)
          B cell homeostasis hematopoietic progenitor cell differentiation adaptive immune response immune system process cell surface receptor signaling pathway negative regulation of B cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline