Product References
Oxygen level is a critical regulator of human B cell differentiation and IgG class switch recombination.
Frontiers in immunology
Koers J,Marsman C,Steuten J,Tol S,Derksen NIL,Ten Brinke A,van Ham SM,Rispens T
25-0279-42 was used in Flow Cytometry to identify multiple mechanisms trough which oxygen pressure governs human B cell differentiation.
Tue Jan 03 00:00:00 EST 2023
Oxygen level is a critical regulator of human B cell differentiation and IgG class switch recombination.
Frontiers in immunology
Koers J,Marsman C,Steuten J,Tol S,Derksen NIL,Ten Brinke A,van Ham SM,Rispens T
25-0279-42 was used in Flow Cytometry to identify multiple mechanisms trough which oxygen pressure governs human B cell differentiation.
Tue Jan 03 00:00:00 EST 2023
Improving naive B cell isolation by absence of CD45RB glycosylation and CD27 expression in combination with BCR isotype.
European journal of immunology
Koers J,Pollastro S,Tol S,Niewold ITG,van Schouwenburg PA,de Vries N,Rispens T
25-0279-42 was used in Flow cytometry/Cell sorting to show that current naive B cell isolation strategies harbor contamination from non-naive B cells, and use of CD27-IgD+ is adequate but can be improved by including markers for CD45RB glycosylation and IgM.
Sat Oct 01 00:00:00 EDT 2022
Dynamics of NK, CD8 and Tfh cell mediated the production of cytokines and antiviral antibodies in Chinese patients with moderate COVID-19.
Journal of cellular and molecular medicine
Yan L,Cai B,Li Y,Wang MJ,An YF,Deng R,Li DD,Wang LC,Xu H,Gao XD,Wang LL
25-0279-42 was used in Flow Cytometry to study the changes of NK, T- and B-cell subsets, inflammatory cytokines and virus-specific antibodies in patients with moderate COVID-19. Subsets were analysed using flow cytometry, cytokines and SARS-CoV-2-specific antibodies were analysed using an electrochemiluminescence immunoassay.
Tue Dec 01 00:00:00 EST 2020
Expression of CD27 and CD28 on γδ T cells from the peripheral blood of patients with allergic rhinitis.
Experimental and therapeutic medicine
Wang Q,Sun Q,Chen Q,Li H,Liu D
Published figure using CD27 monoclonal antibody (Product # 25-0279-42) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2020
Developing Human Skin Contains Lymphocytes Demonstrating a Memory Signature.
Cell reports. Medicine
Dhariwala MO,Karthikeyan D,Vasquez KS,Farhat S,Weckel A,Taravati K,Leitner EG,Clancy S,Pauli M,Piper ML,Cohen JN,Ashouri JF,Lowe MM,Rosenblum MD,Scharschmidt TC
Published figure using CD27 monoclonal antibody (Product # 25-0279-42) in Flow Cytometry
Tue Nov 17 00:00:00 EST 2020
More than one antibody of individual B cells revealed by single-cell immune profiling.
Cell discovery
Shi Z,Zhang Q,Yan H,Yang Y,Wang P,Zhang Y,Deng Z,Yu M,Zhou W,Wang Q,Yang X,Mo X,Zhang C,Huang J,Dai H,Sun B,Zhao Y,Zhang L,Yang YG,Qiu X
Published figure using CD27 monoclonal antibody (Product # 25-0279-42) in Flow Cytometry
Fri Oct 20 00:00:00 EDT 2023
CD19+CD24hiCD38hi B Cells Are Expanded in Juvenile Dermatomyositis and Exhibit a Pro-Inflammatory Phenotype After Activation Through Toll-Like Receptor 7 and Interferon-α.
Frontiers in immunology
Piper CJM,Wilkinson MGL,Deakin CT,Otto GW,Dowle S,Duurland CL,Adams S,Marasco E,Rosser EC,Radziszewska A,Carsetti R,Ioannou Y,Beales PL,Kelberman D,Isenberg DA,Mauri C,Nistala K,Wedderburn LR
Wed Nov 20 00:00:00 EST 2019
Perturbed CD8+ T cell immunity across universal influenza epitopes in the elderly.
Journal of leukocyte biology
Nguyen THO,Sant S,Bird NL,Grant EJ,Clemens EB,Koutsakos M,Valkenburg SA,Gras S,Lappas M,Jaworowski A,Crowe J,Loh L,Kedzierska K
Published figure using CD27 monoclonal antibody (Product # 25-0279-42) in Flow Cytometry
Thu Feb 01 00:00:00 EST 2018
Clonally diverse CD38+HLA-DR+CD8+ T cells persist during fatal H7N9 disease.
Nature communications
Wang Z,Zhu L,Nguyen THO,Wan Y,Sant S,Quiñones-Parra SM,Crawford JC,Eltahla AA,Rizzetto S,Bull RA,Qiu C,Koutsakos M,Clemens EB,Loh L,Chen T,Liu L,Cao P,Ren Y,Kedzierski L,Kotsimbos T,McCaw JM,La Gruta NL,Turner SJ,Cheng AC,Luciani F,Zhang X,Doherty PC,Thomas PG,Xu J,Kedzierska K
25-0279 was used in Flow cytometry/Cell sorting to characterise the circulating CD8+T-cell profiles from patients hospitalised with avian H7N9, seasonal influenza A virus, and influenza vaccinees.
Mon Feb 26 00:00:00 EST 2018
Maintenance of the EBV-specific CD8+ TCRαβ repertoire in immunosuppressed lung transplant recipients.
Immunology and cell biology
Nguyen TH,Bird NL,Grant EJ,Miles JJ,Thomas PG,Kotsimbos TC,Mifsud NA,Kedzierska K
Published figure using CD27 monoclonal antibody (Product # 25-0279-42) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2017
Abnormal phenotypic features of IgM+B cell subsets in patients with chronic hepatitis C virus infection.
Experimental and therapeutic medicine
Kong F,Feng B,Zhang H,Rao H,Wang J,Cong X,Wei L
Published figure using CD27 monoclonal antibody (Product # 25-0279-42) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Antigen receptor-redirected T cells derived from hematopoietic precursor cells lack expression of the endogenous TCR/CD3 receptor and exhibit specific antitumor capacities.
Oncoimmunology
Van Caeneghem Y,De Munter S,Tieppo P,Goetgeluk G,Weening K,Verstichel G,Bonte S,Taghon T,Leclercq G,Kerre T,Debets R,Vermijlen D,Abken H,Vandekerckhove B
Published figure using CD27 monoclonal antibody (Product # 25-0279-42) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
IL-15 enhances the anti-tumor activity of trastuzumab against breast cancer cells but causes fatal side effects in humanized tumor mice (HTM).
Oncotarget
Wege AK,Weber F,Kroemer A,Ortmann O,Nimmerjahn F,Brockhoff G
25-0279 was used in Flow cytometry/Cell sorting to evaluate the effect of IL-15 in trastuzumab treated humanised tumour mice and HER2 positive breast cancer cells.
Tue Jan 10 00:00:00 EST 2017
High-dimensional assessment of B-cell responses to quadrivalent meningococcal conjugate and plain polysaccharide vaccine.
Genome medicine
O'Connor D,Clutterbuck EA,Thompson AJ,Snape MD,Ramasamy MN,Kelly DF,Pollard AJ
25027942 was used in flow cytometry to analyze B-cell responses to meningococcal polysaccharide and conjugate vaccines against Neisseria meningitidis
Mon Jan 30 00:00:00 EST 2017
Restricted isotype, distinct variable gene usage, and high rate of gp120 specificity of HIV-1 envelope-specific B cells in colostrum compared with those in blood of HIV-1-infected, lactating African women.
Mucosal immunology
Sacha CR,Vandergrift N,Jeffries TL,McGuire E,Fouda GG,Liebl B,Marshall DJ,Gurley TC,Stiegel L,Whitesides JF,Friedman J,Badiabo A,Foulger A,Yates NL,Tomaras GD,Kepler TB,Liao HX,Haynes BF,Moody MA,Permar SR
25-0279 was used in Flow cytometry/Cell sorting to suggest HIV-1-specific IgG1-secreting memory B cells selectively home to the mammary gland.
Sun Mar 01 00:00:00 EST 2015
Rapid Development of gp120-Focused Neutralizing B Cell Responses during Acute Simian Immunodeficiency Virus Infection of African Green Monkeys.
Journal of virology
Amos JD,Himes JE,Armand L,Gurley TC,Martinez DR,Colvin L,Beck K,Overman RG,Liao HX,Moody MA,Permar SR
25-0279 was used in Flow cytometry/Cell sorting to compare the envelope (env)-specific antibody responses of african green monkeys and rhesus macaques to simian immunodeficiency virus infection.
Tue Sep 01 00:00:00 EDT 2015
Reduced expression of CD27 by collagenase treatment: implications for interpreting b cell data in tissues.
PloS one
Shen C,Xu H,Alvarez X,Lackner AA,Veazey RS,Wang X
25-0279 was used in Flow cytometry/Cell sorting to suggest that CD27 may be a memory marker for B cells, but not for T cells, since essentially all CD3 T cells expressed CD27.
Wed Jan 13 00:00:00 EST 2016
Phenotypic, genomic and functional characterization reveals no differences between CD138++ and CD138low subpopulations in multiple myeloma cell lines.
PloS one
Paíno T,Sarasquete ME,Paiva B,Krzeminski P,San-Segundo L,Corchete LA,Redondo A,Garayoa M,García-Sanz R,Gutiérrez NC,Ocio EM,San-Miguel JF
25-0279 was used in Flow cytometry/Cell sorting to investigate the phenotype of multiple myeloma cancer stem cells, showing no differences between CD138++ and CD138low subpopulations.
Tue Jan 20 00:00:00 EST 2015
Follicular helper T cells serve as the major CD4 T cell compartment for HIV-1 infection, replication, and production.
The Journal of experimental medicine
Perreau M,Savoye AL,De Crignis E,Corpataux JM,Cubas R,Haddad EK,De Leval L,Graziosi C,Pantaleo G
25-0279 was used in Flow cytometry/Cell sorting to demonstrate that T follicular helper cells serve as the major CD4 T-cell compartment for HIV infection, replication, and production.
Mon Jan 14 00:00:00 EST 2013
Adipose tissue NK cells manifest an activated phenotype in human obesity.
Metabolism: clinical and experimental
O'Rourke RW,Gaston GD,Meyer KA,White AE,Marks DL
25-0279 was used in Flow cytometry/Cell sorting to determine the phenotype of human adipose tissue natural killer cells.
Fri Nov 01 00:00:00 EDT 2013
Rabbit antithymocyte globulin (thymoglobulin) impairs the thymic output of both conventional and regulatory CD4+ T cells after allogeneic hematopoietic stem cell transplantation in adult patients.
Haematologica
Na IK,Wittenbecher F,Dziubianau M,Herholz A,Mensen A,Kunkel D,Blau O,Blau I,Thiel E,Uharek L,Scheibenbogen C,Rieger K,Thiel A
25-0279 was used in Flow cytometry/Cell sorting to evaluate the effects of rabbit antithymocyte globulin-Genzyme™ on T cell kinetics in adult patients undergoing allogeneic hematopoietic stem cell transplantation.
Tue Jan 01 00:00:00 EST 2013
Long-lived Plasmodium falciparum specific memory B cells in naturally exposed Swedish travelers.
European journal of immunology
Ndungu FM,Lundblom K,Rono J,Illingworth J,Eriksson S,Färnert A
25-0279 was used in Flow cytometry/Cell sorting to expose long-lived Plasmodium falciparum specific memory B cells in naturally exposed Swedish travelers.
Fri Nov 01 00:00:00 EDT 2013
Long-term immune reconstitution of naive and memory t cell pools after haploidentical hematopoietic stem cell transplantation.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
Azevedo RI,Soares MV,Albuquerque AS,Tendeiro R,Soares RS,Martins M,Ligeiro D,Victorino RM,Lacerda JF,Sousa AE
25-0279 was used in Flow cytometry/Cell sorting to show that long-term immune reconstitution was successfully achieved after haploidentical hematopoietic stem cell transplantation, a process that appears to have largely relied on de novo T cell production.
Wed May 01 00:00:00 EDT 2013
Chronic exposure to Plasmodium falciparum is associated with phenotypic evidence of B and T cell exhaustion.
Journal of immunology (Baltimore, Md. : 1950)
Illingworth J,Butler NS,Roetynck S,Mwacharo J,Pierce SK,Bejon P,Crompton PD,Marsh K,Ndungu FM
25-0279 was used in Flow cytometry/Cell sorting to study B and T cell exhaustion caused by chronic exposure to Plasmodium falciparum.
Fri Feb 01 00:00:00 EST 2013
B cell analysis of ethnic groups in Mali with differential susceptibility to malaria.
Malaria journal
Portugal S,Doumtabe D,Traore B,Miller LH,Troye-Blomberg M,Doumbo OK,Dolo A,Pierce SK,Crompton PD
25-0279 was used in Flow cytometry/Cell sorting to determine the mechanisms underlying the elevated resistance to malaria observed in the Fulani ethnic group.
Fri May 11 00:00:00 EDT 2012
Long-term immunologically competent human peripheral lymphoid tissue cultures in a 3D bioreactor.
Biotechnology and bioengineering
Kuzin I,Sun H,Moshkani S,Feng C,Mantalaris A,Wu JH,Bottaro A
Published figure using CD27 monoclonal antibody (Product # 25-0279-42) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2011
Vaccine-elicited SIV and HIV envelope-specific IgA and IgG memory B cells in rhesus macaque peripheral blood correlate with functional antibody responses and reduced viremia.
Vaccine
Brocca-Cofano E,McKinnon K,Demberg T,Venzon D,Hidajat R,Xiao P,Daltabuit-Test M,Patterson LJ,Robert-Guroff M
25-0279 was used in Flow cytometry/Cell sorting to evaluate the memory B cell response in immunised rhesus macaques following exposure to simian immunodeficiency virus.
Tue Apr 12 00:00:00 EDT 2011
Chronic lymphocytic leukaemia (CLL) and CLL-type monoclonal B-cell lymphocytosis (MBL) show differential expression of molecules involved in lymphoid tissue homing.
Cytometry. Part B, Clinical cytometry
Rawstron AC,Shingles J,de Tute R,Bennett F,Jack AS,Hillmen P
25-0279 was used in Flow cytometry/Cell sorting to characterise the cell surface markers used to discriminate Chronic lymphocytic leukaemia (CLL)-type monoclonal B-cell lymphocytosis from CLL, or identify the likelihood of stable disease in CLL cases.
Thu Dec 23 00:00:00 EST 2010