Product References
Blockade of TLR2 and TLR4 Attenuates Inflammatory Response and Parasite Load in Cutaneous Leishmaniasis.
Frontiers in immunology
Carneiro PP,Dórea AS,Oliveira WN,Guimarães LH,Brodskyn C,Carvalho EM,Bacellar O
12-9922-42 was used in Flow Cytometry to show that reduction in oxidative burst, IL-1β, TNF and CXCL9 production were observed.
Mon Dec 13 00:00:00 EST 2021
Blockade of TLR2 and TLR4 Attenuates Inflammatory Response and Parasite Load in Cutaneous Leishmaniasis.
Frontiers in immunology
Carneiro PP,Dórea AS,Oliveira WN,Guimarães LH,Brodskyn C,Carvalho EM,Bacellar O
12-9922-42 was used in Flow Cytometry to show that reduction in oxidative burst, IL-1β, TNF and CXCL9 production were observed.
Mon Dec 13 00:00:00 EST 2021
TLR2 on blood monocytes senses dengue virus infection and its expression correlates with disease pathogenesis.
Nature communications
Aguilar-Briseño JA,Upasani V,Ellen BMT,Moser J,Pauzuolis M,Ruiz-Silva M,Heng S,Laurent D,Choeung R,Dussart P,Cantaert T,Smit JM,Rodenhuis-Zybert IA
12-9922 was used in Flow cytometry/Cell sorting to demonstrate that TLR2, together with its co-receptors CD14 and TLR6, is an innate sensor of dengue virus particles.
Tue Jun 23 00:00:00 EDT 2020
TLR2 on blood monocytes senses dengue virus infection and its expression correlates with disease pathogenesis.
Nature communications
Aguilar-Briseño JA,Upasani V,Ellen BMT,Moser J,Pauzuolis M,Ruiz-Silva M,Heng S,Laurent D,Choeung R,Dussart P,Cantaert T,Smit JM,Rodenhuis-Zybert IA
12-9922 was used in Flow cytometry/Cell sorting to demonstrate that TLR2, together with its co-receptors CD14 and TLR6, is an innate sensor of dengue virus particles.
Tue Jun 23 00:00:00 EDT 2020
Patient iPSC-Derived Macrophages to Study Inborn Errors of the IFN-γ Responsive Pathway.
Cells
Haake K,Neehus AL,Buchegger T,Kühnel MP,Blank P,Philipp F,Oleaga-Quintas C,Schulz A,Grimley M,Goethe R,Jonigk D,Kalinke U,Boisson-Dupuis S,Casanova JL,Bustamante J,Lachmann N
12-9922 was used in Flow cytometry/Cell sorting to report a disease modeling platform to study the role of macrophages in patients with inborn errors of IFN-γ immunity.
Wed Feb 19 00:00:00 EST 2020
Activation of the Toll‑like receptor 2 signaling pathway inhibits the proliferation of HCC cells in vitro.
Oncology reports
Chen Y,Huang Z,Chen X,Ye H
Published figure using CD282 (TLR2) monoclonal antibody (Product # 12-9922-42) in Flow Cytometry
Sun Dec 01 00:00:00 EST 2019
Dietary Toll-Like Receptor Stimulants Promote Hepatic Inflammation and Impair Reverse Cholesterol Transport in Mice via Macrophage-Dependent Interleukin-1 Production.
Frontiers in immunology
Faraj TA,Stover C,Erridge C
Published figure using CD282 (TLR2) monoclonal antibody (Product # 12-9922-42) in Flow Cytometry
Mon Oct 19 00:00:00 EDT 2020
n-butanol extract from Folium isatidis inhibits the lipopolysaccharide-induced downregulation of CXCR1 and CXCR2 on human neutrophils.
Molecular medicine reports
Wu B,Wang L,Jiang L,Dong L,Xu F,Lu Y,Jin J,Wang Z,Liang G,Shan X
Published figure using CD282 (TLR2) monoclonal antibody (Product # 12-9922-42) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Toll-like receptor chaperone HSP90B1 and the immune response to Mycobacteria.
PloS one
Graustein AD,Misch EA,Musvosvi M,Shey M,Shah JA,Seshadri C,Ajuogu A,Bowman K,Mulenga H,Veldsman A,Hanekom WA,Hatherill M,Scriba TJ,Hawn TR
Published figure using CD282 (TLR2) monoclonal antibody (Product # 12-9922-42) in Flow Cytometry
Tue May 07 00:00:00 EDT 2019
High-density lipoprotein suppresses tumor necrosis factor alpha production by mycobacteria-infected human macrophages.
Scientific reports
Inoue M,Niki M,Ozeki Y,Nagi S,Chadeka EA,Yamaguchi T,Osada-Oka M,Ono K,Oda T,Mwende F,Kaneko Y,Matsumoto M,Kaneko S,Ichinose Y,Njenga SM,Hamano S,Matsumoto S
12-9922 was used in Flow cytometry/Cell sorting to investigate the role of high-density lipoprotein (HDL) in mycobacteria-infected macrophages, and demonstrate that HDL suppresses the production of pro-infammatory cytokines.
Mon Apr 30 00:00:00 EDT 2018
Toll-Interleukin 1 Receptor Domain-Containing Adaptor Protein 180L Single-Nucleotide Polymorphism Is Associated With Susceptibility to Recurrent Pneumococcal Lower Respiratory Tract Infections in Children.
Frontiers in immunology
Siebert JN,Hamann L,Verolet CM,Gameiro C,Grillet S,Siegrist CA,Posfay-Barbe KM
12-9922 was used in Flow cytometry/Cell sorting to asses whether a defect somewhere along this toll-like receptor signally pathways increases susceptibility to recurrent pneumococcal low respiratory tract infections.
Mon Sep 16 00:00:00 EDT 2019
Early detection of neonatal group B streptococcus sepsis and the possible diagnostic utility of IL-6, IL-8, and CD11b in a human umbilical cord blood in vitro model.
Infection and drug resistance
Nakstad B,Sonerud T,Solevåg AL
Published figure using CD282 (TLR2) monoclonal antibody (Product # 12-9922-42) in Flow Cytometry
Fri Jul 29 00:00:00 EDT 2016
Early detection of neonatal group B streptococcus sepsis and the possible diagnostic utility of IL-6, IL-8, and CD11b in a human umbilical cord blood in vitro model.
Infection and drug resistance
Nakstad B,Sonerud T,Solevåg AL
Published figure using CD282 (TLR2) monoclonal antibody (Product # 12-9922-42) in Flow Cytometry
Fri Jul 29 00:00:00 EDT 2016
Interferon-γ enhances both the anti-bacterial and the pro-inflammatory response of human mast cells to Staphylococcus aureus.
Immunology
Swindle EJ,Brown JM,Rådinger M,DeLeo FR,Metcalfe DD
12-9922 was used in Flow cytometry/Cell sorting to investigate the effect of interferon-γ on the biological responses of human mast cells to Staphylococcus aureus.
Sun Nov 01 00:00:00 EDT 2015
In vivo pre-activation of monocytes in patients with axial spondyloarthritis.
Arthritis research & therapy
Conrad K,Wu P,Sieper J,Syrbe U
12-9922 was used in Flow cytometry/Cell sorting to demonstrate in vivo pre-activation of monocytes in axial spondyloarthritis patients indicated by enhanced cytokine production by monocytes.
Thu Jul 16 00:00:00 EDT 2015
Interactions between alveolar epithelial cells and neutrophils under pro-inflammatory conditions.
European clinical respiratory journal
von Schéele I,Larsson K,Palmberg L
12-9922 was used in Flow cytometry/Cell sorting to investigate communication between cells constituting the first line of defence under infection from chronic obstructive pulmonary disease.
Wed Nov 11 00:00:00 EST 2015
The discriminative capacity of soluble Toll-like receptor (sTLR)2 and sTLR4 in inflammatory diseases.
BMC immunology
Ten Oever J,Kox M,van de Veerdonk FL,Mothapo KM,Slavcovici A,Jansen TL,Tweehuysen L,Giamarellos-Bourboulis EJ,Schneeberger PM,Wever PC,Stoffels M,Simon A,van der Meer JW,Johnson MD,Kullberg BJ,Pickkers P,Pachot A,Joosten LA,Netea MG
12-9922 was used in Flow cytometry/Cell sorting to suggest the possibility to use sTLRs as a diagnostic tool for inflammatory conditions.
Wed Nov 19 00:00:00 EST 2014
Biologically active polymers from spontaneous carotenoid oxidation: a new frontier in carotenoid activity.
PloS one
Johnston JB,Nickerson JG,Daroszewski J,Mogg TJ,Burton GW
12-9922 was used in Flow cytometry/Cell sorting to study carotenoid activity via biologically active polymers from spontaneous carotenoid oxidation.
Thu Jun 25 00:00:00 EDT 2015
Development of monocyte Toll-like receptor 2 and Toll-like receptor 4 in preterm newborns during the first few months of life.
Pediatric research
Shen CM,Lin SC,Niu DM,Kou YR
12-9922 was used in Flow Cytometry to study the development of Toll-like receptor 2 and Toll-like receptor 4 at birth in preterm newborns and during the first few months of life.
Wed May 01 00:00:00 EDT 2013
Reactivation of latent HIV-1 in central memory CD4⁺ T cells through TLR-1/2 stimulation.
Retrovirology
Novis CL,Archin NM,Buzon MJ,Verdin E,Round JL,Lichterfeld M,Margolis DM,Planelles V,Bosque A
12-9922 was used in Flow cytometry/Cell sorting to evaluate the ability of toll-like receptor ligands to reactivate latent HIV-1 within central memory CD4+ T cells.
Thu Oct 24 00:00:00 EDT 2013
Age-related defects in TLR2 signaling diminish the cytokine response by alveolar macrophages during murine pneumococcal pneumonia.
Experimental gerontology
Boyd AR,Shivshankar P,Jiang S,Berton MT,Orihuela CJ
12-9922 was used in Flow cytometry/Cell sorting to investigate the extent of the impact of age-related deficiencies in Toll-like receptor function.
Sun Jul 01 00:00:00 EDT 2012
Toll-like receptor expression in smokers with and without COPD.
Respiratory medicine
von Scheele I,Larsson K,Dahlén B,Billing B,Skedinger M,Lantz AS,Palmberg L
12-9922 was used in Flow cytometry/Cell sorting to study Toll-like receptor expression in smokers with and without chronic obstructive pulmonary disease.
Mon Aug 01 00:00:00 EDT 2011
Bone marrow mononuclear cells up-regulate toll-like receptor expression and produce inflammatory mediators in response to cigarette smoke extract.
PloS one
Zhou J,Eksioglu EA,Fortenbery NR,Chen X,Wang H,Epling-Burnette PK,Djeu JY,Wei S
12-9922 was used in Flow cytometry/Cell sorting to elucidate the effects of cigarette smoke on human bone marrow haematopoiesis and characterise the inflammatory response.
Thu Dec 01 00:00:00 EST 2011
Differential but direct abolishment of human regulatory T cell suppressive capacity by various TLR2 ligands.
Journal of immunology (Baltimore, Md. : 1950)
Oberg HH,Ly TT,Ussat S,Meyer T,Kabelitz D,Wesch D
12-9922 was used in Flow cytometry/Cell sorting to investigate whether TLR2 ligands can abolish human regulatory T cell suppressive capacity.
Sat May 01 00:00:00 EDT 2010
Human neonatal peripheral blood leukocytes demonstrate pathogen-specific coordinate expression of TLR2, TLR4/MD2, and MyD88 during bacterial infection in vivo.
Pediatric research
Zhang JP,Yang Y,Levy O,Chen C
12-9922 was used in Flow cytometry/Cell sorting to suggest key roles for innate immune pathways in neonatal responses to G+ and G- bacterial infection.
Wed Dec 01 00:00:00 EST 2010
Interleukin-25 inhibits interleukin-12 production and Th1 cell-driven inflammation in the gut.
Gastroenterology
Caruso R,Sarra M,Stolfi C,Rizzo A,Fina D,Fantini MC,Pallone F,MacDonald TT,Monteleone G
12-9922 was used in Flow cytometry/Cell sorting to demonstrate that IL-25 inhibits CD14+ cell-derived cytokines and experimental colitis, so could be a treatment for Crohn's disease and ulcerative colitis.
Mon Jun 01 00:00:00 EDT 2009
The role of infection in the development of non-valvular atrial fibrillation: up-regulation of Toll-like receptor 2 expression levels on monocytes.
Journal of cardiology
Ichiki H,Orihara K,Hamasaki S,Ishida S,Oketani N,Iriki Y,Ninomiya Y,Okui H,Kuwahata S,Fujita S,Matsushita T,Yoshifuku S,Oba R,Hirai H,Nagata K,Tei C
12-9922 was used in Flow cytometry/Cell sorting to explore whether expression of TLR2 levels are up-regulated in patients with non-valvular atrial fibrillation.
Sun Feb 01 00:00:00 EST 2009
Expression of cell surface receptors and oxidative metabolism modulation in the clinical continuum of sepsis.
Critical care (London, England)
Martins PS,Brunialti MK,Martos LS,Machado FR,Assunçao MS,Blecher S,Salomao R
12-9922 was used in Flow cytometry/Cell sorting to demonstrate that expression of surface receptors on neutrophils might be modulated across different severities of sepsis depending on the receptor.
Mon Jun 23 00:00:00 EDT 2008
Age-related differences in sensitivity of peripheral blood monocytes to lipopolysaccharide and Staphylococcus aureus toxin B in atopic dermatitis.
The Journal of investigative dermatology
Mandron M,Ariès MF,Boralevi F,Martin H,Charveron M,Taieb A,Davrinche C
Tue Apr 01 00:00:00 EDT 2008
Effects of vitamin D on expression of Toll-like receptors of monocytes from patients with Behcet's disease.
Rheumatology (Oxford, England)
Do JE,Kwon SY,Park S,Lee ES
Sun Jun 01 00:00:00 EDT 2008
Human Langerhans cells selectively activated via Toll-like receptor 2 agonists acquire migratory and CD4+T cell stimulatory capacity.
Journal of leukocyte biology
Peiser M,Koeck J,Kirschning CJ,Wittig B,Wanner R
12-9922 was used in Flow cytometry/Cell sorting to analyse the Toll-like receptor (TLR) phenotype of epidermal Langerhans cells treated with various TLR agonists.
Thu May 01 00:00:00 EDT 2008
Antibodies specific for human or murine Toll-like receptors detect canine leukocytes by flow cytometry.
Veterinary immunology and immunopathology
Burgener IA,Jungi TW
Tue Jul 15 00:00:00 EDT 2008
Differential regulation of CCL-11/eotaxin-1 and CXCL-8/IL-8 by gram-positive and gram-negative bacteria in human airway smooth muscle cells.
Respiratory research
Issa R,Sorrentino R,Sukkar MB,Sriskandan S,Chung KF,Mitchell JA
12-9922 was used in Flow cytometry/Cell sorting to show that Gram-positive or Gram-negative bacteria cause human airway smooth muscle cells to release the neutrophil selective chemokine CXCL-8.
Tue Apr 01 00:00:00 EDT 2008
Immaturity of infection control in preterm and term newborns is associated with impaired toll-like receptor signaling.
The Journal of infectious diseases
Sadeghi K,Berger A,Langgartner M,Prusa AR,Hayde M,Herkner K,Pollak A,Spittler A,Forster-Waldl E
12-9922 was used in Flow cytometry/Cell sorting to show that the reduced functional response to components of the bacterial cell wall might predispose premature newborns to bacterial infection.
Mon Jan 15 00:00:00 EST 2007
Changes in Toll-like receptor (TLR)-2 and TLR4 expression and function but not polymorphisms are associated with acute anterior uveitis.
Investigative ophthalmology & visual science
Chang JH,Hampartzoumian T,Everett B,Lloyd A,McCluskey PJ,Wakefield D
12-9922 was used in Flow cytometry/Cell sorting to suggest that microbial triggers and TLRs play a role in the pathogenesis of acute anterior uveitis.
Sun Apr 01 00:00:00 EDT 2007
Toll-like receptor 3 (TLR3): a new marker of canine monocytes-derived dendritic cells (cMo-DC).
Veterinary immunology and immunopathology
Bonnefont-Rebeix C,Marchal T,Bernaud J,Pin JJ,Leroux C,Lebecque S,Chabanne L,Rigal D
Sun Jul 15 00:00:00 EDT 2007
Toll-like receptor 2, 3, and 4 expression and function in human airway smooth muscle.
The Journal of allergy and clinical immunology
Sukkar MB,Xie S,Khorasani NM,Kon OM,Stanbridge R,Issa R,Chung KF
12-9922 was used in Flow cytometry/Cell sorting to determine the effects of cytokines, TLR ligands, and corticosteroids on TLR expression in human airway smooth muscle cells.
Fri Sep 01 00:00:00 EDT 2006
Induction of interleukin-6 by hepatitis C virus core protein in hepatitis C-associated mixed cryoglobulinemia and B-cell non-Hodgkin's lymphoma.
Clinical cancer research : an official journal of the American Association for Cancer Research
Feldmann G,Nischalke HD,Nattermann J,Banas B,Berg T,Teschendorf C,Schmiegel W,Dührsen U,Halangk J,Iwan A,Sauerbruch T,Caselmann WH,Spengler U
12-9922 was used in Functional assays to study whether recognition of hepatitis C virus proteins by pattern recognition receptors is involved in the pathogenesis of HCV-associated mixed cryoglobulinemia and B-cell non-Hodgkin's lymphoma.
Tue Aug 01 00:00:00 EDT 2006
Expression of Toll-like receptor 2 is up-regulated in monocytes from patients with chronic obstructive pulmonary disease.
Respiratory research
Pons J,Sauleda J,Regueiro V,Santos C,López M,Ferrer J,Agustí AG,Bengoechea JA
12-9922 was used in Flow cytometry/Cell sorting to reveal abnormalities in TLRs expression in chronic obstructive pulmonary disease patients and highlight its potential relationship with systemic inflammation.
Mon Apr 10 00:00:00 EDT 2006
Neonatal immune responses to microbial stimuli: is there an influence of maternal allergy?
The Journal of allergy and clinical immunology
Amoudruz P,Holmlund U,Malmström V,Trollmo C,Bremme K,Scheynius A,Sverremark-Ekström E
12-9922 was used in Flow cytometry/Cell sorting to suggest that cord blood monocytes and cord blood mononuclear blood cell immune responses are influenced by maternal allergy.
Wed Jun 01 00:00:00 EDT 2005
Toll-like receptor 2 expression on human conjunctival epithelial cells: a pathway for Staphylococcus aureus involvement in chronic ocular proinflammatory responses.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Cook EB,Stahl JL,Esnault S,Barney NP,Graziano FM
Fri Apr 01 00:00:00 EST 2005
Peptidoglycan of Staphylococcus aureus upregulates monocyte expression of CD14, Toll-like receptor 2 (TLR2), and TLR4 in human blood: possible implications for priming of lipopolysaccharide signaling.
Infection and immunity
Hadley JS,Wang JE,Foster SJ,Thiemermann C,Hinds CJ
12-9922 was used in Flow cytometry/Cell sorting to show that TLR2 and TLR4 upregulation precedes the synergistic effects of lipopolysaccharide and peptidoglycan on cytokine release.
Tue Nov 01 00:00:00 EST 2005
Corticosteroid and cytokines synergistically enhance toll-like receptor 2 expression in respiratory epithelial cells.
American journal of respiratory cell and molecular biology
Homma T,Kato A,Hashimoto N,Batchelor J,Yoshikawa M,Imai S,Wakiguchi H,Saito H,Matsumoto K
12-9922 was used in Flow cytometry/Cell sorting to investigate the involvement of TLR family molecules in corticosteroid treatment.
Fri Oct 01 00:00:00 EDT 2004
Intracellularly expressed TLR2s and TLR4s contribution to an immunosilent environment at the ocular mucosal epithelium.
Journal of immunology (Baltimore, Md. : 1950)
Ueta M,Nochi T,Jang MH,Park EJ,Igarashi O,Hino A,Kawasaki S,Shikina T,Hiroi T,Kinoshita S,Kiyono H
Wed Sep 01 00:00:00 EDT 2004
Membrane-anchored CD14 is important for induction of interleukin-8 by lipopolysaccharide and peptidoglycan in uroepithelial cells.
Clinical and diagnostic laboratory immunology
Shimizu T,Yokota S,Takahashi S,Kunishima Y,Takeyama K,Masumori N,Takahashi A,Matsukawa M,Itoh N,Tsukamoto T,Fujii N
12-9922 was used in Flow cytometry/Cell sorting to identify factors promoting IL-8 induction in bladder epithelial cells in response to bacterial components.
Wed Sep 01 00:00:00 EDT 2004
Expression of Toll-like receptor 2 on CD16+ blood monocytes and synovial tissue macrophages in rheumatoid arthritis.
Arthritis and rheumatism
Iwahashi M,Yamamura M,Aita T,Okamoto A,Ueno A,Ogawa N,Akashi S,Miyake K,Godowski PJ,Makino H
Sat May 01 00:00:00 EDT 2004
The urokinase receptor can be induced by Borrelia burgdorferi through receptors of the innate immune system.
Infection and immunity
Coleman JL,Benach JL
Wed Oct 01 00:00:00 EDT 2003
Toll-like receptor 2 is expressed by alveolar epithelial cells type II and macrophages in the human lung.
Histochemistry and cell biology
Droemann D,Goldmann T,Branscheid D,Clark R,Dalhoff K,Zabel P,Vollmer E
Sat Feb 01 00:00:00 EST 2003
Immunohistochemical localization of Toll-like receptors 2 and 4 in gingival tissue from patients with periodontitis.
Oral microbiology and immunology
Mori Y,Yoshimura A,Ukai T,Lien E,Espevik T,Hara Y
Sat Feb 01 00:00:00 EST 2003