Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD3 Antibodies

          Invitrogen

          CD3 Monoclonal Antibody (17A2), APC, eBioscience™

          66 Published Figures
          154 References
          View all (584) CD3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD3 Monoclonal Antibody (17A2), APC, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (66)
          CD3 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD3 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 67

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD3 Antibody (17-0032-82) in Flow

          Staining of C57Bl/6 splenocytes with Anti-Human/Mouse CD45R (B220) FITC (Product # 11-0452-82) and 0.25 µg of Rat IgG2b Isotype Control APC (Product # 17-4031-82) (left) or 0.25 µg of Anti-Mouse CD3 APC (right). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Flow Cytometry (Flow)
          CD3 Antibody in Immunocytochemistry (ICC/IF)
          CD3 Antibody in Immunohistochemistry (IHC)

          Product Details

          17-0032-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 3 publications 3 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 146 publications 146 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          17A2

          Conjugate

          APC APC APC
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 700 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 650
          • eFluor 450
          • eFluor 506
          • eFluor 660
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 600
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10597589

          Product Specific Information

          Description: The 17A2 monoclonal antibody reacts with the mouse CD3 complex. CD3 subunits gamma, delta and epsilon are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 17A2 to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation.

          Applications Reported: This 17A2 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 17A2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          The CD3 complex, composed of gamma, delta, epsilon, and zeta subunits, is essential for the assembly, trafficking, and surface expression of the T cell receptor (TCR) complex. These subunits are structurally related members of the immunoglobulin superfamily and are encoded by closely linked genes on human chromosome 11. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, but not on B or NK cells. The CD3 subunits play a crucial role in transducing antigen-recognition signals into the cytoplasm of T cells. The cytoplasmic tails of CD3 subunits contain a double tyrosine-based motif that associates with cytoplasmic signal transduction molecules, mediating T cell activation through the TCR. Crosslinking of the TCR initiates intracellular biochemical pathways that result in cellular activation, proliferation, and potentially growth arrest and cell survival. CD3 is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes, and Purkinje cells in the cerebellum. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. Defects in the CD3 gene are associated with CD3 immunodeficiency, highlighting its importance in immune function and regulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD247; CD247 antigen; CD3 antigen, zeta polypeptide; CD3 TCR complex; CD3-zeta/eta; CD3d; CD3e; CD3g; FLJ18683; Leu-4; T cell antigen receptor complex epsilon subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 eta chain; T-cell receptor T3 gamma chain; T-cell receptor T3 zeta chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell surface glycoprotein CD3 zeta chain; T3/TCR complex

          View more View less

          Gene Aliases: 4930549J05Rik; A430104F18Rik; AI504783; AW552088; Cd247; CD3; Cd3-eta; Cd3-zeta; Cd3d; Cd3e; CD3epsilon; Cd3g; Cd3h; Cd3z; Cd3zeta; Ctg-3; Ctg3; T3d; T3e; T3g; T3z; Tcrk; Tcrz

          View more View less

          UniProt ID: (Mouse) P24161, (Mouse) P04235, (Mouse) P22646, (Mouse) P11942

          View more View less

          Entrez Gene ID: (Mouse) 12503, (Mouse) 12500, (Mouse) 12501, (Mouse) 12502

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding identical protein binding protein heterodimerization activity signal transducer activity SH3 domain binding defense/immunity protein immunoglobulin receptor superfamily
          Process(es)
          cell surface receptor signaling pathway T cell receptor signaling pathway protein complex assembly T cell activation positive thymic T cell selection positive regulation of T cell anergy response to nutrient T cell costimulation positive regulation of interferon-gamma production positive regulation of interleukin-4 production positive regulation of T cell proliferation negative thymic T cell selection positive regulation of interleukin-2 biosynthetic process negative regulation of smoothened signaling pathway positive regulation of alpha-beta T cell proliferation lymphocyte activation positive regulation of peptidyl-tyrosine phosphorylation positive regulation of calcium-mediated signaling positive regulation of T cell activation apoptotic signaling pathway establishment or maintenance of cell polarity protein transport regulation of lymphocyte apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-fcf5f9d6d-l96vj:80/100.66.74.142:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0