Product References
Type H vessel/platelet-derived growth factor receptor β+ perivascular cell disintegration is involved in vascular injury and bone loss in radiation-induced bone damage.
Cell proliferation
Li J,Chen X,Ren L,Chen X,Wu T,Wang Y,Ren X,Cheng B,Xia J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2023
LBP1C-2 from Lycium barbarum maintains skeletal muscle satellite cell pool by interaction with FGFR1.
iScience
Meng J,Lv Z,Chen X,Sun C,Jin C,Ding K,Chen C
12-0311-82 was used in Flow cytometry/Cell sorting to reveal that LBP1C-2 might bind to FGFR1 to activate SCs and promote SC self-renewal through Spry1 upregulation.
Fri May 19 00:00:00 EDT 2023
The Wnt/TCF7L1 transcriptional repressor axis drives primitive endoderm formation by antagonizing naive and formative pluripotency.
Nature communications
Athanasouli P,Balli M,De Jaime-Soguero A,Boel A,Papanikolaou S,van der Veer BK,Janiszewski A,Vanhessche T,Francis A,El Laithy Y,Nigro AL,Aulicino F,Koh KP,Pasque V,Cosma MP,Verfaillie C,Zwijsen A,Heindryckx B,Nikolaou C,Lluis F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Mar 03 00:00:00 EST 2023
Regenerative potential of multinucleated cells: bone marrow adiponectin-positive multinucleated cells take the lead.
Stem cell research & therapy
Moein S,Ahmadbeigi N,Adibi R,Kamali S,Moradzadeh K,Nematollahi P,Nardi NB,Gheisari Y
12-0311-82 was used in Immunocytochemistry to investigate the presence and characteristics of large multinucleated cells (LMCs) in native bone marrow (BM), demonstrating that LMCs play a crucial role in BM regeneration and can give rise to both stromal and hematopoietic lineages, contributing to tissue repair and the reconstitution of the hematopoietic system.
Tue Jul 04 00:00:00 EDT 2023
Autophagy induction during stem cell activation plays a key role in salivary gland self-renewal.
Autophagy
Orhon I,Rocchi C,Villarejo-Zori B,Serrano Martinez P,Baanstra M,Brouwer U,Boya P,Coppes R,Reggiori F
12-0311-82 was used in Flow Cytometry to conclude that autophagy is a key pathway for self-renewal activation in low proliferative adult tissues, and its pharmacological manipulation has the potential to promote tissue regeneration.
Tue Feb 01 00:00:00 EST 2022
Isolation and in vitro characterization of murine young-adult long bone skeletal progenitors.
Frontiers in endocrinology
Loopmans S,Stockmans I,Carmeliet G,Stegen S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Aug 19 00:00:00 EDT 2022
Synovial Fibroblast Sialylation Regulates Cell Migration and Activation of Inflammatory Pathways in Arthritogenesis.
Frontiers in immunology
Wang Y,Pan P,Khan A,Çil Ç,Pineda MA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Apr 08 00:00:00 EDT 2022
Lamin A/C-dependent chromatin architecture safeguards naïve pluripotency to prevent aberrant cardiovascular cell fate and function.
Nature communications
Wang Y,Elsherbiny A,Kessler L,Cordero J,Shi H,Serke H,Lityagina O,Trogisch FA,Mohammadi MM,El-Battrawy I,Backs J,Wieland T,Heineke J,Dobreva G
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Nov 04 00:00:00 EDT 2022
Population-wide gene disruption in the murine lung epithelium via AAV-mediated delivery of CRISPR-Cas9 components.
Molecular therapy. Methods & clinical development
Chen H,Durinck S,Patel H,Foreman O,Mesh K,Eastham J,Caothien R,Newman RJ,Roose-Girma M,Darmanis S,Warming S,Lattanzi A,Liang Y,Haley B
12-0311-82 was used in flow cytometry to demonstrate a uniquely effective approach for somatic engineering of the murine lung, which will greatly facilitate the modeling of disease and therapeutic intervention.
Thu Dec 08 00:00:00 EST 2022
Role of quiescent cells in the homeostatic maintenance of the adult submandibular salivary gland.
iScience
Serrano Martinez P,Maimets M,Bron R,van Os R,de Haan G,Pringle S,Coppes RP
12-0311-82 was used in Flow cytometry/Cell sorting to suggest that an active cycling population of cells is responsible for SMG homeostasis with organoid forming potential.
Fri Oct 21 00:00:00 EDT 2022
Acceleration of chronic wound healing by bio-inorganic polyphosphate: In vitro studies and first clinical applications.
Theranostics
Schepler H,Neufurth M,Wang S,She Z,Schröder HC,Wang X,Müller WEG
12-0311-82 was used in Immunohistochemistry (Paraffin) to investigate whether the administration of polyP can substitute for the energy deficiency in chronic wound healing.
Mon Mar 07 00:00:00 EST 2022
Fibroblast activation protein activated antifibrotic peptide delivery attenuates fibrosis in mouse models of liver fibrosis.
Nature communications
Lee J,Byun J,Shim G,Oh YK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Immunohistochemistry
Mon Mar 21 00:00:00 EDT 2022
Fibroblast activation protein activated antifibrotic peptide delivery attenuates fibrosis in mouse models of liver fibrosis.
Nature communications
Lee J,Byun J,Shim G,Oh YK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Immunohistochemistry
Mon Mar 21 00:00:00 EDT 2022
Photoreceptor protection by mesenchymal stem cell transplantation identifies exosomal MiR-21 as a therapeutic for retinal degeneration.
Cell death and differentiation
Deng CL,Hu CB,Ling ST,Zhao N,Bao LH,Zhou F,Xiong YC,Chen T,Sui BD,Yu XR,Hu CH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
Photoreceptor protection by mesenchymal stem cell transplantation identifies exosomal MiR-21 as a therapeutic for retinal degeneration.
Cell death and differentiation
Deng CL,Hu CB,Ling ST,Zhao N,Bao LH,Zhou F,Xiong YC,Chen T,Sui BD,Yu XR,Hu CH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
Anti-adipogenic signals at the onset of obesity-related inflammation in white adipose tissue.
Cellular and molecular life sciences : CMLS
Caputo T,Tran VDT,Bararpour N,Winkler C,Aguileta G,Trang KB,Giordano Attianese GMP,Wilson A,Thomas A,Pagni M,Guex N,Desvergne B,Gilardi F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Anti-adipogenic signals at the onset of obesity-related inflammation in white adipose tissue.
Cellular and molecular life sciences : CMLS
Caputo T,Tran VDT,Bararpour N,Winkler C,Aguileta G,Trang KB,Giordano Attianese GMP,Wilson A,Thomas A,Pagni M,Guex N,Desvergne B,Gilardi F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
SORLA is required for insulin-induced expansion of the adipocyte precursor pool in visceral fat.
The Journal of cell biology
Schmidt V,Horváth C,Dong H,Blüher M,Qvist P,Wolfrum C,Willnow TE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Dec 06 00:00:00 EST 2021
Tendon and motor phenotypes in the mouse model of recessive osteogenesis imperfecta.
eLife
Grol MW,Haelterman NA,Lim J,Munivez EM,Archer M,Hudson DM,Tufa SF,Keene DR,Lei K,Park D,Kuzawa CD,Ambrose CG,Eyre DR,Lee BH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed May 26 00:00:00 EDT 2021
GRHL3 activates FSCN1 to relax cell-cell adhesions between migrating keratinocytes during wound reepithelialization.
JCI insight
Kashgari G,Venkatesh S,Refuerzo S,Pham B,Bayat A,Klein RH,Ramos R,Ta AP,Plikus MV,Wang PH,Andersen B
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed Sep 08 00:00:00 EDT 2021
The IRE1/XBP1 signaling axis promotes skeletal muscle regeneration through a cell non-autonomous mechanism.
eLife
Roy A,Tomaz da Silva M,Bhat R,Bohnert KR,Iwawaki T,Kumar A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Nov 23 00:00:00 EST 2021
Interleukin-11-expressing fibroblasts have a unique gene signature correlated with poor prognosis of colorectal cancer.
Nature communications
Nishina T,Deguchi Y,Ohshima D,Takeda W,Ohtsuka M,Shichino S,Ueha S,Yamazaki S,Kawauchi M,Nakamura E,Nishiyama C,Kojima Y,Adachi-Akahane S,Hasegawa M,Nakayama M,Oshima M,Yagita H,Shibuya K,Mikami T,Inohara N,Matsushima K,Tada N,Nakano H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Apr 16 00:00:00 EDT 2021
Therapeutic Potential of Mesenchymal Stem Cells in a Pre-Clinical Model of Diabetic Kidney Disease and Obesity.
International journal of molecular sciences
Sávio-Silva C,Soinski-Sousa PE,Simplício-Filho A,Bastos RMC,Beyerstedt S,Rangel ÉB
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu Feb 04 00:00:00 EST 2021
Protocols for isolation and characterization of mouse placental hemogenic endothelial cells.
STAR protocols
Liang G,Huang B,Wang F,Liu F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Dec 17 00:00:00 EST 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue May 11 00:00:00 EDT 2021
Loss of α2-6 sialylation promotes the transformation of synovial fibroblasts into a pro-inflammatory phenotype in arthritis.
Nature communications
Wang Y,Khan A,Antonopoulos A,Bouché L,Buckley CD,Filer A,Raza K,Li KP,Tolusso B,Gremese E,Kurowska-Stolarska M,Alivernini S,Dell A,Haslam SM,Pineda MA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Apr 20 00:00:00 EDT 2021
Wip1 regulates the immunomodulatory effects of murine mesenchymal stem cells in type 1 diabetes mellitus via targeting IFN-α/BST2.
Cell death discovery
Zhou N,Liu W,Zhang W,Liu Y,Li X,Wang Y,Zheng R,Zhang Y
12-0311-82 was used in Flow Cytometry to reveal Wip1 affects MSCs immunomodulation by regulating the expression of IFN-α/BST2.
Fri Oct 29 00:00:00 EDT 2021
Mice with induced pulmonary morbidities display severe lung inflammation and mortality following exposure to SARS-CoV-2.
JCI insight
Falach R,Bar-On L,Lazar S,Kadar T,Mazor O,Aftalion M,Gur D,Evgy Y,Shifman O,Aminov T,Israeli O,Cohen-Gihon I,Zaide G,Gutman H,Vagima Y,Makdasi E,Stein D,Rosenfeld R,Alcalay R,Zahavy E,Levy H,Glinert I,Ben-Shmuel A,Israely T,Melamed S,Politi B,Achdout H,Yitzhaki S,Kronman C,Sabo T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Jun 22 00:00:00 EDT 2021
Mice with induced pulmonary morbidities display severe lung inflammation and mortality following exposure to SARS-CoV-2.
JCI insight
Falach R,Bar-On L,Lazar S,Kadar T,Mazor O,Aftalion M,Gur D,Evgy Y,Shifman O,Aminov T,Israeli O,Cohen-Gihon I,Zaide G,Gutman H,Vagima Y,Makdasi E,Stein D,Rosenfeld R,Alcalay R,Zahavy E,Levy H,Glinert I,Ben-Shmuel A,Israely T,Melamed S,Politi B,Achdout H,Yitzhaki S,Kronman C,Sabo T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Jun 22 00:00:00 EDT 2021
Directly recruited GATA6 + peritoneal cavity macrophages contribute to the repair of intestinal serosal injury.
Nature communications
Honda M,Kadohisa M,Yoshii D,Komohara Y,Hibi T
12-0311-82 was used in flow cytometry experiments to show, using spinning-disk confocal microscopy, that large F4/80hiGATA6+ peritoneal cavity macrophages promptly accumulate at damaged intestinal sites upon intestinal thermal injury and upon dextran sodium sulfate induced colitis in mice via a direct route from the peritoneal cavity.
Wed Dec 15 00:00:00 EST 2021
The AP-1 Transcription Factor Fosl-2 Regulates Autophagy in Cardiac Fibroblasts during Myocardial Fibrogenesis.
International journal of molecular sciences
Seidenberg J,Stellato M,Hukara A,Ludewig B,Klingel K,Distler O,Błyszczuk P,Kania G
12-0311-82 was used in Flow Cytometry to demonstrate that Fosl-2 regulates autophagocytosis and the TGF-β-Fosl-2-autophagy axis controls differentiation of cardiac fibroblasts.
Sat Feb 13 00:00:00 EST 2021
The antitumoral effects of chemerin are independent from leukocyte recruitment and mediated by inhibition of neoangiogenesis.
Oncotarget
Dubois-Vedrenne I,Al Delbany D,De Henau O,Robert V,Vernimmen M,Langa F,Lefort A,Libert F,Wittamer V,Parmentier M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Immunocytochemistry
Tue Sep 14 00:00:00 EDT 2021
The ESCRT-0 subcomplex component Hrs/Hgs is a master regulator of myogenesis via modulation of signaling and degradation pathways.
BMC biology
Coudert L,Osseni A,Gangloff YG,Schaeffer L,Leblanc P
12-0311-82 was used in Flow Cytometry to show for the first time that Hrs/Hgs is a master regulator that modulates myogenesis at different levels through the control of trafficking, signaling, and degradation pathways.
Fri Jul 30 00:00:00 EDT 2021
SPARC regulation of PMN clearance protects from pristane-induced lupus and rheumatoid arthritis.
iScience
Sangaletti S,Botti L,Gulino A,Lecis D,Bassani B,Portararo P,Milani M,Cancila V,De Cecco L,Dugo M,Tripodo C,Colombo MP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Jun 25 00:00:00 EDT 2021
Deficiency of Dietary Fiber Modulates Gut Microbiota Composition, Neutrophil Recruitment and Worsens Experimental Colitis.
Frontiers in immunology
Shen S,Prame Kumar K,Wen SW,Shim R,Wanrooy BJ,Stanley D,Moore RJ,Van TTH,Robert R,Hickey MJ,Wong CHY
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Immunohistochemistry
Mon Jul 05 00:00:00 EDT 2021
Deficiency of Dietary Fiber Modulates Gut Microbiota Composition, Neutrophil Recruitment and Worsens Experimental Colitis.
Frontiers in immunology
Shen S,Prame Kumar K,Wen SW,Shim R,Wanrooy BJ,Stanley D,Moore RJ,Van TTH,Robert R,Hickey MJ,Wong CHY
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Immunohistochemistry
Mon Jul 05 00:00:00 EDT 2021
Protocols for endothelial cell isolation from mouse tissues: brain, choroid, lung, and muscle.
STAR protocols
Conchinha NV,Sokol L,Teuwen LA,Veys K,Dumas SJ,Meta E,García-Caballero M,Geldhof V,Chen R,Treps L,Borri M,de Zeeuw P,Falkenberg KD,Dubois C,Parys M,de Rooij LPMH,Rohlenova K,Goveia J,Schoonjans L,Dewerchin M,Eelen G,Li X,Kalucka J,Carmeliet P
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Sep 17 00:00:00 EDT 2021
Actin fence therapy with exogenous V12Rac1 protects against acute lung injury.
JCI insight
Gusarova GA,Das SR,Islam MN,Westphalen K,Jin G,Shmarakov IO,Li L,Bhattacharya S,Bhattacharya J
12-0311-82 was used in Flow Cytometry to propose a potentially new therapeutic paradigm in which actin enhancement by exogenous Rac1 strengthens the alveolar actin fence, protecting against proinflammatory receptor hyperexpression, and therefore blocking ALI.
Mon Mar 22 00:00:00 EDT 2021
Efficient isolation of interstitial fibroblasts directly from mouse kidneys or indirectly after ex vivo expansion.
STAR protocols
Nakai T,Iwamura Y,Suzuki N
12-0311-82 was used in Flow Cytometry to overcome the isolation difficulties of renal interstitial fibroblasts by mildly dissociating mouse kidneys and coculturing fibroblasts with other kidney cells in semisolid medium.
Fri Dec 17 00:00:00 EST 2021
Testosterone metabolites differentially regulate obesogenesis and fat distribution.
Molecular metabolism
Sebo ZL,Rodeheffer MS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Meningeal lymphatic vessels regulate brain tumor drainage and immunity.
Cell research
Hu X,Deng Q,Ma L,Li Q,Chen Y,Liao Y,Zhou F,Zhang C,Shao L,Feng J,He T,Ning W,Kong Y,Huo Y,He A,Liu B,Zhang J,Adams R,He Y,Tang F,Bian X,Luo J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Endothelial Lactate Controls Muscle Regeneration from Ischemia by Inducing M2-like Macrophage Polarization.
Cell metabolism
Zhang J,Muri J,Fitzgerald G,Gorski T,Gianni-Barrera R,Masschelein E,D'Hulst G,Gilardoni P,Turiel G,Fan Z,Wang T,Planque M,Carmeliet P,Pellerin L,Wolfrum C,Fendt SM,Banfi A,Stockmann C,Soro-Arnáiz I,Kopf M,De Bock K
12-0311-83 was used in Flow Cytometry to conclude that ECs exploit glycolysis for angiocrine lactate shuttling to steer muscle regeneration from ischemia.
Tue Jun 02 00:00:00 EDT 2020
STAT3-BDNF-TrkB signalling promotes alveolar epithelial regeneration after lung injury.
Nature cell biology
Paris AJ,Hayer KE,Oved JH,Avgousti DC,Toulmin SA,Zepp JA,Zacharias WJ,Katzen JB,Basil MC,Kremp MM,Slamowitz AR,Jayachandran S,Sivakumar A,Dai N,Wang P,Frank DB,Eisenlohr LC,Cantu E,Beers MF,Weitzman MD,Morrisey EE,Worthen GS
12-0311 was used in Flow cytometry/Cell sorting to elucidate repair mechanisms of proliferation and trandifferentiation of type II alveolar pneumocytes (AT2 cells) into type I alveolar pneumocytes (AT1 cells) after acute lung injury.
Thu Oct 01 00:00:00 EDT 2020
Therapy-Induced Senescence Drives Bone Loss.
Cancer research
Yao Z,Murali B,Ren Q,Luo X,Faget DV,Cole T,Ricci B,Thotala D,Monahan J,van Deursen JM,Baker D,Faccio R,Schwarz JK,Stewart SA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Membrane expression of the estrogen receptor ERα is required for intercellular communications in the mammary epithelium.
Development (Cambridge, England)
Gagniac L,Rusidzé M,Boudou F,Cagnet S,Adlanmerini M,Jeannot P,Gaide N,Giton F,Besson A,Weyl A,Gourdy P,Raymond-Letron I,Arnal JF,Brisken C,Lenfant F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed Mar 11 00:00:00 EDT 2020
Interaction between ICAM1 in endothelial cells and LFA1 in T cells during the pathogenesis of experimental Parkinson's disease.
Experimental and therapeutic medicine
Li W,Chen S,Luo Y,Xia Y,Ma Q,Yao Q,Wu J
12-0311 was used in Flow cytometry/Cell sorting to investigate the infiltration of T cells in lesions in the central nervous system, in patients with and without Parkinson's Disease.
Sat Aug 01 00:00:00 EDT 2020
Defining the role of pulmonary endothelial cell heterogeneity in the response to acute lung injury.
eLife
Niethamer TK,Stabler CT,Leach JP,Zepp JA,Morley MP,Babu A,Zhou S,Morrisey EE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Feb 24 00:00:00 EST 2020
Defining the role of pulmonary endothelial cell heterogeneity in the response to acute lung injury.
eLife
Niethamer TK,Stabler CT,Leach JP,Zepp JA,Morley MP,Babu A,Zhou S,Morrisey EE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Feb 24 00:00:00 EST 2020
Enhancer remodeling promotes tumor-initiating activity in NRF2-activated non-small cell lung cancers.
Nature communications
Okazaki K,Anzawa H,Liu Z,Ota N,Kitamura H,Onodera Y,Alam MM,Matsumaru D,Suzuki T,Katsuoka F,Tadaka S,Motoike I,Watanabe M,Hayasaka K,Sakurada A,Okada Y,Yamamoto M,Suzuki T,Kinoshita K,Sekine H,Motohashi H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Nov 20 00:00:00 EST 2020
Snail promotes the generation of vascular endothelium by breast cancer cells.
Cell death & disease
Chang Z,Zhang Y,Liu J,Zheng Y,Li H,Kong Y,Li P,Peng H,Shi Y,Cao B,Ran F,Chen Y,Song Y,Ye Q,Ding L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Jun 15 00:00:00 EDT 2020
Sierra: discovery of differential transcript usage from polyA-captured single-cell RNA-seq data.
Genome biology
Patrick R,Humphreys DT,Janbandhu V,Oshlack A,Ho JWK,Harvey RP,Lo KK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed Jul 08 00:00:00 EDT 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu Dec 17 00:00:00 EST 2020
Myh11+ microvascular mural cells and derived mesenchymal stem cells promote retinal fibrosis.
Scientific reports
Ray HC,Corliss BA,Bruce AC,Kesting S,Dey P,Mansour J,Seaman SA,Smolko CM,Mathews C,Dey BK,Owens GK,Peirce SM,Yates PA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Sep 25 00:00:00 EDT 2020
Metastasis-initiating cells induce and exploit a fibroblast niche to fuel malignant colonization of the lungs.
Nature communications
Pein M,Insua-Rodríguez J,Hongu T,Riedel A,Meier J,Wiedmann L,Decker K,Essers MAG,Sinn HP,Spaich S,Sütterlin M,Schneeweiss A,Trumpp A,Oskarsson T
12-0311-82 was used in Flow Cytometry to show that disseminated breast cancer cells evoke phenotypic changes in lung fibroblasts, forming a supportive metastatic niche.
Fri Mar 20 00:00:00 EDT 2020
Diaphanous-related formin mDia2 regulates beta2 integrins to control hematopoietic stem and progenitor cell engraftment.
Nature communications
Mei Y,Han X,Liu Y,Yang J,Sumagin R,Ji P
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Immunohistochemistry
Tue Jun 23 00:00:00 EDT 2020
Diaphanous-related formin mDia2 regulates beta2 integrins to control hematopoietic stem and progenitor cell engraftment.
Nature communications
Mei Y,Han X,Liu Y,Yang J,Sumagin R,Ji P
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Immunohistochemistry
Tue Jun 23 00:00:00 EDT 2020
The bone marrow niche components are adversely affected in sepsis.
Molecular biomedicine
Yin F,Qian H,Duan C,Ning B
12-0311-82 was used in Flow Cytometry to indicate that the components of the BM niche underwent series of adaptive changes in sepsis.
Tue Oct 20 00:00:00 EDT 2020
Virus infection is controlled by hematopoietic and stromal cell sensing of murine cytomegalovirus through STING.
eLife
Piersma SJ,Poursine-Laurent J,Yang L,Barber GN,Parikh BA,Yokoyama WM
12-0311 was used in Flow cytometry/Cell sorting to contrast anti-viral effects of MyD88 versus STING in distinct cell types that are infected with murine CMV (MCMV).
Wed Jul 29 00:00:00 EDT 2020
Single-cell analysis reveals T cell infiltration in old neurogenic niches.
Nature
Dulken BW,Buckley MT,Navarro Negredo P,Saligrama N,Cayrol R,Leeman DS,George BM,Boutet SC,Hebestreit K,Pluvinage JV,Wyss-Coray T,Weissman IL,Vogel H,Davis MM,Brunet A
12-0311 was used in Flow cytometry/Cell sorting to reveal an interaction between T cells and neural stem cells in old brains, opening potential avenues through which to counteract age-related decline in brain function.
Mon Jul 01 00:00:00 EDT 2019
Vascular endothelial growth factor pathway promotes osseointegration and CD31hiEMCNhi endothelium expansion in a mouse tibial implant model: an animal study.
The bone & joint journal
Ji G,Xu R,Niu Y,Li N,Ivashkiv L,Bostrom MPG,Greenblatt MB,Yang X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
Vascular endothelial growth factor pathway promotes osseointegration and CD31hiEMCNhi endothelium expansion in a mouse tibial implant model: an animal study.
The bone & joint journal
Ji G,Xu R,Niu Y,Li N,Ivashkiv L,Bostrom MPG,Greenblatt MB,Yang X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Sat Jun 01 00:00:00 EDT 2019
Cell-specific ablation of Hsp47 defines the collagen-producing cells in the injured heart.
JCI insight
Khalil H,Kanisicak O,Vagnozzi RJ,Johansen AK,Maliken BD,Prasad V,Boyer JG,Brody MJ,Schips T,Kilian KK,Correll RN,Kawasaki K,Nagata K,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu Aug 08 00:00:00 EDT 2019
Single-cell expression profiling reveals dynamic flux of cardiac stromal, vascular and immune cells in health and injury.
eLife
Farbehi N,Patrick R,Dorison A,Xaymardan M,Janbandhu V,Wystub-Lis K,Ho JW,Nordon RE,Harvey RP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Mar 26 00:00:00 EDT 2019
PDGFRα+ stromal adipocyte progenitors transition into epithelial cells during lobulo-alveologenesis in the murine mammary gland.
Nature communications
Joshi PA,Waterhouse PD,Kasaian K,Fang H,Gulyaeva O,Sul HS,Boutros PC,Khokha R
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Apr 15 00:00:00 EDT 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
Metastatic cancers promote cachexia through ZIP14 upregulation in skeletal muscle.
Nature medicine
Wang G,Biswas AK,Ma W,Kandpal M,Coker C,Grandgenett PM,Hollingsworth MA,Jain R,Tanji K,Lόpez-Pintado S,Borczuk A,Hebert D,Jenkitkasemwong S,Hojyo S,Davuluri RV,Knutson MD,Fukada T,Acharyya S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Targeting skeletal endothelium to ameliorate bone loss.
Nature medicine
Xu R,Yallowitz A,Qin A,Wu Z,Shin DY,Kim JM,Debnath S,Ji G,Bostrom MP,Yang X,Zhang C,Dong H,Kermani P,Lalani S,Li N,Liu Y,Poulos MG,Wach A,Zhang Y,Inoue K,Di Lorenzo A,Zhao B,Butler JM,Shim JH,Glimcher LH,Greenblatt MB
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Bone marrow niche trafficking of miR-126 controls the self-renewal of leukemia stem cells in chronic myelogenous leukemia.
Nature medicine
Zhang B,Nguyen LXT,Li L,Zhao D,Kumar B,Wu H,Lin A,Pellicano F,Hopcroft L,Su YL,Copland M,Holyoake TL,Kuo CJ,Bhatia R,Snyder DS,Ali H,Stein AS,Brewer C,Wang H,McDonald T,Swiderski P,Troadec E,Chen CC,Dorrance A,Pullarkat V,Yuan YC,Perrotti D,Carlesso N,Forman SJ,Kortylewski M,Kuo YH,Marcucci G
12-0311 was used in Flow cytometry/Cell sorting to demonstrate how bone marrow endothelial cells supply miR-126 to leukemia stem cells in chronic myelogenous leukemia which modulates their quiescence and self-renewal.
Tue May 01 00:00:00 EDT 2018
Loss of pleckstrin-2 reverts lethality and vascular occlusions in JAK2V617F-positive myeloproliferative neoplasms.
The Journal of clinical investigation
Zhao B,Mei Y,Cao L,Zhang J,Sumagin R,Yang J,Gao J,Schipma MJ,Wang Y,Thorsheim C,Zhao L,Stalker T,Stein B,Wen QJ,Crispino JD,Abrams CS,Ji P
12-0311 was used in Immunohistochemistry to identify pleckstrin-2 as an effector of the JAK2/STAT5 pathway and a key factor in the pathogenesis of JAK2V617F-induced myeloproliferative neoplasms.
Tue Jan 02 00:00:00 EST 2018
Nrf2-Mediated Fibroblast Reprogramming Drives Cellular Senescence by Targeting the Matrisome.
Developmental cell
Hiebert P,Wietecha MS,Cangkrama M,Haertel E,Mavrogonatou E,Stumpe M,Steenbock H,Grossi S,Beer HD,Angel P,Brinckmann J,Kletsas D,Dengjel J,Werner S
12-0311 was used in Flow cytometry/Cell sorting to demonstrate cellular senescence can be driven by Nrf2 activation in fibroblasts.
Mon Jul 16 00:00:00 EDT 2018
Indian Hedgehog Suppresses a Stromal Cell-Driven Intestinal Immune Response.
Cellular and molecular gastroenterology and hepatology
Westendorp BF,Büller NVJA,Karpus ON,van Dop WA,Koster J,Versteeg R,Koelink PJ,Snel CY,Meisner S,Roelofs JJTH,Uhmann A,Ver Loren van Themaat E,Heijmans J,Hahn H,Muncan V,Wildenberg ME,van den Brink GR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu Mar 31 00:00:00 EDT 2022
Invariant Natural Killer T Cells Shape the Gut Microbiota and Regulate Neutrophil Recruitment and Function During Intestinal Inflammation.
Frontiers in immunology
Shen S,Prame Kumar K,Stanley D,Moore RJ,Van TTH,Wen SW,Hickey MJ,Wong CHY
12-0311 was used in intravital imaging to study the role of iNKT cells, neutrophil and gut microbiota in regulating colitis pathology.
Thu Jul 11 00:00:00 EDT 2019
Wnt inhibition promotes vascular specification of embryonic cardiac progenitors.
Development (Cambridge, England)
Reichman DE,Park L,Man L,Redmond D,Chao K,Harvey RP,Taketo MM,Rosenwaks Z,James D
12-0311 was used in Flow cytometry/Cell sorting to investigate the role of Wnt signalling in the specification of cardiac progenitor cells toward downstream fates.
Mon Jan 08 00:00:00 EST 2018
AHNAK Loss in Mice Promotes Type II Pneumocyte Hyperplasia and Lung Tumor Development.
Molecular cancer research : MCR
Park JW,Kim IY,Choi JW,Lim HJ,Shin JH,Kim YN,Lee SH,Son Y,Sohn M,Woo JK,Jeong JH,Lee C,Bae YS,Seong JK
12-0311 was used in Flow cytometry/Cell sorting to suggest that AHNAK functions as a novel tumour suppressor in lung cancer by suppressing alveolar epithelial cell proliferation.
Wed Aug 01 00:00:00 EDT 2018
Adipocyte hypertrophy and lipid dynamics underlie mammary gland remodeling after lactation.
Nature communications
Zwick RK,Rudolph MC,Shook BA,Holtrup B,Roth E,Lei V,Van Keymeulen A,Seewaldt V,Kwei S,Wysolmerski J,Rodeheffer MS,Horsley V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Sep 04 00:00:00 EDT 2018
Stem cell factor is selectively secreted by arterial endothelial cells in bone marrow.
Nature communications
Xu C,Gao X,Wei Q,Nakahara F,Zimmerman SE,Mar J,Frenette PS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Jun 22 00:00:00 EDT 2018
Age-specific biological and molecular profiling distinguishes paediatric from adult acute myeloid leukaemias.
Nature communications
Chaudhury S,O'Connor C,Cañete A,Bittencourt-Silvestre J,Sarrou E,Prendergast Á,Choi J,Johnston P,Wells CA,Gibson B,Keeshan K
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Dec 11 00:00:00 EST 2018
Lsd1 regulates skeletal muscle regeneration and directs the fate of satellite cells.
Nature communications
Tosic M,Allen A,Willmann D,Lepper C,Kim J,Duteil D,Schüle R
12-0311 was used in Flow cytometry/Cell sorting to show that elevated levels of lysine-specific demethylase 1 have a beneficial effect on muscle regeneration and recovery after injury.
Thu Jan 25 00:00:00 EST 2018
AMPK Activation Regulates LTBP4-Dependent TGF-β1 Secretion by Pro-inflammatory Macrophages and Controls Fibrosis in Duchenne Muscular Dystrophy.
Cell reports
Juban G,Saclier M,Yacoub-Youssef H,Kernou A,Arnold L,Boisson C,Ben Larbi S,Magnan M,Cuvellier S,Théret M,Petrof BJ,Desguerre I,Gondin J,Mounier R,Chazaud B
12-0311 was used in Flow cytometry/Cell sorting to conclude that an AMPK-LTBP4 axis in inflammatory macrophages controls the production of TGF-β1, which is further activated by and acts on fibroblastic cells, leading to fibrosis in DMD.
Tue Nov 20 00:00:00 EST 2018
A multi-cellular 3D bioprinting approach for vascularized heart tissue engineering based on HUVECs and iPSC-derived cardiomyocytes.
Scientific reports
Maiullari F,Costantini M,Milan M,Pace V,Chirivì M,Maiullari S,Rainer A,Baci D,Marei HE,Seliktar D,Gargioli C,Bearzi C,Rizzi R
12-0311 was used in Flow cytometry/Cell sorting to develop a novel approach to creating advanced model of cardiac tissue, using heterogeneous, multi-cellular constructs composed of Human Umbilical Vein Endothelial Cells and induced pluripotent cell-derived cardiomyocytes.
Mon Sep 10 00:00:00 EDT 2018
Generation of Vascular Endothelial Cells and Hematopoietic Cells by Blastocyst Complementation.
Stem cell reports
Hamanaka S,Umino A,Sato H,Hayama T,Yanagida A,Mizuno N,Kobayashi T,Kasai M,Suchy FP,Yamazaki S,Masaki H,Yamaguchi T,Nakauchi H
Tue Oct 09 00:00:00 EDT 2018
The ICAM-1 expression level determines the susceptibility of human endothelial cells to simulated microgravity.
Journal of cellular biochemistry
Buravkova LB,Rudimov EG,Andreeva ER,Grigoriev AI
12-0311-82 was used in Flow cytometry/Cell sorting to study the expression profile of the multi-step cascade of adhesion molecules in umbilical cord endothelial cells.
Thu Mar 01 00:00:00 EST 2018
Tumour ischaemia by interferon-γ resembles physiological blood vessel regression.
Nature
Kammertoens T,Friese C,Arina A,Idel C,Briesemeister D,Rothe M,Ivanov A,Szymborska A,Patone G,Kunz S,Sommermeyer D,Engels B,Leisegang M,Textor A,Fehling HJ,Fruttiger M,Lohoff M,Herrmann A,Yu H,Weichselbaum R,Uckert W,Hübner N,Gerhardt H,Beule D,Schreiber H,Blankenstein T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu May 04 00:00:00 EDT 2017
Increased Vascular Permeability in the Bone Marrow Microenvironment Contributes to Disease Progression and Drug Response in Acute Myeloid Leukemia.
Cancer cell
Passaro D,Di Tullio A,Abarrategi A,Rouault-Pierre K,Foster K,Ariza-McNaughton L,Montaner B,Chakravarty P,Bhaw L,Diana G,Lassailly F,Gribben J,Bonnet D
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Sep 11 00:00:00 EDT 2017
Preexisting endothelial cells mediate cardiac neovascularization after injury.
The Journal of clinical investigation
He L,Huang X,Kanisicak O,Li Y,Wang Y,Li Y,Pu W,Liu Q,Zhang H,Tian X,Zhao H,Liu X,Zhang S,Nie Y,Hu S,Miao X,Wang QD,Wang F,Chen T,Xu Q,Lui KO,Molkentin JD,Zhou B
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
FXYD5 Is an Essential Mediator of the Inflammatory Response during Lung Injury.
Frontiers in immunology
Brazee PL,Soni PN,Tokhtaeva E,Magnani N,Yemelyanov A,Perlman HR,Ridge KM,Sznajder JI,Vagin O,Dada LA
12-0311 was used in Flow cytometry/Cell sorting to confirm that FXYD5 induces a range of pro-inflammatory effects.
Fri Mar 18 00:00:00 EDT 2022
Cigarette Smoke Increases Endothelial CXCL16-Leukocyte CXCR6 Adhesion In Vitro and In Vivo. Potential Consequences in Chronic Obstructive Pulmonary Disease.
Frontiers in immunology
Marques P,Collado A,Escudero P,Rius C,González C,Servera E,Piqueras L,Sanz MJ
Wed Nov 20 00:00:00 EST 2019
Obesity reversibly depletes the basal cell population and enhances mammary epithelial cell estrogen receptor alpha expression and progenitor activity.
Breast cancer research : BCR
Chamberlin T,D'Amato JV,Arendt LM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed Nov 29 00:00:00 EST 2017
Mesenchymal Stem Cell Therapy Protects Lungs from Radiation-Induced Endothelial Cell Loss by Restoring Superoxide Dismutase 1 Expression.
Antioxidants & redox signaling
Klein D,Steens J,Wiesemann A,Schulz F,Kaschani F,Röck K,Yamaguchi M,Wirsdörfer F,Kaiser M,Fischer JW,Stuschke M,Jendrossek V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Apr 10 00:00:00 EDT 2017
Estrogen modulates mesenchyme-epidermis interactions in the adult nipple.
Development (Cambridge, England)
Wu HJ,Oh JW,Spandau DF,Tholpady S,Diaz J,Schroeder LJ,Offutt CD,Glick AB,Plikus MV,Koyama S,Foley J
12-0311 was used in Flow cytometry/Cell sorting to show that specialized nipple epidermis is maintained by estrogen-induced repression of TGFβ signalling in the local fibroblasts.
Sat Apr 15 00:00:00 EDT 2017
Cornea lymphatics drive the CD8+ T-cell response to herpes simplex virus-1.
Immunology and cell biology
Gurung HR,Carr MM,Carr DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2017
β3-Adrenergic Regulation of EPC Features Through Manipulation of the Bone Marrow MSC Niche.
Journal of cellular biochemistry
Vafaei R,Nassiri SM,Siavashi V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
Single-Cell Transcriptomic Analysis Defines Heterogeneity and Transcriptional Dynamics in the Adult Neural Stem Cell Lineage.
Cell reports
Dulken BW,Leeman DS,Boutet SC,Hebestreit K,Brunet A
12-0311 was used in Flow cytometry/Cell sorting to characterize adult neural stem cell populations and examine the molecular identities and heterogeneity of in vivo neural stem cell populations.
Tue Jan 17 00:00:00 EST 2017
Dynamic Patterns of Clonal Evolution in Tumor Vasculature Underlie Alterations in Lymphocyte-Endothelial Recognition to Foster Tumor Immune Escape.
Cancer research
Corey DM,Rinkevich Y,Weissman IL
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Mar 15 00:00:00 EDT 2016
Endothelial cell survival during angiogenesis requires the pro-survival protein MCL1.
Cell death and differentiation
Watson EC,Whitehead L,Adams RH,Dewson G,Coultas L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Platelets Guide Leukocytes to Their Sites of Extravasation.
PLoS biology
Zuchtriegel G,Uhl B,Puhr-Westerheide D,Pörnbacher M,Lauber K,Krombach F,Reichel CA
12-0311 was used in Flow cytometry/Cell sorting to uncover a platelet-directed, spatiotemporally organised, multicellular crosstalk that is essential for effective trafficking of leukocytes to the site of inflammation.
Sun May 01 00:00:00 EDT 2016
A New Method to Stabilize C-Kit Expression in Reparative Cardiac Mesenchymal Cells.
Frontiers in cell and developmental biology
Wysoczynski M,Dassanayaka S,Zafir A,Ghafghazi S,Long BW,Noble C,DeMartino AM,Brittian KR,Bolli R,Jones SP
12-0311 was used in Flow cytometry/Cell sorting to investigate a new method for c-kit expressing cell isolation and stabilisation in reparative cardiac mesenchymal cells.
Thu Aug 18 00:00:00 EDT 2016
A cell-autonomous tumour suppressor role of RAF1 in hepatocarcinogenesis.
Nature communications
Jeric I,Maurer G,Cavallo AL,Raguz J,Desideri E,Tarkowski B,Parrini M,Fischer I,Zatloukal K,Baccarini M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed Dec 21 00:00:00 EST 2016
Endothelial to mesenchymal transition is common in atherosclerotic lesions and is associated with plaque instability.
Nature communications
Evrard SM,Lecce L,Michelis KC,Nomura-Kitabayashi A,Pandey G,Purushothaman KR,d'Escamard V,Li JR,Hadri L,Fujitani K,Moreno PR,Benard L,Rimmele P,Cohain A,Mecham B,Randolph GJ,Nabel EG,Hajjar R,Fuster V,Boehm M,Kovacic JC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Jun 24 00:00:00 EDT 2016
Genetic lineage tracing defines myofibroblast origin and function in the injured heart.
Nature communications
Kanisicak O,Khalil H,Ivey MJ,Karch J,Maliken BD,Correll RN,Brody MJ,J Lin SC,Aronow BJ,Tallquist MD,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Fri Jul 22 00:00:00 EDT 2016
BMP and Hedgehog Regulate Distinct AGM Hematopoietic Stem Cells Ex Vivo.
Stem cell reports
Crisan M,Solaimani Kartalaei P,Neagu A,Karkanpouna S,Yamada-Inagawa T,Purini C,Vink CS,van der Linden R,van Ijcken W,Chuva de Sousa Lopes SM,Monteiro R,Mummery C,Dzierzak E
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Mar 08 00:00:00 EST 2016
Long-Term In Vitro Expansion of Salivary Gland Stem Cells Driven by Wnt Signals.
Stem cell reports
Maimets M,Rocchi C,Bron R,Pringle S,Kuipers J,Giepmans BN,Vries RG,Clevers H,de Haan G,van Os R,Coppes RP
12-0311 was used in Flow cytometry/Cell sorting to identify Wnt signalling as a key driver of self-renewal in adult SGSCs that allows extensive in vitro expansion and rescue of SG function in irradiated mice upon transplantation.
Tue Jan 12 00:00:00 EST 2016
Neural innervation stimulates splenic TFF2 to arrest myeloid cell expansion and cancer.
Nature communications
Dubeykovskaya Z,Si Y,Chen X,Worthley DL,Renz BW,Urbanska AM,Hayakawa Y,Xu T,Westphalen CB,Dubeykovskiy A,Chen D,Friedman RA,Asfaha S,Nagar K,Tailor Y,Muthupalani S,Fox JG,Kitajewski J,Wang TC
12-0311 was used in Flow cytometry/Cell sorting to show that TFF2 is released by vagally modulated memory T cells to suppress the expansion of myeloid-derived suppressor cells through CXCR4.
Thu Feb 04 00:00:00 EST 2016
The Lymphatic Endothelial mCLCA1 Antibody Induces Proliferation and Growth of Lymph Node Lymphatic Sinuses.
PloS one
Jordan-Williams KL,Ramanujam N,Farr AG,Ruddell A
12-0311 was used in Flow cytometry/Cell sorting to investigate the induction of proliferation and growth of lymph node lymphatic sinuses.
Mon Jul 10 00:00:00 EDT 2017
Self-renewing diploid Axin2(+) cells fuel homeostatic renewal of the liver.
Nature
Wang B,Zhao L,Fish M,Logan CY,Nusse R
Thu Aug 13 00:00:00 EDT 2015
Lineage conversion induced by pluripotency factors involves transient passage through an iPSC stage.
Nature biotechnology
Bar-Nur O,Verheul C,Sommer AG,Brumbaugh J,Schwarz BA,Lipchina I,Huebner AJ,Mostoslavsky G,Hochedlinger K
12-0311 was used in Immunocytochemistry to elucidate the conditions necessary for the generation of induced neural stem cells from fibroblasts using OKSM factors.
Wed Jul 01 00:00:00 EDT 2015
Host microbiota constantly control maturation and function of microglia in the CNS.
Nature neuroscience
Erny D,Hrabě de Angelis AL,Jaitin D,Wieghofer P,Staszewski O,David E,Keren-Shaul H,Mahlakoiv T,Jakobshagen K,Buch T,Schwierzeck V,Utermöhlen O,Chun E,Garrett WS,McCoy KD,Diefenbach A,Staeheli P,Stecher B,Amit I,Prinz M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
The C-terminal CGHC motif of protein disulfide isomerase supports thrombosis.
The Journal of clinical investigation
Zhou J,Wu Y,Wang L,Rauova L,Hayes VM,Poncz M,Essex DW
12-0311 was used in Flow cytometry/Cell sorting to investigate the contribution of protein disulfide isomerase CGHC sites during physiological reaction such as thrombosis.
Tue Nov 03 00:00:00 EST 2015
Imaging the dynamic platelet-neutrophil response in sterile liver injury and repair in mice.
Hepatology (Baltimore, Md.)
Slaba I,Wang J,Kolaczkowska E,McDonald B,Lee WY,Kubes P
12-0311 was used in flow-cytomery to study the role of platelets in sterile liver injury and repair.
Sun Nov 01 00:00:00 EDT 2015
Vascular endothelial growth factor c/vascular endothelial growth factor receptor 3 signaling regulates chemokine gradients and lymphocyte migration from tissues to lymphatics.
Transplantation
Iwami D,Brinkman CC,Bromberg JS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Conditional immortalization of primary adipocyte precursor cells.
Adipocyte
Church C,Brown M,Rodeheffer MS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Aug 10 00:00:00 EDT 2015
Targeted Gene Correction in Osteopetrotic-Induced Pluripotent Stem Cells for the Generation of Functional Osteoclasts.
Stem cell reports
Neri T,Muggeo S,Paulis M,Caldana ME,Crisafulli L,Strina D,Focarelli ML,Faggioli F,Recordati C,Scaramuzza S,Scanziani E,Mantero S,Buracchi C,Sobacchi C,Lombardo A,Naldini L,Vezzoni P,Villa A,Ficara F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Oct 13 00:00:00 EDT 2015
Platelet aggregation but not activation and degranulation during the acute post-ischemic reperfusion phase in livers with no underlying disease.
Journal of clinical and translational research
van Golen RF,Stevens KM,Colarusso P,Jaeschke H,Heger M
12-0311 was used in Immunofluorescence to understand platelet function during hepatic ischaemia/reperfusion injury.
Thu Mar 31 00:00:00 EDT 2022
O-GlcNAcylation Negatively Regulates Cardiomyogenic Fate in Adult Mouse Cardiac Mesenchymal Stromal Cells.
PloS one
Zafir A,Bradley JA,Long BW,Muthusamy S,Li Q,Hill BG,Wysoczynski M,Prabhu SD,Bhatnagar A,Bolli R,Jones SP
12-0311 was used in Flow cytometry/Cell sorting to investigate how O-GlcNAcylation of intracellular proteins is involved in the regulation of mesenchymal stromal cell lineage commitment.
Wed Jun 29 00:00:00 EDT 2016
Rejuvenation of the muscle stem cell population restores strength to injured aged muscles.
Nature medicine
Cosgrove BD,Gilbert PM,Porpiglia E,Mourkioti F,Lee SP,Corbel SY,Llewellyn ME,Delp SL,Blau HM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Sat Mar 01 00:00:00 EST 2014
c-kit+ cells minimally contribute cardiomyocytes to the heart.
Nature
van Berlo JH,Kanisicak O,Maillet M,Vagnozzi RJ,Karch J,Lin SC,Middleton RC,Marbán E,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu May 15 00:00:00 EDT 2014
Mesenchymal-endothelial transition contributes to cardiac neovascularization.
Nature
Ubil E,Duan J,Pillai IC,Rosa-Garrido M,Wu Y,Bargiacchi F,Lu Y,Stanbouly S,Huang J,Rojas M,Vondriska TM,Stefani E,Deb A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu Oct 30 00:00:00 EDT 2014
Autophagy regulates the therapeutic potential of mesenchymal stem cells in experimental autoimmune encephalomyelitis.
Autophagy
Dang S,Xu H,Xu C,Cai W,Li Q,Cheng Y,Jin M,Wang RX,Peng Y,Zhang Y,Wu C,He X,Wan B,Zhang Y
12-0311 was used in Flow cytometry/Cell sorting to study how inflammatory microenvironment-induced autophagy can downregulate the immunosuppressive function of mesenchymal stem cells.
Tue Jul 01 00:00:00 EDT 2014
Cell-autonomous heparanase modulates self-renewal and migration in bone marrow-derived mesenchymal stem cells.
Journal of biomedical science
Cheng CC,Lee YH,Lin SP,Huangfu WC,Liu IH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu Mar 13 00:00:00 EDT 2014
Blocking junctional adhesion molecule C enhances dendritic cell migration and boosts the immune responses against Leishmania major.
PLoS pathogens
Ballet R,Emre Y,Jemelin S,Charmoy M,Tacchini-Cottier F,Imhof BA
12-0311 was used in Flow cytometry/Cell sorting to illustrate a key role for the vascular adhesion molecule JAM-C in controlling leukocyte migration and subsequent immune events in response to pathogen infections.
Mon Dec 01 00:00:00 EST 2014
Characterization of Cre recombinase activity for in vivo targeting of adipocyte precursor cells.
Stem cell reports
Krueger KC,Costa MJ,Du H,Feldman BJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Tue Dec 09 00:00:00 EST 2014
Dual modulation of the mitochondrial permeability transition pore and redox signaling synergistically promotes cardiomyocyte differentiation from pluripotent stem cells.
Journal of the American Heart Association
Cho SW,Park JS,Heo HJ,Park SW,Song S,Kim I,Han YM,Yamashita JK,Youm JB,Han J,Koh GY
12-0311 was used in Flow cytometry/Cell sorting to show that mPTP inhibition by CsA alters mitochondrial oxidative metabolism and redox signaling.
Thu Mar 13 00:00:00 EDT 2014
Dual modulation of the mitochondrial permeability transition pore and redox signaling synergistically promotes cardiomyocyte differentiation from pluripotent stem cells.
Journal of the American Heart Association
Cho SW,Park JS,Heo HJ,Park SW,Song S,Kim I,Han YM,Yamashita JK,Youm JB,Han J,Koh GY
12-0311 was used in Flow cytometry/Cell sorting to show that mPTP inhibition by CsA alters mitochondrial oxidative metabolism and redox signaling.
Thu Mar 13 00:00:00 EDT 2014
Self-recognition of the endothelium enables regulatory T-cell trafficking and defines the kinetics of immune regulation.
Nature communications
Fu H,Kishore M,Gittens B,Wang G,Coe D,Komarowska I,Infante E,Ridley AJ,Cooper D,Perretti M,Marelli-Berg FM
12-0311 was used in Flow cytometry/Cell sorting to explain the mechanisms underlying the powerful migratory capacity of Treg cells and their ability to maintain optimal T cell ratios.
Fri Mar 14 00:00:00 EDT 2014
Anti-remodeling and anti-fibrotic effects of the neuregulin-1β glial growth factor 2 in a large animal model of heart failure.
Journal of the American Heart Association
Galindo CL,Kasasbeh E,Murphy A,Ryzhov S,Lenihan S,Ahmad FA,Williams P,Nunnally A,Adcock J,Song Y,Harrell FE,Tran TL,Parry TJ,Iaci J,Ganguly A,Feoktistov I,Stephenson MK,Caggiano AO,Sawyer DB,Cleator JH
12-0311 was used in Flow cytometry/Cell sorting to suggest that GGF2/NRG-1b prevents adverse remodeling after injury in part via anti-fibrotic effects in the heart.
Thu Oct 23 00:00:00 EDT 2014
miR-126 and miR-126* repress recruitment of mesenchymal stem cells and inflammatory monocytes to inhibit breast cancer metastasis.
Nature cell biology
Zhang Y,Yang P,Sun T,Li D,Xu X,Rui Y,Li C,Chong M,Ibrahim T,Mercatali L,Amadori D,Lu X,Xie D,Li QJ,Wang XF
12-0311 was used in Flow cytometry/Cell sorting to investigate the role of miR-126 and miR-126* in tumour progression.
Fri Mar 01 00:00:00 EST 2013
The expression of Sox17 identifies and regulates haemogenic endothelium.
Nature cell biology
Clarke RL,Yzaguirre AD,Yashiro-Ohtani Y,Bondue A,Blanpain C,Pear WS,Speck NA,Keller G
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2013
VEGF-A regulated by progesterone governs uterine angiogenesis and vascular remodelling during pregnancy.
EMBO molecular medicine
Kim M,Park HJ,Seol JW,Jang JY,Cho YS,Kim KR,Choi Y,Lydon JP,Demayo FJ,Shibuya M,Ferrara N,Sung HK,Nagy A,Alitalo K,Koh GY
12-0311 was used in Immunohistochemistry to investigate the role of two vascular growth factor receptors in the regulation of angiogenesis in rapidly growing and regressing uteri.
Sun Sep 01 00:00:00 EDT 2013
Isolation, characterization, and transplantation of cardiac endothelial cells.
BioMed research international
Pratumvinit B,Reesukumal K,Janebodin K,Ieronimakis N,Reyes M
12-0311 was used in Immunofluorescence to develop a method to expand the study of endothelial cell biology for multiple clinical applications..
Tue Jun 17 00:00:00 EDT 2014
Etv2 rescues Flk1 mutant embryoid bodies.
Genesis (New York, N.Y. : 2000)
Rasmussen TL,Martin CM,Walter CA,Shi X,Perlingeiro R,Koyano-Nakagawa N,Garry DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2013
Mouse models and techniques for the isolation of the diabetic endothelium.
ISRN endocrinology
Darrow AL,Maresh JG,Shohet RV
12-0311 was used in Flow cytometry/Cell sorting to validate mouse models and endothelial isolation techniques for assessing diabetic endothelial dysfunction and vascular responses in vivo.
Wed Jul 10 00:00:00 EDT 2013
Endothelial cells provide an instructive niche for the differentiation and functional polarization of M2-like macrophages.
Blood
He H,Xu J,Warren CM,Duan D,Li X,Wu L,Iruela-Arispe ML
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu Oct 11 00:00:00 EDT 2012
ROCK suppression promotes differentiation and expansion of endothelial cells from embryonic stem cell-derived Flk1(+) mesodermal precursor cells.
Blood
Joo HJ,Choi DK,Lim JS,Park JS,Lee SH,Song S,Shin JH,Lim DS,Kim I,Hwang KC,Koh GY
12-0311 was used in Flow cytometry/Cell sorting to demonstrate a versatile method for obtaining an ample amount of ESC-derived ECs for use in therapeutic neovascularisation.
Thu Sep 27 00:00:00 EDT 2012
Biological and clinical availability of adipose-derived stem cells for pelvic dead space repair.
Stem cells translational medicine
Takahashi H,Haraguchi N,Nishikawa S,Miyazaki S,Suzuki Y,Mizushima T,Nishimura J,Takemasa I,Yamamoto H,Mimori K,Ishii H,Doki Y,Mori M
12-0311 was used in Flow cytometry/Cell sorting to clarify the utility of adipose-derived stem cells and resected intraperitoneal fat pads for pelvic cavity repair using a mouse model.
Thu Nov 01 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
The endothelial antigen ESAM monitors hematopoietic stem cell status between quiescence and self-renewal.
Journal of immunology (Baltimore, Md. : 1950)
Sudo T,Yokota T,Oritani K,Satoh Y,Sugiyama T,Ishida T,Shibayama H,Ezoe S,Fujita N,Tanaka H,Maeda T,Nagasawa T,Kanakura Y
12-0311 was used in Flow cytometry/Cell sorting to demonstrate that the intensity of ESAM expression is useful to trace activated HSCs.
Sun Jul 01 00:00:00 EDT 2012
Role of A2B adenosine receptors in regulation of paracrine functions of stem cell antigen 1-positive cardiac stromal cells.
The Journal of pharmacology and experimental therapeutics
Ryzhov S,Goldstein AE,Novitskiy SV,Blackburn MR,Biaggioni I,Feoktistov I
12-0311-82 was used in Flow Cytometry to investigate whether the stimulation of adenosine receptors on cardiac Sca-1(+) cells up-regulates their secretion of proangiogenic factors.
Fri Jun 01 00:00:00 EDT 2012
A model of DENV-3 infection that recapitulates severe disease and highlights the importance of IFN-γ in host resistance to infection.
PLoS neglected tropical diseases
Costa VV,Fagundes CT,Valadão DF,Cisalpino D,Dias AC,Silveira KD,Kangussu LM,Ávila TV,Bonfim MR,Bonaventura D,Silva TA,Sousa LP,Rachid MA,Vieira LQ,Menezes GB,de Paula AM,Atrasheuskaya A,Ignatyev G,Teixeira MM,Souza DG
12-0311-82 was used in Immunohistochemistry to develop an animal model of dengue virus (DENV-3) infection in immunocompetent mice.
Thu Aug 30 00:00:00 EDT 2012
Reprogramming adult dermis to a neonatal state through epidermal activation of β-catenin.
Development (Cambridge, England)
Collins CA,Kretzschmar K,Watt FM
12-0311 was used in Flow cytometry/Cell sorting to display the plasticity of the characteristics of the dermis in response to cues from the epidermis.
Thu Dec 01 00:00:00 EST 2011
Differentiation of an embryonic stem cell to hemogenic endothelium by defined factors: essential role of bone morphogenetic protein 4.
Development (Cambridge, England)
Chiang PM,Wong PC
12-0311 was used in Immunohistochemistry to identify a set of factors that are necessary and sufficient to differentiate ES cells into definitive hematopoietic precursors.
Fri Jul 01 00:00:00 EDT 2011
Experimental myocardial infarction triggers canonical Wnt signaling and endothelial-to-mesenchymal transition.
Disease models & mechanisms
Aisagbonhi O,Rai M,Ryzhov S,Atria N,Feoktistov I,Hatzopoulos AK
12-0311 was used in Flow cytometry/Cell sorting to demonstrates that canonical Wnt activation and EndMT are molecular and cellular responses to myocardial infarction (MI).
Fri Jul 01 00:00:00 EDT 2011
Intramuscular VEGF activates an SDF1-dependent progenitor cell cascade and an SDF1-independent muscle paracrine cascade for cardiac repair.
American journal of physiology. Heart and circulatory physiology
Zisa D,Shabbir A,Mastri M,Taylor T,Aleksic I,McDaniel M,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2011
Transcriptional analysis of the endothelial response to diabetes reveals a role for galectin-3.
Physiological genomics
Darrow AL,Shohet RV,Maresh JG
12-0311 was used in Flow cytometry/Cell sorting to survey transcriptional responses in the vascular endothelia of mice receiving a diabetogenic, high-fat diet.
Thu Oct 20 00:00:00 EDT 2011
Angiogenesis gene expression in murine endothelial cells during post-pneumonectomy lung growth.
Respiratory research
Lin M,Chamoto K,Gibney BC,Lee GS,Collings-Simpson D,Houdek J,Konerding MA,Tsuda A,Mentzer SJ
12-0311 was used in Flow cytometry/Cell sorting to investigate the genes involved in an experimental model of alveolar capillary angiogenesis, identifying 22 genes expressed in CD31+ cells with a significantly increased transcription.
Wed Jul 27 00:00:00 EDT 2011
Antigen-specific immunity and cross-priming by epithelial ovarian carcinoma-induced CD11b(+)Gr-1(+) cells.
Journal of immunology (Baltimore, Md. : 1950)
Tomihara K,Guo M,Shin T,Sun X,Ludwig SM,Brumlik MJ,Zhang B,Curiel TJ,Shin T
12-0311 was used in Flow cytometry/Cell sorting to investigate CD11b(+)Gr-1(+) cells found in epithelial ovarian cancer-bearing mice, which are induced by ovarian cancer cells and undergo antigen-specific immunity and cross-priming.
Tue Jun 01 00:00:00 EDT 2010
The endothelial antigen ESAM marks primitive hematopoietic progenitors throughout life in mice.
Blood
Yokota T,Oritani K,Butz S,Kokame K,Kincade PW,Miyata T,Vestweber D,Kanakura Y
12-0311 was used in Flow cytometry/Cell sorting to study hematopoietic stem cell (HSC) characteristics for improved understanding of their development and stage-related functions, showing that the endothelial antigen ESAM marks primitive hematopoietic progenitors throughout life in mice.
Thu Mar 26 00:00:00 EDT 2009
Auf1/Hnrnpd-deficient mice develop pruritic inflammatory skin disease.
The Journal of investigative dermatology
Sadri N,Schneider RJ
Sun Mar 01 00:00:00 EST 2009
Heart failure therapy mediated by the trophic activities of bone marrow mesenchymal stem cells: a noninvasive therapeutic regimen.
American journal of physiology. Heart and circulatory physiology
Shabbir A,Zisa D,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2009
Intramuscular VEGF repairs the failing heart: role of host-derived growth factors and mobilization of progenitor cells.
American journal of physiology. Regulatory, integrative and comparative physiology
Zisa D,Shabbir A,Mastri M,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 12-0311-82) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2009
Deficient ghrelin receptor-mediated signaling compromises thymic stromal cell microenvironment by accelerating thymic adiposity.
The Journal of biological chemistry
Youm YH,Yang H,Sun Y,Smith RG,Manley NR,Vandanmagsar B,Dixit VD
12-0311 was used in Immunohistochemistry on frozen tissues to study the expression of Ghrl and ghrelin receptor in thymic stromal cells during physiological ageing.
Fri Mar 13 00:00:00 EDT 2009
Analysis of the IKKbeta/NF-kappaB signaling pathway during embryonic angiogenesis.
Developmental dynamics : an official publication of the American Association of Anatomists
Hou Y,Li F,Karin M,Ostrowski MC
12-0311 was used in Flow cytometry/Cell sorting to describe the IKKβ/NF-κB signalling pathway involved in embryonic angiogenesis, which Ikkβ locus disruption in foetal liver vasculature revealed to be responsible for regulation of hepatocyte proliferation and apoptosis, however not for hematopoiesis.
Wed Oct 01 00:00:00 EDT 2008