Search Thermo Fisher Scientific
This antibody recognizes two isoforms of human CD32, Fc gammaRIIA and Fc gammaRIIB.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.