Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: This 9E8 monoclonal antibody reacts with human CD367, also known as dendritic cell immunoreceptor (DCIR) or Clec4A. This C-type lectin is expressed on monocytes, granulocytes, B cells, and plasmacytoid dendritic cells in peripheral blood. DCIR is also expressed on mature and immature monocyte-derived dendritic cells in vitro. DCIR is not expressed on NK or T cells. With its cytoplasmic ITIM domain, DCIR is an inhibitory receptor that recruits the SHP-1 and SHP-2 phosphatases upon ligand binding. Studies have demonstrated that DCIR plays a role in the interaction between dendritic cells and HIV-1.
Applications Reported: This 9E8 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This 9E8 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in inflammatory and immune response. Multiple transcript variants encoding distinct isoforms have been identified for this gene. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 6; C-type lectin DDB27; C-type lectin domain family 4 member A; C-type lectin domain family 4, member A; C-type lectin superfamily member 6; CD367; Dendritic cell immunoreceptor; Lectin-like immunoreceptor
Gene Aliases: CD367; CLEC4A; CLECSF6; DCIR; DDB27; HDCGC13P; LLIR
UniProt ID: (Human) Q9UMR7
Entrez Gene ID: (Human) 50856
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support