Product References
Impact of broad-spectrum antibiotics on the gut-microbiota-spleen-brain axis.
Brain, behavior, & immunity - health
Wan X,Eguchi A,Sakamoto A,Fujita Y,Yang Y,Qu Y,Hatano M,Mori C,Hashimoto K
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2023
Deletion of CD36 exhibits limited impact on normal hematopoiesis and the leukemia microenvironment.
Cellular & molecular biology letters
Meng Y,Pospiech M,Ali A,Chandwani R,Vergel M,Onyemaechi S,Yaghmour G,Lu R,Alachkar H
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed May 24 00:00:00 EDT 2023
Deletion of CD36 exhibits limited impact on normal hematopoiesis and the leukemia microenvironment.
Cellular & molecular biology letters
Meng Y,Pospiech M,Ali A,Chandwani R,Vergel M,Onyemaechi S,Yaghmour G,Lu R,Alachkar H
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed May 24 00:00:00 EDT 2023
Spleen-targeted neoantigen DNA vaccine for personalized immunotherapy of hepatocellular carcinoma.
EMBO molecular medicine
Wu M,Luo Z,Cai Z,Mao Q,Li Z,Li H,Zhang C,Zhang Y,Zhong A,Wu L,Liu X
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Oct 11 00:00:00 EDT 2023
Klebsiella pneumoniae Induces Inflammatory Bowel Disease Through Caspase-11-Mediated IL18 in the Gut Epithelial Cells.
Cellular and molecular gastroenterology and hepatology
Zhang Q,Su X,Zhang C,Chen W,Wang Y,Yang X,Liu D,Zhang Y,Yang R
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Feb 23 00:00:00 EST 2023
Purinergic receptor P2Y12 boosts autoimmune hepatitis through hexokinase 2-dependent glycolysis in T cells.
International journal of biological sciences
Zhuang W,Liu X,Liu G,Lv J,Qin H,Wang C,Xie L,Saimaier K,Han S,Shi C,Hua Q,Zhang R,Du C
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Aug 03 00:00:00 EDT 2023
Calcaratarin D, a labdane diterpenoid, attenuates mouse asthma via modulating alveolar macrophage function.
British journal of pharmacology
Liao W,Foo HYC,Tran TNQ,Chai CLL,Wong WSF
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
Palmitoyl transferases act as novel drug targets for pancreatic cancer.
Journal of translational medicine
Lin Z,Lv Z,Liu X,Huang K
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Apr 10 00:00:00 EDT 2023
GSK-3 Inhibitor Elraglusib Enhances Tumor-Infiltrating Immune Cell Activation in Tumor Biopsies and Synergizes with Anti-PD-L1 in a Murine Model of Colorectal Cancer.
International journal of molecular sciences
Huntington KE,Louie AD,Srinivasan PR,Schorl C,Lu S,Silverberg D,Newhouse D,Wu Z,Zhou L,Borden BA,Giles FJ,Dooner M,Carneiro BA,El-Deiry WS
25-0042-82 was used in Flow cytometry/Cell sorting to investigate the immunomodulatory properties of the GSK-3 inhibitor elraglusib, revealing its capacity to enhance immune cell-mediated tumor cell killing, sensitize colorectal cancer cells to immune-mediated cytotoxicity, reduce immunosuppressive factors, and improve survival when combined with immune checkpoint blockade therapy in preclinical models and clinical samples.
Thu Jun 29 00:00:00 EDT 2023
SARS-CoV-2 spike protein promotes inflammatory cytokine activation and aggravates rheumatoid arthritis.
Cell communication and signaling : CCS
Lee AR,Woo JS,Lee SY,Lee YS,Jung J,Lee CR,Park SH,Cho ML
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Mar 02 00:00:00 EST 2023
Platelet factors attenuate inflammation and rescue cognition in ageing.
Nature
Schroer AB,Ventura PB,Sucharov J,Misra R,Chui MKK,Bieri G,Horowitz AM,Smith LK,Encabo K,Tenggara I,Couthouis J,Gross JD,Chan JM,Luke A,Villeda SA
25-0042-82 was used in Flow cytometry/Cell sorting to show that platelet factors transfer the benefits of young blood to the ageing brain.
Tue Aug 01 00:00:00 EDT 2023
TRAF6 controls T cell homeostasis by maintaining the equilibrium of MALT1 scaffolding and protease functions.
Frontiers in immunology
O'Neill TJ,Gewies A,Seeholzer T,Krappmann D
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Feb 13 00:00:00 EST 2023
Group 2 Innate Lymphoid Cells Protect Mice from Abdominal Aortic Aneurysm Formation via IL5 and Eosinophils.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Liu T,Deng Z,Fang W,Zhang X,Zhang S,Wang M,Luo S,Meng Z,Liu J,Sukhova GK,Li D,McKenzie ANJ,Libby P,Shi GP,Guo J
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
miR-4759 suppresses breast cancer through immune checkpoint blockade.
Computational and structural biotechnology journal
Lin YZ,Liu SH,Wu WR,Shen YC,Wang YL,Liao CC,Lin PL,Chang H,Liu LC,Cheng WC,Wang SC
25-0042-82 was used in Flow Cytometry to demonstrate miR-4759 as a novel non-coding RNA which promotes anti-tumor immunity of breast cancer.
Sat Jan 15 00:00:00 EST 2022
Aryl hydrocarbon receptor activation ameliorates experimental colitis by modulating the tolerogenic dendritic and regulatory T cell formation.
Cell & bioscience
Cui X,Ye Z,Wang D,Yang Y,Jiao C,Ma J,Tang N,Zhang H
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat Apr 23 00:00:00 EDT 2022
IL-33 induces thymic involution-associated naive T cell aging and impairs host control of severe infection.
Nature communications
Xu L,Wei C,Chen Y,Wu Y,Shou X,Chen W,Lu D,Sun H,Li W,Yu B,Wang X,Zhang X,Yu Y,Lei Z,Tang R,Zhu J,Li Y,Lu L,Zhou H,Zhou S,Su C,Chen X
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat Nov 12 00:00:00 EST 2022
Fringe GlcNAc-transferases differentially extend O-fucose on endogenous NOTCH1 in mouse activated T cells.
The Journal of biological chemistry
Matsumoto K,Kumar V,Varshney S,Nairn AV,Ito A,Pennarubia F,Moremen KW,Stanley P,Haltiwanger RS
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2022
Breast cancer cell-derived extracellular vesicles promote CD8+ T cell exhaustion via TGF-β type II receptor signaling.
Nature communications
Xie F,Zhou X,Su P,Li H,Tu Y,Du J,Pan C,Wei X,Zheng M,Jin K,Miao L,Wang C,Meng X,van Dam H,Ten Dijke P,Zhang L,Zhou F
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2022
Redox regulation of age-associated defects in generation and maintenance of T cell self-tolerance and immunity to foreign antigens.
Cell reports
Hester AK,Semwal MK,Cepeda S,Xiao Y,Rueda M,Wimberly K,Venables T,Dileepan T,Kraig E,Griffith AV
25-0042-82 was used in Flow cytometry/Cell sorting to study the effects and mechanisms of age on thymic dysfunction.
Tue Feb 15 00:00:00 EST 2022
Dendritic Cell-Based Vaccines Recruit Neutrophils to the Local Draining Lymph Nodes to Prime Natural Killer Cell Responses.
Cells
Chan L,Mehrani Y,Wood GA,Bridle BW,Karimi K
25-0042-82 was used in Flow cytometry/Cell sorting to suggest that neutrophil-mediated anti-tumor immunity induced by a DC vaccine platform could be targeted to provide innovative strategies to enhance its clinical efficacy.
Wed Dec 28 00:00:00 EST 2022
TNF hampers intestinal tissue repair in colitis by restricting IL-22 bioavailability.
Mucosal immunology
Ninnemann J,Winsauer C,Bondareva M,Kühl AA,Lozza L,Durek P,Lissner D,Siegmund B,Kaufmann SHE,Mashreghi MF,Nedospasov SA,Kruglov AA
25-0042-82 was used in Flow cytometry/Cell sorting to show how anti-TNF therapy induces mucosal healing by increasing IL-22 availability and implicates new therapeutic opportunities for IBD.
Fri Apr 01 00:00:00 EDT 2022
Niche-specific MHC II and PD-L1 regulate CD4+CD8αα+ intraepithelial lymphocyte differentiation.
The Journal of experimental medicine
Moon S,Park Y,Hyeon S,Kim YM,Kim JH,Kim H,Park S,Lee KJ,Koo BK,Ha SJ,Lee SW
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Apr 05 00:00:00 EDT 2021
Romidepsin (FK228) regulates the expression of the immune checkpoint ligand PD-L1 and suppresses cellular immune functions in colon cancer.
Cancer immunology, immunotherapy : CII
Shi Y,Fu Y,Zhang X,Zhao G,Yao Y,Guo Y,Ma G,Bai S,Li H
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Potential therapeutic effects of interleukin-35 on the differentiation of naïve T cells into Helios+Foxp3+ Tregs in clinical and experimental acute respiratory distress syndrome.
Molecular immunology
Wang C,Xie K,Li K,Lin S,Xu F
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Anisakis pegreffii Extract Induces Airway Inflammation with Airway Remodeling in a Murine Model System.
BioMed research international
Choi JH,Kim JY,Yi MH,Kim M,Yong TS
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Jan 12 00:00:00 EST 2022
Feline adipose tissue-derived mesenchymal stem cells pretreated with IFN-γ enhance immunomodulatory effects through the PGE₂ pathway.
Journal of veterinary science
Park SG,An JH,Li Q,Chae HK,Park SM,Lee JH,Ahn JO,Song WJ,Youn HY
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
Imbalance between T helper 1 and regulatory T cells plays a detrimental role in experimental Parkinson's disease in mice.
The Journal of international medical research
Li W,Luo Y,Xu H,Ma Q,Yao Q
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Intratumoral expression of interleukin 23 variants using oncolytic vaccinia virus elicit potent antitumor effects on multiple tumor models via tumor microenvironment modulation.
Theranostics
Chen L,Chen H,Ye J,Ge Y,Wang H,Dai E,Ren J,Liu W,Ma C,Ju S,Guo ZS,Liu Z,Bartlett DL
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Aug 04 00:00:00 EDT 2021
Antitumor efficacy of a neoantigen cancer vaccine delivered by electroporation is influenced by microbiota composition.
Oncoimmunology
Lione L,Salvatori E,Petrazzuolo A,Massacci A,Maggio R,Confroti A,Compagnone M,Aurisicchio L,Ciliberto G,Palombo F
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Mar 23 00:00:00 EDT 2021
Evaluation of DNA-Launched Virus-Like Particle Vaccines in an Immune Competent Mouse Model of Chikungunya Virus Infection.
Vaccines
Rayner JO,Kim JH,Roberts RW,Wood RR,Fouty B,Solodushko V
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Apr 02 00:00:00 EDT 2021
Gut Microbiota Dysbiosis Induced by Intracerebral Hemorrhage Aggravates Neuroinflammation in Mice.
Frontiers in microbiology
Yu X,Zhou G,Shao B,Zhou H,Xu C,Yan F,Wang L,Chen G,Li J,Fu X
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed May 26 00:00:00 EDT 2021
Near-infrared photoimmunotherapy targeting human-EGFR in a mouse tumor model simulating current and future clinical trials.
EBioMedicine
Okada R,Furusawa A,Vermeer DW,Inagaki F,Wakiyama H,Kato T,Nagaya T,Choyke PL,Spanos WC,Allen CT,Kobayashi H
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat May 01 00:00:00 EDT 2021
Activation of Bone Marrow Adaptive Immunity in Type 2 Diabetes: Rescue by Co-stimulation Modulator Abatacept.
Frontiers in immunology
Santopaolo M,Sullivan N,Thomas AC,Alvino VV,Nicholson LB,Gu Y,Spinetti G,Kallikourdis M,Blom A,Madeddu P
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Jul 02 00:00:00 EDT 2021
Engineered macrophages as near-infrared light activated drug vectors for chemo-photodynamic therapy of primary and bone metastatic breast cancer.
Nature communications
Huang Y,Guan Z,Dai X,Shen Y,Wei Q,Ren L,Jiang J,Xiao Z,Jiang Y,Liu D,Huang Z,Xu X,Luo Y,Zhao C
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Jul 14 00:00:00 EDT 2021
Histological, behavioural and flow cytometric datasets relating to acute ischaemic stroke in young, aged and ApoE-/- mice in the presence and absence of immunomodulation with fingolimod.
Data in brief
Diaz Diaz AC,Malone K,Shearer JA,Moore AC,Waeber C
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Ddb1 Is Essential for the Expansion of CD4+ Helper T Cells by Regulating Cell Cycle Progression and Cell Death.
Frontiers in immunology
Yang L,Chen W,Li L,Xiao Y,Fan S,Zhang Q,Xia T,Li M,Hong Y,Zhao T,Li Q,Liu WH,Xiao N
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Dec 20 00:00:00 EST 2021
Local Sustained GM-CSF Delivery by Genetically Engineered Encapsulated Cells Enhanced Both Cellular and Humoral SARS-CoV-2 Spike-Specific Immune Response in an Experimental Murine Spike DNA Vaccination Model.
Vaccines
Vernet R,Charrier E,Cosset E,Fièvre S,Tomasello U,Grogg J,Mach N
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon May 10 00:00:00 EDT 2021
Sequencing, cloning, and antigen binding analysis of monoclonal antibodies isolated from single mouse B cells.
STAR protocols
Viant C,Escolano A,Chen ST,Nussenzweig MC
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Jun 18 00:00:00 EDT 2021
Small-molecule antagonist of VLA-4 (GW559090) attenuated neuro-inflammation by targeting Th17 cell trafficking across the blood-retinal barrier in experimental autoimmune uveitis.
Journal of neuroinflammation
Chen YH,Eskandarpour M,Zhang X,Galatowicz G,Greenwood J,Lightman S,Calder V
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Feb 18 00:00:00 EST 2021
Analysis of T cells in mouse lymphoid tissue and blood with flow cytometry.
STAR protocols
Skordos I,Demeyer A,Beyaert R
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Mar 19 00:00:00 EDT 2021
STING activation normalizes the intraperitoneal vascular-immune microenvironment and suppresses peritoneal carcinomatosis of colon cancer.
Journal for immunotherapy of cancer
Lee SJ,Yang H,Kim WR,Lee YS,Lee WS,Kong SJ,Lee HJ,Kim JH,Cheon J,Kang B,Chon HJ,Kim C
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Construction, characterization, and immunization of nanoparticles that display a diverse array of influenza HA trimers.
PloS one
Cohen AA,Yang Z,Gnanapragasam PNP,Ou S,Dam KA,Wang H,Bjorkman PJ
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Oct 15 00:00:00 EDT 2021
Plasmacytoid Dendritic Cells and Type I Interferon Promote Extrafollicular B Cell Responses to Extracellular Self-DNA.
Immunity
Soni C,Perez OA,Voss WN,Pucella JN,Serpas L,Mehl J,Ching KL,Goike J,Georgiou G,Ippolito GC,Sisirak V,Reizis B
25-0042-82 was used in Flow Cytometry to establish extrafollicular B cell differentiation into short-lived AFCs as a key mechanism of anti-DNA autoreactivity and reveal a major contribution of pDCs, endosomal Toll-like receptors (TLRs), and IFN-I to this pathway.
Tue Jun 16 00:00:00 EDT 2020
Protein arginine methyltransferase 5 promotes cholesterol biosynthesis-mediated Th17 responses and autoimmunity.
The Journal of clinical investigation
Webb LM,Sengupta S,Edell C,Piedra-Quintero ZL,Amici SA,Miranda JN,Bevins M,Kennemer A,Laliotis G,Tsichlis PN,Guerau-de-Arellano M
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2020
Nucleoside-modified mRNA vaccination partially overcomes maternal antibody inhibition of de novo immune responses in mice.
Science translational medicine
Willis E,Pardi N,Parkhouse K,Mui BL,Tam YK,Weissman D,Hensley SE
25-0042-82 was used in Flow Cytometry to demonstrate that influenza virus-specific maternal antibodies inhibited de novo antibody responses in mouse pups elicited by influenza virus infection or administration of conventional influenza vaccines.
Wed Jan 08 00:00:00 EST 2020
Methnaridine is an orally bioavailable, fast-killing and long-acting antimalarial agent that cures Plasmodium infections in mice.
British journal of pharmacology
Wang W,Yao J,Chen Z,Sun Y,Shi Y,Wei Y,Zhou H,Yu Y,Li S,Duan L
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2020
Increased baseline RASGRP1 signals enhance stem cell fitness during native hematopoiesis.
Oncogene
Karra L,Romero-Moya D,Ksionda O,Krush M,Gu Z,Mues M,Depeille P,Mullighan C,Roose JP
25-0042 was used in Flow cytometry/Cell sorting to expand RASGRP1 expression surveys in pediatric T-ALL and generated a RoLoRiG mouse model crossed to Mx1CRE to determine the consequences of induced RASGRP1 overexpression in primary hematopoietic cells.
Sun Nov 01 00:00:00 EDT 2020
DKK2 blockage-mediated immunotherapy enhances anti-angiogenic therapy of Kras mutated colorectal cancer.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Hu J,Wang Z,Chen Z,Li A,Sun J,Zheng M,Wu J,Shen T,Qiao J,Li L,Li B,Wu D,Xiao Q
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Increased Tim-3 expression on TILs during treatment with the Anchored GM-CSF vaccine and anti-PD-1 antibodies is inversely correlated with response in prostate cancer.
Journal of Cancer
Zhang X,Chen H,Li G,Zhou X,Shi Y,Zou F,Chen Y,Gao J,Yang S,Wu S,Long Z
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Comparison of rapamycin and methylprednisolone for treating inflammatory muscle disease in a murine model of experimental autoimmune myositis.
Experimental and therapeutic medicine
Kang J,Feng D,Yang F,Tian X,Han W,Jia H
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Effects of CD4+ T lymphocytes from ovariectomized mice on bone marrow mesenchymal stem cell proliferation and osteogenic differentiation.
Experimental and therapeutic medicine
Shao BY,Wang L,Yu Y,Chen L,Gan N,Huang WM
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2020
Dietary Glucose Consumption Promotes RALDH Activity in Small Intestinal CD103+CD11b+ Dendritic Cells.
Frontiers in immunology
Ko HJ,Hong SW,Verma R,Jung J,Lee M,Kim N,Kim D,Surh CD,Kim KS,Rudra D,Im SH
25-0042 was used in Flow cytometry/Cell sorting to reveal previously underappreciated role of dietary glucose concentration in establishing regulatory properties in intestinal DCs, thereby extending a potential therapeutic module against intestinal inflammation.
Mon Apr 19 00:00:00 EDT 2021
Dietary Glucose Consumption Promotes RALDH Activity in Small Intestinal CD103+CD11b+ Dendritic Cells.
Frontiers in immunology
Ko HJ,Hong SW,Verma R,Jung J,Lee M,Kim N,Kim D,Surh CD,Kim KS,Rudra D,Im SH
25-0042 was used in Flow cytometry/Cell sorting to reveal previously underappreciated role of dietary glucose concentration in establishing regulatory properties in intestinal DCs, thereby extending a potential therapeutic module against intestinal inflammation.
Mon Apr 19 00:00:00 EDT 2021
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
Oncolytic vaccinia virus delivering tethered IL-12 enhances antitumor effects with improved safety.
Journal for immunotherapy of cancer
Ge Y,Wang H,Ren J,Liu W,Chen L,Chen H,Ye J,Dai E,Ma C,Ju S,Guo ZS,Liu Z,Bartlett DL
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
A novel antibody targeting TIM-3 resulting in receptor internalization for cancer immunotherapy.
Antibody therapeutics
Kuang Z,Li L,Zhang P,Chen B,Wu M,Ni H,Yi S,Zou J,Liu J
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2020
A novel antibody targeting TIM-3 resulting in receptor internalization for cancer immunotherapy.
Antibody therapeutics
Kuang Z,Li L,Zhang P,Chen B,Wu M,Ni H,Yi S,Zou J,Liu J
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2020
Mechanisms of interleukin 4 mediated increase in efficacy of vaccines against opioid use disorders.
NPJ vaccines
Crouse B,Robinson C,Huseby Kelcher A,Laudenbach M,Abrahante JE,Pravetoni M
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Oct 29 00:00:00 EDT 2020
CD226 Deletion Reduces Type 1 Diabetes in the NOD Mouse by Impairing Thymocyte Development and Peripheral T Cell Activation.
Frontiers in immunology
Shapiro MR,Yeh WI,Longfield JR,Gallagher J,Infante CM,Wellford S,Posgai AL,Atkinson MA,Campbell-Thompson M,Lieberman SM,Serreze DV,Geurts AM,Chen YG,Brusko TM
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed May 26 00:00:00 EDT 2021
Distinct actions of the fermented beverage kefir on host behaviour, immunity and microbiome gut-brain modules in the mouse.
Microbiome
van de Wouw M,Walsh AM,Crispie F,van Leuven L,Lyte JM,Boehme M,Clarke G,Dinan TG,Cotter PD,Cryan JF
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon May 18 00:00:00 EDT 2020
Senolytics prevent mt-DNA-induced inflammation and promote the survival of aged organs following transplantation.
Nature communications
Iske J,Seyda M,Heinbokel T,Maenosono R,Minami K,Nian Y,Quante M,Falk CS,Azuma H,Martin F,Passos JF,Niemann CU,Tchkonia T,Kirkland JL,Elkhal A,Tullius SG
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Aug 27 00:00:00 EDT 2020
Loss of testosterone impairs anti-tumor neutrophil function.
Nature communications
Markman JL,Porritt RA,Wakita D,Lane ME,Martinon D,Noval Rivas M,Luu M,Posadas EM,Crother TR,Arditi M
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Mar 31 00:00:00 EDT 2020
A Commercial Probiotic Induces Tolerogenic and Reduces Pathogenic Responses in Experimental Autoimmune Encephalomyelitis.
Cells
Calvo-Barreiro L,Eixarch H,Ponce-Alonso M,Castillo M,Lebrón-Galán R,Mestre L,Guaza C,Clemente D,Del Campo R,Montalban X,Espejo C
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Apr 07 00:00:00 EDT 2020
Mitochondrial Oxidative Phosphorylation Regulates the Fate Decision between Pathogenic Th17 and Regulatory T Cells.
Cell reports
Shin B,Benavides GA,Geng J,Koralov SB,Hu H,Darley-Usmar VM,Harrington LE
25-0042 was used in Flow cytometry/Cell sorting to study the role of mitochondrial oxidative phosphorylation in pathogenic Th17 differentiation.
Tue Feb 11 00:00:00 EST 2020
Axenic Caenorhabditis elegans antigen protects against development of type-1 diabetes in NOD mice.
Journal of translational autoimmunity
Jackson-Thompson BM,Torrero M,Mitre BK,Long J,Packiam M,Mitre E
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Prodrug-Loaded Zirconium Carbide Nanosheets as a Novel Biophotonic Nanoplatform for Effective Treatment of Cancer.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Liu Q,Xie Z,Qiu M,Shim I,Yang Y,Xie S,Yang Q,Wang D,Chen S,Fan T,Ding B,Guo Z,Adah D,Yao X,Zhang Y,Wu H,Wu Z,Wei C,Wang H,Kim HS,Zou Q,Yan Q,Cai Z,Kim JS,Liu LP,Zhang H,Cao Y
25-0042-82 was used in Flow Cytometry to unravel a promising biphotonic strategy for precision treatment of cancer.
Tue Dec 01 00:00:00 EST 2020
Clonal copy-number mosaicism in autoreactive T lymphocytes in diabetic NOD mice.
Genome research
Alriyami M,Marchand L,Li Q,Du X,Olivier M,Polychronakos C
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sun Dec 01 00:00:00 EST 2019
A unique mutator phenotype reveals complementary oncogenic lesions leading to acute leukemia.
JCI insight
Yin M,Baslan T,Walker RL,Zhu YJ,Freeland A,Matsukawa T,Sridharan S,Nussenzweig A,Pruitt SC,Lowe SW,Meltzer PS,Aplan PD
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Dec 05 00:00:00 EST 2019
Mutation of an L-Type Calcium Channel Gene Leads to T Lymphocyte Dysfunction.
Frontiers in immunology
Fenninger F,Han J,Stanwood SR,Nohara LL,Arora H,Choi KB,Munro L,Pfeifer CG,Shanina I,Horwitz MS,Jefferies WA
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Oct 29 00:00:00 EDT 2020
Systemic inhibition of tissue-nonspecific alkaline phosphatase alters the brain-immune axis in experimental sepsis.
Scientific reports
Brichacek AL,Benkovic SA,Chakraborty S,Nwafor DC,Wang W,Jun S,Dakhlallah D,Geldenhuys WJ,Pinkerton AB,Millán JL,Brown CM
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Dec 11 00:00:00 EST 2019
Chloroquine modulates inflammatory autoimmune responses through Nurr1 in autoimmune diseases.
Scientific reports
Park TY,Jang Y,Kim W,Shin J,Toh HT,Kim CH,Yoon HS,Leblanc P,Kim KS
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Oct 29 00:00:00 EDT 2019
Ufbp1 promotes plasma cell development and ER expansion by modulating distinct branches of UPR.
Nature communications
Zhu H,Bhatt B,Sivaprakasam S,Cai Y,Liu S,Kodeboyina SK,Patel N,Savage NM,Sharma A,Kaufman RJ,Li H,Singh N
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Mar 06 00:00:00 EST 2019
Mitophagy in Intestinal Epithelial Cells Triggers Adaptive Immunity during Tumorigenesis.
Cell
Ziegler PK,Bollrath J,Pallangyo CK,Matsutani T,Canli Ö,De Oliveira T,Diamanti MA,Müller N,Gamrekelashvili J,Putoczki T,Horst D,Mankan AK,Öner MG,Müller S,Müller-Höcker J,Kirchner T,Slotta-Huspenina J,Taketo MM,Reinheckel T,Dröse S,Larner AC,Wels WS,Ernst M,Greten TF,Arkan MC,Korn T,Wirth D,Greten FR
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Jun 28 00:00:00 EDT 2018
T-Cell-Intrinsic Receptor Interacting Protein 2 Regulates Pathogenic T Helper 17 Cell Differentiation.
Immunity
Shimada K,Porritt RA,Markman JL,O'Rourke JG,Wakita D,Noval Rivas M,Ogawa C,Kozhaya L,Martins GA,Unutmaz D,Baloh RH,Crother TR,Chen S,Arditi M
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Nov 20 00:00:00 EST 2018
TCF-1 and HEB cooperate to establish the epigenetic and transcription profiles of CD4+CD8+ thymocytes.
Nature immunology
Emmanuel AO,Arnovitz S,Haghi L,Mathur PS,Mondal S,Quandt J,Okoreeh MK,Maienschein-Cline M,Khazaie K,Dose M,Gounari F
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
Macrophage-specific MHCII expression is regulated by a remote Ciita enhancer controlled by NFAT5.
The Journal of experimental medicine
Buxadé M,Huerga Encabo H,Riera-Borrull M,Quintana-Gallardo L,López-Cotarelo P,Tellechea M,Martínez-Martínez S,Redondo JM,Martín-Caballero J,Flores JM,Bosch E,Rodríguez-Fernández JL,Aramburu J,López-Rodríguez C
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Nov 05 00:00:00 EST 2018
Irg1 expression in myeloid cells prevents immunopathology during M. tuberculosis infection.
The Journal of experimental medicine
Nair S,Huynh JP,Lampropoulou V,Loginicheva E,Esaulova E,Gounder AP,Boon ACM,Schwarzkopf EA,Bradstreet TR,Edelson BT,Artyomov MN,Stallings CL,Diamond MS
Mon Apr 02 00:00:00 EDT 2018
Effector CD4 T cells with progenitor potential mediate chronic intestinal inflammation.
The Journal of experimental medicine
Shin B,Kress RL,Kramer PA,Darley-Usmar VM,Bellis SL,Harrington LE
25-0042 was used in Flow cytometry/Cell sorting to indicate that the chronicity of T cell-mediated inflammation is perpetuated by specific effector CD4 T cells with stem-like properties.
Mon Jul 02 00:00:00 EDT 2018
Bryostatin-1 alleviates experimental multiple sclerosis.
Proceedings of the National Academy of Sciences of the United States of America
Kornberg MD,Smith MD,Shirazi HA,Calabresi PA,Snyder SH,Kim PM
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Feb 27 00:00:00 EST 2018
Quantitative Methods for Measuring Repair Rates and Innate-Immune Cell Responses in Wounded Mouse Skin.
Frontiers in immunology
Li Z,Gothard E,Coles MC,Ambler CA
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Mar 27 00:00:00 EDT 2019
Influenza-Activated ILC1s Contribute to Antiviral Immunity Partially Influenced by Differential GITR Expression.
Frontiers in immunology
Vashist N,Trittel S,Ebensen T,Chambers BJ,Guzmán CA,Riese P
25-0042 was used in Flow cytometry/Cell sorting to discover the regulation behind ILC1s during infection and their capacity to interact with cells of the adaptive immune system.
Fri May 10 00:00:00 EDT 2019
STAT1 is a sex-specific tumor suppressor in colitis-associated colorectal cancer.
Molecular oncology
Crnčec I,Modak M,Gordziel C,Svinka J,Scharf I,Moritsch S,Pathria P,Schlederer M,Kenner L,Timelthaler G,Müller M,Strobl B,Casanova E,Bayer E,Mohr T,Stöckl J,Friedrich K,Eferl R
25-0042 was used in Flow cytometry/Cell sorting to investigate sex-specific STAT1 functions in colitis and colitis-associated colorectal cancer.
Sun Apr 01 00:00:00 EDT 2018
The intestinal microbiota determines the colitis-inducing potential of T-bet-deficient Th cells in mice.
European journal of immunology
Zimmermann J,Durek P,Kühl AA,Schattenberg F,Maschmeyer P,Siracusa F,Lehmann K,Westendorf K,Weber M,Riedel R,Müller S,Radbruch A,Chang HD
25-0042 was used in Flow cytometry/Cell sorting to study the dominance of the microbiota over particular T cell differentiation programs in the pathogenesis of chronic intestinal inflammation.
Mon Jan 01 00:00:00 EST 2018
Protease-activated receptor-2 signaling through β-arrestin-2 mediates Alternaria alkaline serine protease-induced airway inflammation.
American journal of physiology. Lung cellular and molecular physiology
Yee MC,Nichols HL,Polley D,Saifeddine M,Pal K,Lee K,Wilson EH,Daines MO,Hollenberg MD,Boitano S,DeFea KA
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
Granulocyte-macrophage colony-stimulating factor neuroprotective activities in Alzheimer's disease mice.
Journal of neuroimmunology
Kiyota T,Machhi J,Lu Y,Dyavarshetty B,Nemati M,Yokoyama I,Mosley RL,Gendelman HE
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Jun 15 00:00:00 EDT 2018
Immunological effects of a low-dose cytarabine, aclarubicin and granulocyte-colony stimulating factor priming regimen on a mouse leukemia model.
Oncology letters
Chen J,Yang N,Liu H,Yao H,Wang J,Yang Y,Zhang W
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2018
Protective function of Bu Shen Huo Xue formula on the immunity of B6AF1 mice with experimental autoimmune premature ovarian failure.
Experimental and therapeutic medicine
Wang P,Lu Y,Chen S,Chen Y,Hu C,Zuo Y
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Runx-dependent and silencer-independent repression of a maturation enhancer in the Cd4 gene.
Nature communications
Kojo S,Yasmin N,Muroi S,Tenno M,Taniuchi I
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Sep 05 00:00:00 EDT 2018
Runx-dependent and silencer-independent repression of a maturation enhancer in the Cd4 gene.
Nature communications
Kojo S,Yasmin N,Muroi S,Tenno M,Taniuchi I
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Sep 05 00:00:00 EDT 2018
Modifying the cancer-immune set point using vaccinia virus expressing re-designed interleukin-2.
Nature communications
Liu Z,Ge Y,Wang H,Ma C,Feist M,Ju S,Guo ZS,Bartlett DL
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Nov 08 00:00:00 EST 2018
Rap1 deficiency-provoked paracrine dysfunction impairs immunosuppressive potency of mesenchymal stem cells in allograft rejection of heart transplantation.
Cell death & disease
Ding Y,Liang X,Zhang Y,Yi L,Shum HC,Chen Q,Chan BP,Fan H,Liu Z,Tergaonkar V,Qi Z,Tse HF,Lian Q
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Mar 07 00:00:00 EST 2018
Inactivation of TRPM7 kinase in mice results in enlarged spleens, reduced T-cell proliferation and diminished store-operated calcium entry.
Scientific reports
Beesetty P,Wieczerzak KB,Gibson JN,Kaitsuka T,Luu CT,Matsushita M,Kozak JA
25-0042 was used in Flow cytometry/Cell sorting to study the role of TRPM7 channel kinase as a novel modulator of blastogenesis, proliferation and store-operated calcium entry.
Wed Feb 14 00:00:00 EST 2018
Spred1 Safeguards Hematopoietic Homeostasis against Diet-Induced Systemic Stress.
Cell stem cell
Tadokoro Y,Hoshii T,Yamazaki S,Eto K,Ema H,Kobayashi M,Ueno M,Ohta K,Arai Y,Hara E,Harada K,Oshima M,Oshima H,Arai F,Yoshimura A,Nakauchi H,Hirao A
25-0042-82 was used in Flow Cytometry to demonstrate the involvement of diet-induced stress on hematopoietic stem cell self-renewal.
Thu May 03 00:00:00 EDT 2018
Hypothalamic Macrophage Inducible Nitric Oxide Synthase Mediates Obesity-Associated Hypothalamic Inflammation.
Cell reports
Lee CH,Kim HJ,Lee YS,Kang GM,Lim HS,Lee SH,Song DK,Kwon O,Hwang I,Son M,Byun K,Sung YH,Kim S,Kim JB,Choi EY,Kim YB,Kim K,Kweon MN,Sohn JW,Kim MS
25-0042 was used in Flow cytometry to uncover a critical role for hypothalamic macrophage-expressed inducible nitric oxide synthase in hypothalamic inflammation and abnormal glucose metabolism in cases of overnutrition-induced obesity.
Tue Oct 23 00:00:00 EDT 2018
Notch Signaling Facilitates In Vitro Generation of Cross-Presenting Classical Dendritic Cells.
Cell reports
Kirkling ME,Cytlak U,Lau CM,Lewis KL,Resteu A,Khodadadi-Jamayran A,Siebel CW,Salmon H,Merad M,Tsirigos A,Collin M,Bigley V,Reizis B
25-0042 was used in Flow cytometry/Cell sorting to examine the role of Notch signalling in the in vitro generation of classical dendritic cells.
Tue Jun 19 00:00:00 EDT 2018
MERTK inhibition alters the PD-1 axis and promotes anti-leukemia immunity.
JCI insight
Lee-Sherick AB,Jacobsen KM,Henry CJ,Huey MG,Parker RE,Page LS,Hill AA,Wang X,Frye SV,Earp HS,Jordan CT,DeRyckere D,Graham DK
25-0042 was used in Flow cytometry/Cell sorting to demonstrate both the direct therapeutic effects of MERTK inhibition on leukaemia cells and the induction of anti-leukaemia immunity via suppression of the coinhibitory PD-1 axis.
Fri Nov 02 00:00:00 EDT 2018
CD69 enhances immunosuppressive function of regulatory T-cells and attenuates colitis by prompting IL-10 production.
Cell death & disease
Yu L,Yang F,Zhang F,Guo D,Li L,Wang X,Liang T,Wang J,Cai Z,Jin H
25-0042 was used in Flow cytometry/Cell sorting to study the importance of the C-type lectin receptor CD69 to the suppressive function of Tregs.
Wed Sep 05 00:00:00 EDT 2018
Ubiquitination of tumor suppressor PML regulates prometastatic and immunosuppressive tumor microenvironment.
The Journal of clinical investigation
Wang YT,Chen J,Chang CW,Jen J,Huang TY,Chen CM,Shen R,Liang SY,Cheng IC,Yang SC,Lai WW,Cheng KH,Hsieh TS,Lai MZ,Cheng HC,Wang YC,Chen RH
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Excessive expression of miR-27 impairs Treg-mediated immunological tolerance.
The Journal of clinical investigation
Cruz LO,Hashemifar SS,Wu CJ,Cho S,Nguyen DT,Lin LL,Khan AA,Lu LF
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
IL-36 Signaling Facilitates Activation of the NLRP3 Inflammasome and IL-23/IL-17 Axis in Renal Inflammation and Fibrosis.
Journal of the American Society of Nephrology : JASN
Chi HH,Hua KF,Lin YC,Chu CL,Hsieh CY,Hsu YJ,Ka SM,Tsai YL,Liu FC,Chen A
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2017
CD4+ Th2 cells are directly regulated by IL-10 during allergic airway inflammation.
Mucosal immunology
Coomes SM,Kannan Y,Pelly VS,Entwistle LJ,Guidi R,Perez-Lloret J,Nikolov N,Müller W,Wilson MS
25-0042 was used in Flow cytometry/Cell sorting to identify that pathogenic Th2 cells are an important direct target of IL-10 and an insight into IL-10 mediated regualation of allergic airway inflammation.
Sun Jan 01 00:00:00 EST 2017
IL-2/Anti-IL-2 Complex Attenuates Inflammation and BBB Disruption in Mice Subjected to Traumatic Brain Injury.
Frontiers in neurology
Gao W,Li F,Zhou Z,Xu X,Wu Y,Zhou S,Yin D,Sun D,Xiong J,Jiang R,Zhang J
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Discrimination of the heterogeneity of bone marrow‑derived dendritic cells.
Molecular medicine reports
Wang J,Dai X,Hsu C,Ming C,He Y,Zhang J,Wei L,Zhou P,Wang CY,Yang J,Gong N
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Nov 01 00:00:00 EDT 2017
Immunological effects of vaccines combined with granulocyte colony-stimulating factor on a murine WEHI-3 leukemia model.
Oncology letters
Chen J,Zhang M,Zhou F,Wang J,Niu B,Zhang W
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Effect of follicular helper T cells on the pathogenesis of asthma.
Experimental and therapeutic medicine
Ma X,Sun Z,Zhai P,Yu W,Wang T,Li F,Ding J
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Batf3-dependent CD8α+ Dendritic Cells Aggravates Atherosclerosis via Th1 Cell Induction and Enhanced CCL5 Expression in Plaque Macrophages.
EBioMedicine
Li Y,Liu X,Duan W,Tian H,Zhu G,He H,Yao S,Yi S,Song W,Tang H
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Isolation of cell type-specific apoptotic bodies by fluorescence-activated cell sorting.
Scientific reports
Atkin-Smith GK,Paone S,Zanker DJ,Duan M,Phan TK,Chen W,Hulett MD,Poon IK
25004282 was used in flow cytometry to establish a method to isolate apoptotic bodies from cultured cells to 99% purity
Fri Jan 06 00:00:00 EST 2017
IL-7-dependent STAT1 activation limits homeostatic CD4+ T cell expansion.
JCI insight
Le Saout C,Luckey MA,Villarino AV,Smith M,Hasley RB,Myers TG,Imamichi H,Park JH,O'Shea JJ,Lane HC,Catalfamo M
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Nov 16 00:00:00 EST 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Differential regulation of C5a receptor 1 in innate immune cells during the allergic asthma effector phase.
PloS one
Ender F,Wiese AV,Schmudde I,Sun J,Vollbrandt T,König P,Laumonnier Y,Köhl J
25-0042 was used in Flow cytometry/Cell sorting to demonstrate a complex regulation pattern of C5aR1 in the airways, lung tissue and lymph nodes of mice.
Thu Aug 17 00:00:00 EDT 2017
Type I IFN and not TNF, is Essential for Cyclic Di-nucleotide-elicited CTL by a Cytosolic Cross-presentation Pathway.
EBioMedicine
Lirussi D,Ebensen T,Schulze K,Trittel S,Duran V,Liebich I,Kalinke U,Guzmán CA
25-0042 was used in Flow Cytometry to demonstrate that stimulation of interferon-γ-secreting CD8+ cytotoxic T lymphocytes is significantly decreased after vaccination in the absence of type I interferon signalling.
Tue Aug 01 00:00:00 EDT 2017
Longitudinal intravital imaging of the femoral bone marrow reveals plasticity within marrow vasculature.
Nature communications
Reismann D,Stefanowski J,Günther R,Rakhymzhan A,Matthys R,Nützi R,Zehentmeier S,Schmidt-Bleek K,Petkau G,Chang HD,Naundorf S,Winter Y,Melchers F,Duda G,Hauser AE,Niesner RA
25-0042 was used in Flow cytometry/Cell sorting to develop longitudinal intravital imaging of the bone marrow to analyse cellular dynamics within the deep marrow.
Mon Dec 18 00:00:00 EST 2017
CD69 controls the uptake of L-tryptophan through LAT1-CD98 and AhR-dependent secretion of IL-22 in psoriasis.
Nature immunology
Cibrian D,Saiz ML,de la Fuente H,Sánchez-Díaz R,Moreno-Gonzalo O,Jorge I,Ferrarini A,Vázquez J,Punzón C,Fresno M,Vicente-Manzanares M,Daudén E,Fernández-Salguero PM,Martín P,Sánchez-Madrid F
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Oligodendrocyte death results in immune-mediated CNS demyelination.
Nature neuroscience
Traka M,Podojil JR,McCarthy DP,Miller SD,Popko B
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
Antitumor adaptive immunity remains intact following inhibition of autophagy and antimalarial treatment.
The Journal of clinical investigation
Starobinets H,Ye J,Broz M,Barry K,Goldsmith J,Marsh T,Rostker F,Krummel M,Debnath J
25-0042 was used in Flow cytometry/Cell sorting to evaluate how autophagy inhibition impacts the antitumor immune response in immune-competent mouse models of melanoma and mammary cancer.
Thu Dec 01 00:00:00 EST 2016
The composition of the microbiota modulates allograft rejection.
The Journal of clinical investigation
Lei YM,Chen L,Wang Y,Stefka AT,Molinero LL,Theriault B,Aquino-Michaels K,Sivan AS,Nagler CR,Gajewski TF,Chong AS,Bartman C,Alegre ML
25-0042 was used in Flow cytometry/Cell sorting to test the influence of host and donor microbiota on skin transplant rejection.
Fri Jul 01 00:00:00 EDT 2016
STAT3/5-Dependent IL9 Overexpression Contributes to Neoplastic Cell Survival in Mycosis Fungoides.
Clinical cancer research : an official journal of the American Association for Cancer Research
Vieyra-Garcia PA,Wei T,Naym DG,Fredholm S,Fink-Puches R,Cerroni L,Odum N,O'Malley JT,Gniadecki R,Wolf P
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
Genomic and immunologic factors associated with viral pathogenesis in a lethal EV71 infected neonatal mouse model.
Molecular medicine reports
Yue Y,Li P,Song N,Li B,Li Z,Guo Y,Zhang W,Wei MQ,Gai Z,Meng H,Wang J,Qin L
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
Deletion of Wiskott-Aldrich syndrome protein triggers Rac2 activity and increased cross-presentation by dendritic cells.
Nature communications
Baptista MA,Keszei M,Oliveira M,Sunahara KK,Andersson J,Dahlberg CI,Worth AJ,Liedén A,Kuo IC,Wallin RP,Snapper SB,Eidsmo L,Scheynius A,Karlsson MC,Bouma G,Burns SO,Forsell MN,Thrasher AJ,Nylén S,Westerberg LS
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Jul 18 00:00:00 EDT 2016
Affinity and dose of TCR engagement yield proportional enhancer and gene activity in CD4+ T cells.
eLife
Allison KA,Sajti E,Collier JG,Gosselin D,Troutman TD,Stone EL,Hedrick SM,Glass CK
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Jul 04 00:00:00 EDT 2016
Sex bias in CNS autoimmune disease mediated by androgen control of autoimmune regulator.
Nature communications
Zhu ML,Bakhru P,Conley B,Nelson JS,Free M,Martin A,Starmer J,Wilson EM,Su MA
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Apr 13 00:00:00 EDT 2016
Irradiation of necrotic cancer cells, employed for pulsing dendritic cells (DCs), potentiates DC vaccine-induced antitumor immunity against high-grade glioma.
Oncoimmunology
Vandenberk L,Garg AD,Verschuere T,Koks C,Belmans J,Beullens M,Agostinis P,De Vleeschouwer S,Van Gool SW
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2016
Exploring the induction of preproinsulin-specific Foxp3(+) CD4(+) Treg cells that inhibit CD8(+) T cell-mediated autoimmune diabetes by DNA vaccination.
Scientific reports
Stifter K,Schuster C,Schlosser M,Boehm BO,Schirmbeck R
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Jul 11 00:00:00 EDT 2016
Timing of CSF-1/CSF-1R signaling blockade is critical to improving responses to CTLA-4 based immunotherapy.
Oncoimmunology
Holmgaard RB,Brachfeld A,Gasmi B,Jones DR,Mattar M,Doman T,Murphy M,Schaer D,Wolchok JD,Merghoub T
25-0042 was used in Flow cytometry/Cell sorting to show that both CSF-1 and its receptor (CSF-1R) are frequently expressed in tumors from cancer patients, and that this expression correlates with tumor-infiltration of MDSCs.
Fri Jul 01 00:00:00 EDT 2016
IL-1 signaling modulates activation of STAT transcription factors to antagonize retinoic acid signaling and control the TH17 cell-iTreg cell balance.
Nature immunology
Basu R,Whitley SK,Bhaumik S,Zindl CL,Schoeb TR,Benveniste EN,Pear WS,Hatton RD,Weaver CT
25-0042 was used in Flow cytometry/Cell sorting to assess the ability of IL-1 to override retinoic acid-mediated expression of the transcription factor Foxp3 and induce protective TH17 cell responses.
Sun Mar 01 00:00:00 EST 2015
A Threshold Level of Intratumor CD8+ T-cell PD1 Expression Dictates Therapeutic Response to Anti-PD1.
Cancer research
Ngiow SF,Young A,Jacquelot N,Yamazaki T,Enot D,Zitvogel L,Smyth MJ
25-0042 was used in Flow cytometry/Cell sorting to provide a framework to interrogate intra-tumour CD8(+) T-cell PD1 and immune PDL1 levels and response in human cancer.
Tue Sep 15 00:00:00 EDT 2015
Atypical chemokine receptor 1 deficiency reduces atherogenesis in ApoE-knockout mice.
Cardiovascular research
Wan W,Liu Q,Lionakis MS,Marino AP,Anderson SA,Swamydas M,Murphy PM
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
B-cell very late antigen-4 deficiency reduces leukocyte recruitment and susceptibility to central nervous system autoimmunity.
Annals of neurology
Lehmann-Horn K,Sagan SA,Bernard CC,Sobel RA,Zamvil SS
25-0042 was used in Flow cytometry/Cell sorting to report that the selective inhibition of VLA-4 expression on B cells impedes CNS accumulation of B cells, and recruitment of Th17 cells and macrophages.
Fri May 01 00:00:00 EDT 2015
Single-Dose CpG Immunization Protects Against a Heterosubtypic Challenge and Generates Antigen-Specific Memory T Cells.
Frontiers in immunology
Vogel AJ,Brown DM
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Jul 10 00:00:00 EDT 2015
IFNγ and IL-12 Restrict Th2 Responses during Helminth/Plasmodium Co-Infection and Promote IFNγ from Th2 Cells.
PLoS pathogens
Coomes SM,Pelly VS,Kannan Y,Okoye IS,Czieso S,Entwistle LJ,Perez-Lloret J,Nikolov N,Potocnik AJ,Biró J,Langhorne J,Wilson MS
25-0042 was used in Flow cytometry/Cell sorting to show that co-infection with Plasmodium spp. may contribute to the chronicity of helminth infection by reducing the frequency of anti-helminth Th2 cells.
Wed Jul 01 00:00:00 EDT 2015
Premature thymic involution is independent of structural plasticity of the thymic stroma.
European journal of immunology
Franckaert D,Schlenner SM,Heirman N,Gill J,Skogberg G,Ekwall O,Put K,Linterman MA,Dooley J,Liston A
25-0042 was used in Flow cytometry/Cell sorting to investigate thymic involution in FVB/N mice, showing that disruption of the epithelial-endothelial relationship and a progressive loss of pro-T cells precedes involution, and loss of pro-T cells is sufficient to drive premature involution, suggesting that pro-T cells are the main driver of involution.
Fri May 01 00:00:00 EDT 2015
A novel Cd8-cis-regulatory element preferentially directs expression in CD44hiCD62L+ CD8+ T cells and in CD8αα+ dendritic cells.
Journal of leukocyte biology
Sakaguchi S,Hombauer M,Hassan H,Tanaka H,Yasmin N,Naoe Y,Bilic I,Moser MA,Hainberger D,Mayer H,Seiser C,Bergthaler A,Taniuchi I,Ellmeier W
25-0042 was used in Flow cytometry/Cell sorting to identify a novel Cd8 enhancer that directs expression in CD44(hi)CD62L(+) CD8(+) T cells, including innate-like and antigen-specific effector/memory CD8(+) T cells and in CD8αα(+) DCs.
Wed Apr 01 00:00:00 EDT 2015
A tumor lysate is an effective vaccine antigen for the stimulation of CD4(+) T-cell function and subsequent induction of antitumor immunity mediated by CD8(+) T cells.
Cancer biology & therapy
Kawahara M,Takaku H
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Sep 12 00:00:00 EDT 2016
Immunogenicity and protective efficacy of DMT liposome-adjuvanted tuberculosis subunit CTT3H vaccine.
Human vaccines & immunotherapeutics
Teng X,Tian M,Li J,Tan S,Yuan X,Yu Q,Jing Y,Zhang Z,Yue T,Zhou L,Fan X
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Mar 11 00:00:00 EST 2016
Enhanced T-cell activation and differentiation in lymphocytes from transgenic mice expressing ubiquitination-resistant 2KR LAT molecules.
Gene therapy
Rodriguez-Peña AB,Gomez-Rodriguez J,Kortum RL,Palmer DC,Yu Z,Guittard GC,Wohlfert EA,Silver PB,Misplon JA,Sommers CL,Feigenbaum L,Epstein SL,Caspi RR,Belkaid Y,Restifo NP,Samelson LE,Balagopalan L
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
Induction of Th17 cells by segmented filamentous bacteria in the murine intestine.
Journal of immunological methods
Farkas AM,Panea C,Goto Y,Nakato G,Galan-Diez M,Narushima S,Honda K,Ivanov II
25-0042 was used in Flow cytometry/Cell sorting to study the ability of segmented filamentous bacteria (SFB) to modulate the host immune system through induction of IgA and Th17 cells, and examine the kinetics of SFB colonisation and Th17 cell induction.
Mon Jun 01 00:00:00 EDT 2015
Recombinant adeno-vaccine expressing enterovirus 71-like particles against hand, foot, and mouth disease.
PLoS neglected tropical diseases
Tsou YL,Lin YW,Shao HY,Yu SL,Wu SR,Lin HY,Liu CC,Huang C,Chong P,Chow YH
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
dNP2 is a blood-brain barrier-permeable peptide enabling ctCTLA-4 protein delivery to ameliorate experimental autoimmune encephalomyelitis.
Nature communications
Lim S,Kim WJ,Kim YH,Lee S,Koo JH,Lee JA,Yoon H,Kim DH,Park HJ,Kim HM,Lee HG,Yun Kim J,Lee JU,Hun Shin J,Kyun Kim L,Doh J,Kim H,Lee SK,Bothwell ALM,Suh M,Choi JM
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Sep 15 00:00:00 EDT 2015
Near-infrared photoactivatable control of Ca(2+) signaling and optogenetic immunomodulation.
eLife
He L,Zhang Y,Ma G,Tan P,Li Z,Zang S,Wu X,Jing J,Fang S,Zhou L,Wang Y,Huang Y,Hogan PG,Han G,Zhou Y
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2015
STAT4 controls GM-CSF production by both Th1 and Th17 cells during EAE.
Journal of neuroinflammation
McWilliams IL,Rajbhandari R,Nozell S,Benveniste E,Harrington LE
25-0042 was used in Flow cytometry/Cell sorting to establish the role of STAT4 in the regulation of cytokine production during experimental autoimmune encephalomyelitis pathogenesis.
Tue Jun 30 00:00:00 EDT 2015
Antibiotics in neonatal life increase murine susceptibility to experimental psoriasis.
Nature communications
Zanvit P,Konkel JE,Jiao X,Kasagi S,Zhang D,Wu R,Chia C,Ajami NJ,Smith DP,Petrosino JF,Abbatiello B,Nakatsukasa H,Chen Q,Belkaid Y,Chen ZJ,Chen W
25-0042 was used in Flow cytometry/Cell sorting to show that the addition of antibiotics in neonatal life increase murine susceptibility to experimental psoriasis.
Tue Sep 29 00:00:00 EDT 2015
Distortion of the Major Histocompatibility Complex Class I Binding Groove to Accommodate an Insulin-derived 10-Mer Peptide.
The Journal of biological chemistry
Motozono C,Pearson JA,De Leenheer E,Rizkallah PJ,Beck K,Trimby A,Sewell AK,Wong FS,Cole DK
25-0042 was used in Flow cytometry/Cell sorting to study how distortion in the MHC class I binding groove can accommodate extra C-terminal peptide residues..
Fri Jul 31 00:00:00 EDT 2015
Immune complexes stimulate CCR7-dependent dendritic cell migration to lymph nodes.
Nature medicine
Clatworthy MR,Aronin CE,Mathews RJ,Morgan NY,Smith KG,Germain RN
25-0042 was used in Flow cytometry/Cell sorting to study how immune complexes stimulate CCR7-dependent dendritic cell migration to lymph nodes.
Mon Dec 01 00:00:00 EST 2014
Immune complexes stimulate CCR7-dependent dendritic cell migration to lymph nodes.
Nature medicine
Clatworthy MR,Aronin CE,Mathews RJ,Morgan NY,Smith KG,Germain RN
25-0042 was used in Flow cytometry/Cell sorting to study how immune complexes stimulate CCR7-dependent dendritic cell migration to lymph nodes.
Mon Dec 01 00:00:00 EST 2014
Unexpected link between an antibiotic, pannexin channels and apoptosis.
Nature
Poon IK,Chiu YH,Armstrong AJ,Kinchen JM,Juncadella IJ,Bayliss DA,Ravichandran KS
25-0042 was used in Flow cytometry/Cell sorting to show that the quinolone antibiotic trovafloxacin is a novel PANX1 inhibitor.
Thu Mar 20 00:00:00 EDT 2014
Quantitative and temporal requirements revealed for Zap70 catalytic activity during T cell development.
Nature immunology
Au-Yeung BB,Melichar HJ,Ross JO,Cheng DA,Zikherman J,Shokat KM,Robey EA,Weiss A
25-0042 was used in Flow cytometry/Cell sorting to show that CD4(+)CD8(+) thymocytes integrate multiple, transient, Zap70-dependent signals over more than 36 h to reach a cumulative threshold for positive selection.
Tue Jul 01 00:00:00 EDT 2014
Identifying gut microbe-host phenotype relationships using combinatorial communities in gnotobiotic mice.
Science translational medicine
Faith JJ,Ahern PP,Ridaura VK,Cheng J,Gordon JI
25-0042 was used in Flow cytometry/Cell sorting to describe a method in which a clonally arrayed collection of cultured, sequenced bacteria was generated from one of several human fecal microbiota samples found to transmit a particular phenotype to recipient germ-free mice.
Wed Jan 22 00:00:00 EST 2014
TLR2 engagement on CD4(+) T cells enhances effector functions and protective responses to Mycobacterium tuberculosis.
European journal of immunology
Reba SM,Li Q,Onwuzulike S,Ding X,Karim AF,Hernandez Y,Fulton SA,Harding CV,Lancioni CL,Nagy N,Rodriguez ME,Wearsch PA,Rojas RE
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu May 01 00:00:00 EDT 2014
Site-specific chemokine expression regulates central nervous system inflammation and determines clinical phenotype in autoimmune encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950)
Stoolman JS,Duncker PC,Huber AK,Segal BM
25-0042 was used in Flow cytometry/Cell sorting to investigate the role of CXCL2 in atypical EAE, showing that site-specific chemokine expression regulates CNS inflammation and determines clinical phenotype.
Tue Jul 15 00:00:00 EDT 2014
T Cell factor 1 represses CD8+ effector T cell formation and function.
Journal of immunology (Baltimore, Md. : 1950)
Tiemessen MM,Baert MR,Kok L,van Eggermond MC,van den Elsen PJ,Arens R,Staal FJ
25-0042 was used in Flow cytometry/Cell sorting to explore the role of the Wnt-responsive transcription factor T cell factor 1 in the initial phases of CD8(+) T effector cell formation.
Mon Dec 01 00:00:00 EST 2014
Lasting antibody responses are mediated by a combination of newly formed and established bone marrow plasma cells drawn from clonally distinct precursors.
Journal of immunology (Baltimore, Md. : 1950)
Chernova I,Jones DD,Wilmore JR,Bortnick A,Yucel M,Hershberg U,Allman D
25-0042 was used in Flow cytometry/Cell sorting to investigate the role of subpopulations of bone marrow plasma cells in long-term humoral responses to T cell-dependent Ag.
Sat Nov 15 00:00:00 EST 2014
Site-dependent recruitment of inflammatory cells determines the effective dose of Leishmania major.
Infection and immunity
Ribeiro-Gomes FL,Roma EH,Carneiro MB,Doria NA,Sacks DL,Peters NC
25-0042 was used in Flow cytometry/Cell sorting to investigate the effect of Leishmania major intradermal infection site on the inflammatory cell population and effective dose.
Tue Jul 01 00:00:00 EDT 2014
The mucosal adjuvant cyclic di-AMP exerts immune stimulatory effects on dendritic cells and macrophages.
PloS one
Škrnjug I,Rueckert C,Libanova R,Lienenklaus S,Weiss S,Guzmán CA
25-0042 was used in Flow cytometry/Cell sorting to assess changes in cell surface marker expression of dendritic cells and macrophages in response to c-di-AMP.
Wed Jan 14 00:00:00 EST 2015
Cyclic GMP-AMP displays mucosal adjuvant activity in mice.
PloS one
Škrnjug I,Guzmán CA,Rueckert C
25-0042 was used in Flow cytometry/Cell sorting to explore the efficacy of cGAMP as a mucosal adjuvant in mice.
Fri Oct 09 00:00:00 EDT 2015
Gene Expression Driven by a Strong Viral Promoter in MVA Increases Vaccination Efficiency by Enhancing Antibody Responses and Unmasking CD8⁺ T Cell Epitopes.
Vaccines
Becker PD,Nörder M,Weissmann S,Ljapoci R,Erfle V,Drexler I,Guzmán CA
25-0042 was used in Flow cytometry/Cell sorting to evaluate the effect of overexpressing recombinant Ag using modified vaccinia virus early/late promoter H5 (mPH5), showing that overexpression enhances antibody responses and increases vaccination efficiency.
Tue Jul 22 00:00:00 EDT 2014
Leukocyte population dynamics and detection of IL-9 as a major cytokine at the mouse fetal-maternal interface.
PloS one
Habbeddine M,Verbeke P,Karaz S,Bobé P,Kanellopoulos-Langevin C
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Jun 10 00:00:00 EDT 2015
The efficacy of tumor debulking surgery is improved by adjuvant immunotherapy using imiquimod and anti-CD40.
BMC cancer
Khong A,Cleaver AL,Fahmi Alatas M,Wylie BC,Connor T,Fisher SA,Broomfield S,Lesterhuis WJ,Currie AJ,Lake RA,Robinson BW
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Wed Dec 17 00:00:00 EST 2014
P2X7 receptor modulates inflammatory and functional pulmonary changes induced by silica.
PloS one
Monção-Ribeiro LC,Faffe DS,Santana PT,Vieira FS,da Graça CL,Marques-da-Silva C,Machado MN,Caruso-Neves C,Zin WA,Borojevic R,Takiya CM,Coutinho-Silva R
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2015
MHC class II-dependent B cell APC function is required for induction of CNS autoimmunity independent of myelin-specific antibodies.
The Journal of experimental medicine
Molnarfi N,Schulze-Topphoff U,Weber MS,Patarroyo JC,Prod'homme T,Varrin-Doyer M,Shetty A,Linington C,Slavin AJ,Hidalgo J,Jenne DE,Wekerle H,Sobel RA,Bernard CC,Shlomchik MJ,Zamvil SS
25-0042 was used in Flow cytometry/Cell sorting to investigate the contribution of B cells to antigen presentation and pathogenic T cell activation in the multiple sclerosis model experimental autoimmune encephalomyelitis.
Mon Dec 16 00:00:00 EST 2013
Genetic deletion of chemokine receptor Ccr7 exacerbates atherogenesis in ApoE-deficient mice.
Cardiovascular research
Wan W,Lionakis MS,Liu Q,Roffê E,Murphy PM
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
Group X secreted phospholipase A2 limits the development of atherosclerosis in LDL receptor-null mice.
Arteriosclerosis, thrombosis, and vascular biology
Ait-Oufella H,Herbin O,Lahoute C,Coatrieux C,Loyer X,Joffre J,Laurans L,Ramkhelawon B,Blanc-Brude O,Karabina S,Girard CA,Payré C,Yamamoto K,Binder CJ,Murakami M,Tedgui A,Lambeau G,Mallat Z
25-0042 was used in Flow cytometry/Cell sorting to study the proatherogenic properties of PLA2G10, a type of several secreted phospholipase A2.
Fri Mar 01 00:00:00 EST 2013
Group V secretory phospholipase A2 is involved in macrophage activation and is sufficient for macrophage effector functions in allergic pulmonary inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Ohta S,Imamura M,Xing W,Boyce JA,Balestrieri B
25-0042 was used in Flow cytometry/Cell sorting to identify a unique function for gV-sPLA2 in the activation and recruitment of immune cells.
Sat Jun 15 00:00:00 EDT 2013
Aurintricarboxylic acid ameliorates experimental autoimmune encephalomyelitis by blocking chemokine-mediated pathogenic cell migration and infiltration.
Journal of immunology (Baltimore, Md. : 1950)
Zhang F,Wei W,Chai H,Xie X
25-0042 was used in Immunofluorescence on frozen tissues to investigate the effect of aurintricarboxylic acid (ATA) on experimental autoimmune encephalomyelitis (EAE), showing that ATA ameliorates EAE by blocking chemokine-mediated pathogenic cell migration and infiltration.
Fri Feb 01 00:00:00 EST 2013
Myeloid dendritic cells (DCs) of mice susceptible to paracoccidioidomycosis suppress T cell responses whereas myeloid and plasmacytoid DCs from resistant mice induce effector and regulatory T cells.
Infection and immunity
Pina A,de Araujo EF,Felonato M,Loures FV,Feriotti C,Bernardino S,Barbuto JA,Calich VL
25-0042 was used in Flow cytometry/Cell sorting to highlight the deleterious effect of excessive innate proinflammatory reaction and provide new evidence for the importance of immunomodulation during pulmonary paracoccidioidomycosis.
Mon Apr 01 00:00:00 EDT 2013
Nod2 activates NF-kB in CD4+ T cells but its expression is dispensable for T cell-induced colitis.
PloS one
Zanello G,Goethel A,Forster K,Geddes K,Philpott DJ,Croitoru K
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Thu Jul 31 00:00:00 EDT 2014
Characterization of two distinct lymphoproliferative diseases caused by ectopic expression of the Notch ligand DLL4 on T cells.
PloS one
Xiong H,Maraver A,Latkowski JA,Henderson T,Schlessinger K,Ding Y,Shen J,Tadokoro CE,Lafaille JJ
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Mon Feb 23 00:00:00 EST 2015
Immune response is an important aspect of the antitumor effect produced by a CD40L-encoding oncolytic adenovirus.
Cancer research
Diaconu I,Cerullo V,Hirvinen ML,Escutenaire S,Ugolini M,Pesonen SK,Bramante S,Parviainen S,Kanerva A,Loskog AS,Eliopoulos AG,Pesonen S,Hemminki A
25-0042 was used in Flow cytometry/Cell sorting to test whether a transcriptionally targeted oncolytic adenovirus, which features a capsid modification and is armed with CD40L, can result in oncolytic antitumor activity and stimulate an immune response.
Tue May 01 00:00:00 EDT 2012
PSGL-1 regulates the migration and proliferation of CD8(+) T cells under homeostatic conditions.
Journal of immunology (Baltimore, Md. : 1950)
Veerman KM,Carlow DA,Shanina I,Priatel JJ,Horwitz MS,Ziltener HJ
25-0042 was used in Flow cytometry/Cell sorting to demonstrate a novel, pleiotropic influence of PSGL-1 deficiency on several aspects of T cell homeostasis.
Wed Feb 15 00:00:00 EST 2012
A critical role for Rictor in T lymphopoiesis.
Journal of immunology (Baltimore, Md. : 1950)
Tang F,Wu Q,Ikenoue T,Guan KL,Liu Y,Zheng P
25-0042 was used in Flow cytometry/Cell sorting to investigate the signalling pathway required for T lymphopoiesis, showing a critical role for Rictor.
Wed Aug 15 00:00:00 EDT 2012
Evaluation of Lassa virus vaccine immunogenicity in a CBA/J-ML29 mouse model.
Vaccine
Goicochea MA,Zapata JC,Bryant J,Davis H,Salvato MS,Lukashevich IS
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Feb 14 00:00:00 EST 2012
The stalk domain and the glycosylation status of the activating natural killer cell receptor NKp30 are important for ligand binding.
The Journal of biological chemistry
Hartmann J,Tran TV,Kaudeer J,Oberle K,Herrmann J,Quagliano I,Abel T,Cohnen A,Gatterdam V,Jacobs A,Wollscheid B,Tampé R,Watzl C,Diefenbach A,Koch J
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Sep 07 00:00:00 EDT 2012
SH2D2A modulates T cell mediated protection to a B cell derived tumor in transgenic mice.
PloS one
Berge T,Grønningsæter IH,Lorvik KB,Abrahamsen G,Granum S,Sundvold-Gjerstad V,Corthay A,Bogen B,Spurkland A
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Tue Aug 26 00:00:00 EDT 2014
Nuclear export of the NF-κB inhibitor IκBα is required for proper B cell and secondary lymphoid tissue formation.
Immunity
Wuerzberger-Davis SM,Chen Y,Yang DT,Kearns JD,Bates PW,Lynch C,Ladell NC,Yu M,Podd A,Zeng H,Huang TT,Wen R,Hoffmann A,Wang D,Miyamoto S
Published figure using CD4 monoclonal antibody (Product # 25-0042-82) in Flow Cytometry
Fri Feb 25 00:00:00 EST 2011
Thymus-derived rather than tumor-induced regulatory T cells predominate in brain tumors.
Neuro-oncology
Wainwright DA,Sengupta S,Han Y,Lesniak MS
25-0042 was used in Flow cytometry/Cell sorting to investigate whether brain-tumour-resident Tregs are thymus-derived natural Tregs or induced Tregs, showing that thymus-derived Tregs predominate.
Thu Dec 01 00:00:00 EST 2011
Ly49D-mediated ITAM signaling in immature thymocytes impairs development by bypassing the pre-TCR checkpoint.
Journal of immunology (Baltimore, Md. : 1950)
Merck E,Lees RK,Voyle RB,Held W,MacDonald HR
25-0042 was used in Flow cytometry/Cell sorting to investigate the role of Ly49D-mediated ITAM signaling, showing that it impairs development by bypassing the pre-TCR checkpoint.
Fri Jul 01 00:00:00 EDT 2011
B cell depletion reduces the development of atherosclerosis in mice.
The Journal of experimental medicine
Ait-Oufella H,Herbin O,Bouaziz JD,Binder CJ,Uyttenhove C,Laurans L,Taleb S,Van Vré E,Esposito B,Vilar J,Sirvent J,Van Snick J,Tedgui A,Tedder TF,Mallat Z
25-0042 was used in Flow cytometry/Cell sorting to challenge the current paradigm that B cell activation plays an overall protective role in atherogenesis, and identify new antiatherogenic strategies based on B cell modulation.
Mon Aug 02 00:00:00 EDT 2010
Partial restoration of T-cell function in aged mice by in vitro blockade of the PD-1/ PD-L1 pathway.
Aging cell
Lages CS,Lewkowich I,Sproles A,Wills-Karp M,Chougnet C
25-0042 was used in Flow cytometry/Cell sorting to suggest that blockade of the PD-1/PD-L1 is not likely to be efficient at restoring exhausted T-cell responses in aged hosts.
Fri Oct 01 00:00:00 EDT 2010
Foxa2 programs Th2 cell-mediated innate immunity in the developing lung.
Journal of immunology (Baltimore, Md. : 1950)
Chen G,Wan H,Luo F,Zhang L,Xu Y,Lewkowich I,Wills-Karp M,Whitsett JA
25-0042 was used in Flow cytometry/Cell sorting to demonstrate that selective expression of Foxa2 plays a key role in regulating Th2 cell-mediated pulmonary inflammation.
Tue Jun 01 00:00:00 EDT 2010
Resistance to dengue virus infection in mice is potentiated by CXCL10 and is independent of CXCL10-mediated leukocyte recruitment.
Journal of immunology (Baltimore, Md. : 1950)
Ip PP,Liao F
25-0042 was used in Flow cytometry/Cell sorting to study how CXCL10(-/-) mice are more susceptible than wild-type mice to dengue virus infection.
Sat May 15 00:00:00 EDT 2010
Modified vaccinia virus Ankara exerts potent immune modulatory activities in a murine model.
PloS one
Nörder M,Becker PD,Drexler I,Link C,Erfle V,Guzmán CA
25-0042 was used in Flow cytometry/Cell sorting to assess the immune response to vaccine antigens co-administered or not co-administered with modified vaccinia virus Ankara.
Wed Jun 30 00:00:00 EDT 2010
Regulation of cytokine production by virus-specific CD8 T cells in the lungs.
Journal of virology
Fulton RB,Olson MR,Varga SM
25-0042 was used in Flow cytometry/Cell sorting to show that both virus-induced inflammation and the host tissue environment influence the capacity of virus-specific T cells to produce cytokines.
Fri Aug 01 00:00:00 EDT 2008
IL-10 regulates movement of intestinally derived CD4+ T cells to the liver.
Journal of immunology (Baltimore, Md. : 1950)
Bliss SK,Bliss SP,Beiting DP,Alcaraz A,Appleton JA
Fri Jun 15 00:00:00 EDT 2007
Alloantigen recognition is critical for CD8 T cell-mediated graft anti-tumor activity against murine BCL1 lymphoma after myeloablative bone marrow transplantation.
Bone marrow transplantation
Pillai A,Teo P,George T,Mukhopadhyay A,Dejbakhsh-Jones S,Strober S
25-0042 was used in Flow cytometry/Cell sorting to determine whether whole bone marrow cells or splenic CD8 T cells from C57BL/6 donor mice, are capable of mediating graft anti-tumor activity against a BALB/c B-cell lymphoma.
Sat Sep 01 00:00:00 EDT 2007
Converting IL-15 to a superagonist by binding to soluble IL-15R{alpha}.
Proceedings of the National Academy of Sciences of the United States of America
Rubinstein MP,Kovar M,Purton JF,Cho JH,Boyman O,Surh CD,Sprent J
Tue Jun 13 00:00:00 EDT 2006