Product References
Impact of broad-spectrum antibiotics on the gut-microbiota-spleen-brain axis.
Brain, behavior, & immunity - health
Wan X,Eguchi A,Sakamoto A,Fujita Y,Yang Y,Qu Y,Hatano M,Mori C,Hashimoto K
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2023
Klebsiella pneumoniae Induces Inflammatory Bowel Disease Through Caspase-11-Mediated IL18 in the Gut Epithelial Cells.
Cellular and molecular gastroenterology and hepatology
Zhang Q,Su X,Zhang C,Chen W,Wang Y,Yang X,Liu D,Zhang Y,Yang R
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Feb 23 00:00:00 EST 2023
Calcaratarin D, a labdane diterpenoid, attenuates mouse asthma via modulating alveolar macrophage function.
British journal of pharmacology
Liao W,Foo HYC,Tran TNQ,Chai CLL,Wong WSF
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
Palmitoyl transferases act as novel drug targets for pancreatic cancer.
Journal of translational medicine
Lin Z,Lv Z,Liu X,Huang K
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Apr 10 00:00:00 EDT 2023
SARS-CoV-2 spike protein promotes inflammatory cytokine activation and aggravates rheumatoid arthritis.
Cell communication and signaling : CCS
Lee AR,Woo JS,Lee SY,Lee YS,Jung J,Lee CR,Park SH,Cho ML
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Mar 02 00:00:00 EST 2023
TRAF6 controls T cell homeostasis by maintaining the equilibrium of MALT1 scaffolding and protease functions.
Frontiers in immunology
O'Neill TJ,Gewies A,Seeholzer T,Krappmann D
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Feb 13 00:00:00 EST 2023
Group 2 Innate Lymphoid Cells Protect Mice from Abdominal Aortic Aneurysm Formation via IL5 and Eosinophils.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Liu T,Deng Z,Fang W,Zhang X,Zhang S,Wang M,Luo S,Meng Z,Liu J,Sukhova GK,Li D,McKenzie ANJ,Libby P,Shi GP,Guo J
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
Complete Freund's Adjuvant Induces a Fibroblast-like Synoviocytes (FLS) Metabolic and Migratory Phenotype in Resident Fibroblasts of the Inoculated Footpad at the Earliest Stage of Adjuvant-Induced Arthritis.
Cells
González-Chávez SA,Chaparro-Barrera E,Alvarado-Jáquez MF,Cuevas-Martínez R,Ochoa-Albíztegui RE,Pacheco-Tena C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry
Wed Mar 08 00:00:00 EST 2023
Aryl hydrocarbon receptor activation ameliorates experimental colitis by modulating the tolerogenic dendritic and regulatory T cell formation.
Cell & bioscience
Cui X,Ye Z,Wang D,Yang Y,Jiao C,Ma J,Tang N,Zhang H
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat Apr 23 00:00:00 EDT 2022
IL-33 induces thymic involution-associated naive T cell aging and impairs host control of severe infection.
Nature communications
Xu L,Wei C,Chen Y,Wu Y,Shou X,Chen W,Lu D,Sun H,Li W,Yu B,Wang X,Zhang X,Yu Y,Lei Z,Tang R,Zhu J,Li Y,Lu L,Zhou H,Zhou S,Su C,Chen X
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat Nov 12 00:00:00 EST 2022
Fringe GlcNAc-transferases differentially extend O-fucose on endogenous NOTCH1 in mouse activated T cells.
The Journal of biological chemistry
Matsumoto K,Kumar V,Varshney S,Nairn AV,Ito A,Pennarubia F,Moremen KW,Stanley P,Haltiwanger RS
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2022
Breast cancer cell-derived extracellular vesicles promote CD8+ T cell exhaustion via TGF-β type II receptor signaling.
Nature communications
Xie F,Zhou X,Su P,Li H,Tu Y,Du J,Pan C,Wei X,Zheng M,Jin K,Miao L,Wang C,Meng X,van Dam H,Ten Dijke P,Zhang L,Zhou F
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2022
Black raspberry extract inhibits regulatory T-cell activity in a murine model of head and neck squamous cell carcinoma chemoprevention.
Frontiers in immunology
Ryan NM,Lamenza FF,Upadhaya P,Pracha H,Springer A,Swingler M,Siddiqui A,Oghumu S
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry
Mon Aug 29 00:00:00 EDT 2022
cDC1 coordinate innate and adaptive responses in the omentum required for T cell priming and memory.
Science immunology
Christian DA,Adams TA,Shallberg LA,Phan AT,Smith TE,Abraha M,Perry J,Ruthel G,Clark JT,Pritchard GH,Aronson LR,Gossa S,McGavern DB,Kedl RM,Hunter CA
58-0042-82 was used in Immunohistochemistry to investigate how conventional type 1 dendritic cells (cDC1s) in the peritoneal cavity's omentum coordinate innate responses to microbial challenge and provide secondary signals required for CD8+ T cell expansion and memory formation, particularly after intraperitoneal immunization with Toxoplasma gondii.
Fri Sep 30 00:00:00 EDT 2022
Clonally expanded CD8 T cells characterize amyotrophic lateral sclerosis-4.
Nature
Campisi L,Chizari S,Ho JSY,Gromova A,Arnold FJ,Mosca L,Mei X,Fstkchyan Y,Torre D,Beharry C,Garcia-Forn M,Jiménez-Alcázar M,Korobeynikov VA,Prazich J,Fayad ZA,Seldin MM,De Rubeis S,Bennett CL,Ostrow LW,Lunetta C,Squatrito M,Byun M,Shneider NA,Jiang N,La Spada AR,Marazzi I
58-0042-82 was used in Flow cytometry/Cell sorting to provide evidence of an antigen-specific CD8 T cell response in ALS4, which could be used to unravel disease mechanisms and as a potential biomarker of disease state.
Wed Jun 01 00:00:00 EDT 2022
Niche-specific MHC II and PD-L1 regulate CD4+CD8αα+ intraepithelial lymphocyte differentiation.
The Journal of experimental medicine
Moon S,Park Y,Hyeon S,Kim YM,Kim JH,Kim H,Park S,Lee KJ,Koo BK,Ha SJ,Lee SW
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Apr 05 00:00:00 EDT 2021
Niche-specific MHC II and PD-L1 regulate CD4+CD8αα+ intraepithelial lymphocyte differentiation.
The Journal of experimental medicine
Moon S,Park Y,Hyeon S,Kim YM,Kim JH,Kim H,Park S,Lee KJ,Koo BK,Ha SJ,Lee SW
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Apr 05 00:00:00 EDT 2021
Romidepsin (FK228) regulates the expression of the immune checkpoint ligand PD-L1 and suppresses cellular immune functions in colon cancer.
Cancer immunology, immunotherapy : CII
Shi Y,Fu Y,Zhang X,Zhao G,Yao Y,Guo Y,Ma G,Bai S,Li H
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Potential therapeutic effects of interleukin-35 on the differentiation of naïve T cells into Helios+Foxp3+ Tregs in clinical and experimental acute respiratory distress syndrome.
Molecular immunology
Wang C,Xie K,Li K,Lin S,Xu F
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Feline adipose tissue-derived mesenchymal stem cells pretreated with IFN-γ enhance immunomodulatory effects through the PGE₂ pathway.
Journal of veterinary science
Park SG,An JH,Li Q,Chae HK,Park SM,Lee JH,Ahn JO,Song WJ,Youn HY
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
Imbalance between T helper 1 and regulatory T cells plays a detrimental role in experimental Parkinson's disease in mice.
The Journal of international medical research
Li W,Luo Y,Xu H,Ma Q,Yao Q
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Intratumoral expression of interleukin 23 variants using oncolytic vaccinia virus elicit potent antitumor effects on multiple tumor models via tumor microenvironment modulation.
Theranostics
Chen L,Chen H,Ye J,Ge Y,Wang H,Dai E,Ren J,Liu W,Ma C,Ju S,Guo ZS,Liu Z,Bartlett DL
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Aug 04 00:00:00 EDT 2021
Tissue plasminogen activator worsens experimental autoimmune encephalomyelitis by complementary actions on lymphoid and myeloid cell responses.
Journal of neuroinflammation
Hélie P,Camacho-Toledano C,Lesec L,Seillier C,Miralles AJ,Ortega MC,Guérit S,Lebas H,Bardou I,Vila-Del Sol V,Vivien D,Le Mauff B,Clemente D,Docagne F,Toutirais O
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry
Sat Feb 20 00:00:00 EST 2021
Antitumor efficacy of a neoantigen cancer vaccine delivered by electroporation is influenced by microbiota composition.
Oncoimmunology
Lione L,Salvatori E,Petrazzuolo A,Massacci A,Maggio R,Confroti A,Compagnone M,Aurisicchio L,Ciliberto G,Palombo F
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Mar 23 00:00:00 EDT 2021
Evaluation of DNA-Launched Virus-Like Particle Vaccines in an Immune Competent Mouse Model of Chikungunya Virus Infection.
Vaccines
Rayner JO,Kim JH,Roberts RW,Wood RR,Fouty B,Solodushko V
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Apr 02 00:00:00 EDT 2021
Increased Levels of IL-16 in the Central Nervous System during Neuroinflammation Are Associated with Infiltrating Immune Cells and Resident Glial Cells.
Biology
Hridi SU,Barbour M,Wilson C,Franssen AJ,Harte T,Bushell TJ,Jiang HR
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry (Frozen)
Thu May 27 00:00:00 EDT 2021
Gut Microbiota Dysbiosis Induced by Intracerebral Hemorrhage Aggravates Neuroinflammation in Mice.
Frontiers in microbiology
Yu X,Zhou G,Shao B,Zhou H,Xu C,Yan F,Wang L,Chen G,Li J,Fu X
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed May 26 00:00:00 EDT 2021
Epithelial Mutant p53 Promotes Resistance to Anti-PD-1-Mediated Oral Cancer Immunoprevention in Carcinogen-Induced Mouse Models.
Cancers
Wang J,Hu Y,Escamilla-Rivera V,Gonzalez CL,Tang L,Wang B,El-Naggar AK,Myers JN,Caulin C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry
Tue Mar 23 00:00:00 EDT 2021
Near-infrared photoimmunotherapy targeting human-EGFR in a mouse tumor model simulating current and future clinical trials.
EBioMedicine
Okada R,Furusawa A,Vermeer DW,Inagaki F,Wakiyama H,Kato T,Nagaya T,Choyke PL,Spanos WC,Allen CT,Kobayashi H
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat May 01 00:00:00 EDT 2021
Activation of Bone Marrow Adaptive Immunity in Type 2 Diabetes: Rescue by Co-stimulation Modulator Abatacept.
Frontiers in immunology
Santopaolo M,Sullivan N,Thomas AC,Alvino VV,Nicholson LB,Gu Y,Spinetti G,Kallikourdis M,Blom A,Madeddu P
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Jul 02 00:00:00 EDT 2021
Engineered macrophages as near-infrared light activated drug vectors for chemo-photodynamic therapy of primary and bone metastatic breast cancer.
Nature communications
Huang Y,Guan Z,Dai X,Shen Y,Wei Q,Ren L,Jiang J,Xiao Z,Jiang Y,Liu D,Huang Z,Xu X,Luo Y,Zhao C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Jul 14 00:00:00 EDT 2021
Histological, behavioural and flow cytometric datasets relating to acute ischaemic stroke in young, aged and ApoE-/- mice in the presence and absence of immunomodulation with fingolimod.
Data in brief
Diaz Diaz AC,Malone K,Shearer JA,Moore AC,Waeber C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Ddb1 Is Essential for the Expansion of CD4+ Helper T Cells by Regulating Cell Cycle Progression and Cell Death.
Frontiers in immunology
Yang L,Chen W,Li L,Xiao Y,Fan S,Zhang Q,Xia T,Li M,Hong Y,Zhao T,Li Q,Liu WH,Xiao N
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Dec 20 00:00:00 EST 2021
Local Sustained GM-CSF Delivery by Genetically Engineered Encapsulated Cells Enhanced Both Cellular and Humoral SARS-CoV-2 Spike-Specific Immune Response in an Experimental Murine Spike DNA Vaccination Model.
Vaccines
Vernet R,Charrier E,Cosset E,Fièvre S,Tomasello U,Grogg J,Mach N
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon May 10 00:00:00 EDT 2021
Sequencing, cloning, and antigen binding analysis of monoclonal antibodies isolated from single mouse B cells.
STAR protocols
Viant C,Escolano A,Chen ST,Nussenzweig MC
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Jun 18 00:00:00 EDT 2021
Small-molecule antagonist of VLA-4 (GW559090) attenuated neuro-inflammation by targeting Th17 cell trafficking across the blood-retinal barrier in experimental autoimmune uveitis.
Journal of neuroinflammation
Chen YH,Eskandarpour M,Zhang X,Galatowicz G,Greenwood J,Lightman S,Calder V
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Feb 18 00:00:00 EST 2021
Analysis of T cells in mouse lymphoid tissue and blood with flow cytometry.
STAR protocols
Skordos I,Demeyer A,Beyaert R
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Mar 19 00:00:00 EDT 2021
STING activation normalizes the intraperitoneal vascular-immune microenvironment and suppresses peritoneal carcinomatosis of colon cancer.
Journal for immunotherapy of cancer
Lee SJ,Yang H,Kim WR,Lee YS,Lee WS,Kong SJ,Lee HJ,Kim JH,Cheon J,Kang B,Chon HJ,Kim C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Construction, characterization, and immunization of nanoparticles that display a diverse array of influenza HA trimers.
PloS one
Cohen AA,Yang Z,Gnanapragasam PNP,Ou S,Dam KA,Wang H,Bjorkman PJ
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Oct 15 00:00:00 EDT 2021
Serum Amyloid A Proteins Induce Pathogenic Th17 Cells and Promote Inflammatory Disease.
Cell
Lee JY,Hall JA,Kroehling L,Wu L,Najar T,Nguyen HH,Lin WY,Yeung ST,Silva HM,Li D,Hine A,Loke P,Hudesman D,Martin JC,Kenigsberg E,Merad M,Khanna KM,Littman DR
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunocytochemistry
Thu Jan 09 00:00:00 EST 2020
Protein arginine methyltransferase 5 promotes cholesterol biosynthesis-mediated Th17 responses and autoimmunity.
The Journal of clinical investigation
Webb LM,Sengupta S,Edell C,Piedra-Quintero ZL,Amici SA,Miranda JN,Bevins M,Kennemer A,Laliotis G,Tsichlis PN,Guerau-de-Arellano M
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2020
Methnaridine is an orally bioavailable, fast-killing and long-acting antimalarial agent that cures Plasmodium infections in mice.
British journal of pharmacology
Wang W,Yao J,Chen Z,Sun Y,Shi Y,Wei Y,Zhou H,Yu Y,Li S,Duan L
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2020
DKK2 blockage-mediated immunotherapy enhances anti-angiogenic therapy of Kras mutated colorectal cancer.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Hu J,Wang Z,Chen Z,Li A,Sun J,Zheng M,Wu J,Shen T,Qiao J,Li L,Li B,Wu D,Xiao Q
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Increased Tim-3 expression on TILs during treatment with the Anchored GM-CSF vaccine and anti-PD-1 antibodies is inversely correlated with response in prostate cancer.
Journal of Cancer
Zhang X,Chen H,Li G,Zhou X,Shi Y,Zou F,Chen Y,Gao J,Yang S,Wu S,Long Z
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Comparison of rapamycin and methylprednisolone for treating inflammatory muscle disease in a murine model of experimental autoimmune myositis.
Experimental and therapeutic medicine
Kang J,Feng D,Yang F,Tian X,Han W,Jia H
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Effects of CD4+ T lymphocytes from ovariectomized mice on bone marrow mesenchymal stem cell proliferation and osteogenic differentiation.
Experimental and therapeutic medicine
Shao BY,Wang L,Yu Y,Chen L,Gan N,Huang WM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2020
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
Oncolytic vaccinia virus delivering tethered IL-12 enhances antitumor effects with improved safety.
Journal for immunotherapy of cancer
Ge Y,Wang H,Ren J,Liu W,Chen L,Chen H,Ye J,Dai E,Ma C,Ju S,Guo ZS,Liu Z,Bartlett DL
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
A novel antibody targeting TIM-3 resulting in receptor internalization for cancer immunotherapy.
Antibody therapeutics
Kuang Z,Li L,Zhang P,Chen B,Wu M,Ni H,Yi S,Zou J,Liu J
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2020
Angioimmunoblastic T-cell lymphoma-like lymphadenopathy in mice transgenic for human RHOA with p.Gly17Val mutation.
Oncoimmunology
Lee GJ,Jun Y,Yoo HY,Jeon YK,Lee D,Lee S,Kim J
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry
Wed May 13 00:00:00 EDT 2020
Mechanisms of interleukin 4 mediated increase in efficacy of vaccines against opioid use disorders.
NPJ vaccines
Crouse B,Robinson C,Huseby Kelcher A,Laudenbach M,Abrahante JE,Pravetoni M
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Oct 29 00:00:00 EDT 2020
CD226 Deletion Reduces Type 1 Diabetes in the NOD Mouse by Impairing Thymocyte Development and Peripheral T Cell Activation.
Frontiers in immunology
Shapiro MR,Yeh WI,Longfield JR,Gallagher J,Infante CM,Wellford S,Posgai AL,Atkinson MA,Campbell-Thompson M,Lieberman SM,Serreze DV,Geurts AM,Chen YG,Brusko TM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed May 26 00:00:00 EDT 2021
Distinct actions of the fermented beverage kefir on host behaviour, immunity and microbiome gut-brain modules in the mouse.
Microbiome
van de Wouw M,Walsh AM,Crispie F,van Leuven L,Lyte JM,Boehme M,Clarke G,Dinan TG,Cotter PD,Cryan JF
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon May 18 00:00:00 EDT 2020
Senolytics prevent mt-DNA-induced inflammation and promote the survival of aged organs following transplantation.
Nature communications
Iske J,Seyda M,Heinbokel T,Maenosono R,Minami K,Nian Y,Quante M,Falk CS,Azuma H,Martin F,Passos JF,Niemann CU,Tchkonia T,Kirkland JL,Elkhal A,Tullius SG
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Aug 27 00:00:00 EDT 2020
Loss of testosterone impairs anti-tumor neutrophil function.
Nature communications
Markman JL,Porritt RA,Wakita D,Lane ME,Martinon D,Noval Rivas M,Luu M,Posadas EM,Crother TR,Arditi M
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Mar 31 00:00:00 EDT 2020
Positive Effects of Preventive Nutrition Supplement on Anticancer Radiotherapy in Lung Cancer Bearing Mice.
Cancers
Liu YM,Wu TH,Chiu YH,Wang H,Li TL,Hsia S,Chan YL,Wu CJ
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry
Fri Aug 28 00:00:00 EDT 2020
Cancer-Specific Loss of p53 Leads to a Modulation of Myeloid and T Cell Responses.
Cell reports
Blagih J,Zani F,Chakravarty P,Hennequart M,Pilley S,Hobor S,Hock AK,Walton JB,Morton JP,Gronroos E,Mason S,Yang M,McNeish I,Swanton C,Blyth K,Vousden KH
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Jan 14 00:00:00 EST 2020
Prodrug-Loaded Zirconium Carbide Nanosheets as a Novel Biophotonic Nanoplatform for Effective Treatment of Cancer.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Liu Q,Xie Z,Qiu M,Shim I,Yang Y,Xie S,Yang Q,Wang D,Chen S,Fan T,Ding B,Guo Z,Adah D,Yao X,Zhang Y,Wu H,Wu Z,Wei C,Wang H,Kim HS,Zou Q,Yan Q,Cai Z,Kim JS,Liu LP,Zhang H,Cao Y
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry
Tue Dec 01 00:00:00 EST 2020
STING couples with PI3K to regulate actin reorganization during BCR activation.
Science advances
Jing Y,Dai X,Yang L,Kang D,Jiang P,Li N,Cheng J,Li J,Miller H,Ren B,Gong Q,Yin W,Liu Z,Mattila PK,Ning Q,Sun J,Yu B,Liu C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry
Wed Apr 01 00:00:00 EDT 2020
Airway Microbiota-Host Interactions Regulate Secretory Leukocyte Protease Inhibitor Levels and Influence Allergic Airway Inflammation.
Cell reports
Jaeger N,McDonough RT,Rosen AL,Hernandez-Leyva A,Wilson NG,Lint MA,Russler-Germain EV,Chai JN,Bacharier LB,Hsieh CS,Kau AL
58-0042 was used in Flow cytometry/Cell sorting to investigate how chronic colonization impacts mucosal immunity and the development of allergic airway inflammation (AAI).
Tue Nov 03 00:00:00 EST 2020
A Commercial Probiotic Induces Tolerogenic and Reduces Pathogenic Responses in Experimental Autoimmune Encephalomyelitis.
Cells
Calvo-Barreiro L,Eixarch H,Ponce-Alonso M,Castillo M,Lebrón-Galán R,Mestre L,Guaza C,Clemente D,Del Campo R,Montalban X,Espejo C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Apr 07 00:00:00 EDT 2020
Axenic Caenorhabditis elegans antigen protects against development of type-1 diabetes in NOD mice.
Journal of translational autoimmunity
Jackson-Thompson BM,Torrero M,Mitre BK,Long J,Packiam M,Mitre E
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
T Cell Recruitment to the Intestinal Stem Cell Compartment Drives Immune-Mediated Intestinal Damage after Allogeneic Transplantation.
Immunity
Fu YY,Egorova A,Sobieski C,Kuttiyara J,Calafiore M,Takashima S,Clevers H,Hanash AM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunocytochemistry
Tue Jul 16 00:00:00 EDT 2019
T Cell Recruitment to the Intestinal Stem Cell Compartment Drives Immune-Mediated Intestinal Damage after Allogeneic Transplantation.
Immunity
Fu YY,Egorova A,Sobieski C,Kuttiyara J,Calafiore M,Takashima S,Clevers H,Hanash AM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunocytochemistry
Tue Jul 16 00:00:00 EDT 2019
Clonal copy-number mosaicism in autoreactive T lymphocytes in diabetic NOD mice.
Genome research
Alriyami M,Marchand L,Li Q,Du X,Olivier M,Polychronakos C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sun Dec 01 00:00:00 EST 2019
A unique mutator phenotype reveals complementary oncogenic lesions leading to acute leukemia.
JCI insight
Yin M,Baslan T,Walker RL,Zhu YJ,Freeland A,Matsukawa T,Sridharan S,Nussenzweig A,Pruitt SC,Lowe SW,Meltzer PS,Aplan PD
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Dec 05 00:00:00 EST 2019
Phosphorylation of PD-1-Y248 is a marker of PD-1-mediated inhibitory function in human T cells.
Scientific reports
Bardhan K,Aksoylar HI,Le Bourgeois T,Strauss L,Weaver JD,Delcuze B,Charest A,Patsoukis N,Boussiotis VA
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Nov 21 00:00:00 EST 2019
Mutation of an L-Type Calcium Channel Gene Leads to T Lymphocyte Dysfunction.
Frontiers in immunology
Fenninger F,Han J,Stanwood SR,Nohara LL,Arora H,Choi KB,Munro L,Pfeifer CG,Shanina I,Horwitz MS,Jefferies WA
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Oct 29 00:00:00 EDT 2020
Systemic inhibition of tissue-nonspecific alkaline phosphatase alters the brain-immune axis in experimental sepsis.
Scientific reports
Brichacek AL,Benkovic SA,Chakraborty S,Nwafor DC,Wang W,Jun S,Dakhlallah D,Geldenhuys WJ,Pinkerton AB,Millán JL,Brown CM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Dec 11 00:00:00 EST 2019
Chloroquine modulates inflammatory autoimmune responses through Nurr1 in autoimmune diseases.
Scientific reports
Park TY,Jang Y,Kim W,Shin J,Toh HT,Kim CH,Yoon HS,Leblanc P,Kim KS
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Oct 29 00:00:00 EDT 2019
Ufbp1 promotes plasma cell development and ER expansion by modulating distinct branches of UPR.
Nature communications
Zhu H,Bhatt B,Sivaprakasam S,Cai Y,Liu S,Kodeboyina SK,Patel N,Savage NM,Sharma A,Kaufman RJ,Li H,Singh N
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Mar 06 00:00:00 EST 2019
Mitophagy in Intestinal Epithelial Cells Triggers Adaptive Immunity during Tumorigenesis.
Cell
Ziegler PK,Bollrath J,Pallangyo CK,Matsutani T,Canli Ö,De Oliveira T,Diamanti MA,Müller N,Gamrekelashvili J,Putoczki T,Horst D,Mankan AK,Öner MG,Müller S,Müller-Höcker J,Kirchner T,Slotta-Huspenina J,Taketo MM,Reinheckel T,Dröse S,Larner AC,Wels WS,Ernst M,Greten TF,Arkan MC,Korn T,Wirth D,Greten FR
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Jun 28 00:00:00 EDT 2018
T-Cell-Intrinsic Receptor Interacting Protein 2 Regulates Pathogenic T Helper 17 Cell Differentiation.
Immunity
Shimada K,Porritt RA,Markman JL,O'Rourke JG,Wakita D,Noval Rivas M,Ogawa C,Kozhaya L,Martins GA,Unutmaz D,Baloh RH,Crother TR,Chen S,Arditi M
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Nov 20 00:00:00 EST 2018
TCF-1 and HEB cooperate to establish the epigenetic and transcription profiles of CD4+CD8+ thymocytes.
Nature immunology
Emmanuel AO,Arnovitz S,Haghi L,Mathur PS,Mondal S,Quandt J,Okoreeh MK,Maienschein-Cline M,Khazaie K,Dose M,Gounari F
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
Macrophage-specific MHCII expression is regulated by a remote Ciita enhancer controlled by NFAT5.
The Journal of experimental medicine
Buxadé M,Huerga Encabo H,Riera-Borrull M,Quintana-Gallardo L,López-Cotarelo P,Tellechea M,Martínez-Martínez S,Redondo JM,Martín-Caballero J,Flores JM,Bosch E,Rodríguez-Fernández JL,Aramburu J,López-Rodríguez C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Nov 05 00:00:00 EST 2018
Bryostatin-1 alleviates experimental multiple sclerosis.
Proceedings of the National Academy of Sciences of the United States of America
Kornberg MD,Smith MD,Shirazi HA,Calabresi PA,Snyder SH,Kim PM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Feb 27 00:00:00 EST 2018
Quantitative Methods for Measuring Repair Rates and Innate-Immune Cell Responses in Wounded Mouse Skin.
Frontiers in immunology
Li Z,Gothard E,Coles MC,Ambler CA
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Mar 27 00:00:00 EDT 2019
Protease-activated receptor-2 signaling through β-arrestin-2 mediates Alternaria alkaline serine protease-induced airway inflammation.
American journal of physiology. Lung cellular and molecular physiology
Yee MC,Nichols HL,Polley D,Saifeddine M,Pal K,Lee K,Wilson EH,Daines MO,Hollenberg MD,Boitano S,DeFea KA
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
Granulocyte-macrophage colony-stimulating factor neuroprotective activities in Alzheimer's disease mice.
Journal of neuroimmunology
Kiyota T,Machhi J,Lu Y,Dyavarshetty B,Nemati M,Yokoyama I,Mosley RL,Gendelman HE
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Jun 15 00:00:00 EDT 2018
Immunological effects of a low-dose cytarabine, aclarubicin and granulocyte-colony stimulating factor priming regimen on a mouse leukemia model.
Oncology letters
Chen J,Yang N,Liu H,Yao H,Wang J,Yang Y,Zhang W
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2018
Protective function of Bu Shen Huo Xue formula on the immunity of B6AF1 mice with experimental autoimmune premature ovarian failure.
Experimental and therapeutic medicine
Wang P,Lu Y,Chen S,Chen Y,Hu C,Zuo Y
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Runx-dependent and silencer-independent repression of a maturation enhancer in the Cd4 gene.
Nature communications
Kojo S,Yasmin N,Muroi S,Tenno M,Taniuchi I
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Sep 05 00:00:00 EDT 2018
Modifying the cancer-immune set point using vaccinia virus expressing re-designed interleukin-2.
Nature communications
Liu Z,Ge Y,Wang H,Ma C,Feist M,Ju S,Guo ZS,Bartlett DL
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Nov 08 00:00:00 EST 2018
Rap1 deficiency-provoked paracrine dysfunction impairs immunosuppressive potency of mesenchymal stem cells in allograft rejection of heart transplantation.
Cell death & disease
Ding Y,Liang X,Zhang Y,Yi L,Shum HC,Chen Q,Chan BP,Fan H,Liu Z,Tergaonkar V,Qi Z,Tse HF,Lian Q
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Mar 07 00:00:00 EST 2018
Ubiquitination of tumor suppressor PML regulates prometastatic and immunosuppressive tumor microenvironment.
The Journal of clinical investigation
Wang YT,Chen J,Chang CW,Jen J,Huang TY,Chen CM,Shen R,Liang SY,Cheng IC,Yang SC,Lai WW,Cheng KH,Hsieh TS,Lai MZ,Cheng HC,Wang YC,Chen RH
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Excessive expression of miR-27 impairs Treg-mediated immunological tolerance.
The Journal of clinical investigation
Cruz LO,Hashemifar SS,Wu CJ,Cho S,Nguyen DT,Lin LL,Khan AA,Lu LF
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
IL-36 Signaling Facilitates Activation of the NLRP3 Inflammasome and IL-23/IL-17 Axis in Renal Inflammation and Fibrosis.
Journal of the American Society of Nephrology : JASN
Chi HH,Hua KF,Lin YC,Chu CL,Hsieh CY,Hsu YJ,Ka SM,Tsai YL,Liu FC,Chen A
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2017
IL-2/Anti-IL-2 Complex Attenuates Inflammation and BBB Disruption in Mice Subjected to Traumatic Brain Injury.
Frontiers in neurology
Gao W,Li F,Zhou Z,Xu X,Wu Y,Zhou S,Yin D,Sun D,Xiong J,Jiang R,Zhang J
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Discrimination of the heterogeneity of bone marrow‑derived dendritic cells.
Molecular medicine reports
Wang J,Dai X,Hsu C,Ming C,He Y,Zhang J,Wei L,Zhou P,Wang CY,Yang J,Gong N
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Nov 01 00:00:00 EDT 2017
PD-1 Blockade Prevents the Development and Progression of Carcinogen-Induced Oral Premalignant Lesions.
Cancer prevention research (Philadelphia, Pa.)
Wang J,Xie T,Wang B,William WN,Heymach JV,El-Naggar AK,Myers JN,Caulin C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Immunohistochemistry
Fri Dec 01 00:00:00 EST 2017
Immunological effects of vaccines combined with granulocyte colony-stimulating factor on a murine WEHI-3 leukemia model.
Oncology letters
Chen J,Zhang M,Zhou F,Wang J,Niu B,Zhang W
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Effect of follicular helper T cells on the pathogenesis of asthma.
Experimental and therapeutic medicine
Ma X,Sun Z,Zhai P,Yu W,Wang T,Li F,Ding J
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
IL-7-dependent STAT1 activation limits homeostatic CD4+ T cell expansion.
JCI insight
Le Saout C,Luckey MA,Villarino AV,Smith M,Hasley RB,Myers TG,Imamichi H,Park JH,O'Shea JJ,Lane HC,Catalfamo M
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Nov 16 00:00:00 EST 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Batf3-dependent CD8α+ Dendritic Cells Aggravates Atherosclerosis via Th1 Cell Induction and Enhanced CCL5 Expression in Plaque Macrophages.
EBioMedicine
Li Y,Liu X,Duan W,Tian H,Zhu G,He H,Yao S,Yi S,Song W,Tang H
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Isolation of cell type-specific apoptotic bodies by fluorescence-activated cell sorting.
Scientific reports
Atkin-Smith GK,Paone S,Zanker DJ,Duan M,Phan TK,Chen W,Hulett MD,Poon IK
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Jan 06 00:00:00 EST 2017
CD69 controls the uptake of L-tryptophan through LAT1-CD98 and AhR-dependent secretion of IL-22 in psoriasis.
Nature immunology
Cibrian D,Saiz ML,de la Fuente H,Sánchez-Díaz R,Moreno-Gonzalo O,Jorge I,Ferrarini A,Vázquez J,Punzón C,Fresno M,Vicente-Manzanares M,Daudén E,Fernández-Salguero PM,Martín P,Sánchez-Madrid F
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Oligodendrocyte death results in immune-mediated CNS demyelination.
Nature neuroscience
Traka M,Podojil JR,McCarthy DP,Miller SD,Popko B
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
STAT3/5-Dependent IL9 Overexpression Contributes to Neoplastic Cell Survival in Mycosis Fungoides.
Clinical cancer research : an official journal of the American Association for Cancer Research
Vieyra-Garcia PA,Wei T,Naym DG,Fredholm S,Fink-Puches R,Cerroni L,Odum N,O'Malley JT,Gniadecki R,Wolf P
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
Genomic and immunologic factors associated with viral pathogenesis in a lethal EV71 infected neonatal mouse model.
Molecular medicine reports
Yue Y,Li P,Song N,Li B,Li Z,Guo Y,Zhang W,Wei MQ,Gai Z,Meng H,Wang J,Qin L
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
Deletion of Wiskott-Aldrich syndrome protein triggers Rac2 activity and increased cross-presentation by dendritic cells.
Nature communications
Baptista MA,Keszei M,Oliveira M,Sunahara KK,Andersson J,Dahlberg CI,Worth AJ,Liedén A,Kuo IC,Wallin RP,Snapper SB,Eidsmo L,Scheynius A,Karlsson MC,Bouma G,Burns SO,Forsell MN,Thrasher AJ,Nylén S,Westerberg LS
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Jul 18 00:00:00 EDT 2016
Affinity and dose of TCR engagement yield proportional enhancer and gene activity in CD4+ T cells.
eLife
Allison KA,Sajti E,Collier JG,Gosselin D,Troutman TD,Stone EL,Hedrick SM,Glass CK
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Jul 04 00:00:00 EDT 2016
Sex bias in CNS autoimmune disease mediated by androgen control of autoimmune regulator.
Nature communications
Zhu ML,Bakhru P,Conley B,Nelson JS,Free M,Martin A,Starmer J,Wilson EM,Su MA
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Apr 13 00:00:00 EDT 2016
Irradiation of necrotic cancer cells, employed for pulsing dendritic cells (DCs), potentiates DC vaccine-induced antitumor immunity against high-grade glioma.
Oncoimmunology
Vandenberk L,Garg AD,Verschuere T,Koks C,Belmans J,Beullens M,Agostinis P,De Vleeschouwer S,Van Gool SW
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2016
Exploring the induction of preproinsulin-specific Foxp3(+) CD4(+) Treg cells that inhibit CD8(+) T cell-mediated autoimmune diabetes by DNA vaccination.
Scientific reports
Stifter K,Schuster C,Schlosser M,Boehm BO,Schirmbeck R
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Jul 11 00:00:00 EDT 2016
Atypical chemokine receptor 1 deficiency reduces atherogenesis in ApoE-knockout mice.
Cardiovascular research
Wan W,Liu Q,Lionakis MS,Marino AP,Anderson SA,Swamydas M,Murphy PM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Single-Dose CpG Immunization Protects Against a Heterosubtypic Challenge and Generates Antigen-Specific Memory T Cells.
Frontiers in immunology
Vogel AJ,Brown DM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Jul 10 00:00:00 EDT 2015
A tumor lysate is an effective vaccine antigen for the stimulation of CD4(+) T-cell function and subsequent induction of antitumor immunity mediated by CD8(+) T cells.
Cancer biology & therapy
Kawahara M,Takaku H
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Sep 12 00:00:00 EDT 2016
Immunogenicity and protective efficacy of DMT liposome-adjuvanted tuberculosis subunit CTT3H vaccine.
Human vaccines & immunotherapeutics
Teng X,Tian M,Li J,Tan S,Yuan X,Yu Q,Jing Y,Zhang Z,Yue T,Zhou L,Fan X
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Mar 11 00:00:00 EST 2016
Enhanced T-cell activation and differentiation in lymphocytes from transgenic mice expressing ubiquitination-resistant 2KR LAT molecules.
Gene therapy
Rodriguez-Peña AB,Gomez-Rodriguez J,Kortum RL,Palmer DC,Yu Z,Guittard GC,Wohlfert EA,Silver PB,Misplon JA,Sommers CL,Feigenbaum L,Epstein SL,Caspi RR,Belkaid Y,Restifo NP,Samelson LE,Balagopalan L
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
dNP2 is a blood-brain barrier-permeable peptide enabling ctCTLA-4 protein delivery to ameliorate experimental autoimmune encephalomyelitis.
Nature communications
Lim S,Kim WJ,Kim YH,Lee S,Koo JH,Lee JA,Yoon H,Kim DH,Park HJ,Kim HM,Lee HG,Yun Kim J,Lee JU,Hun Shin J,Kyun Kim L,Doh J,Kim H,Lee SK,Bothwell ALM,Suh M,Choi JM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Sep 15 00:00:00 EDT 2015
dNP2 is a blood-brain barrier-permeable peptide enabling ctCTLA-4 protein delivery to ameliorate experimental autoimmune encephalomyelitis.
Nature communications
Lim S,Kim WJ,Kim YH,Lee S,Koo JH,Lee JA,Yoon H,Kim DH,Park HJ,Kim HM,Lee HG,Yun Kim J,Lee JU,Hun Shin J,Kyun Kim L,Doh J,Kim H,Lee SK,Bothwell ALM,Suh M,Choi JM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Sep 15 00:00:00 EDT 2015
Recombinant adeno-vaccine expressing enterovirus 71-like particles against hand, foot, and mouth disease.
PLoS neglected tropical diseases
Tsou YL,Lin YW,Shao HY,Yu SL,Wu SR,Lin HY,Liu CC,Huang C,Chong P,Chow YH
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Near-infrared photoactivatable control of Ca(2+) signaling and optogenetic immunomodulation.
eLife
He L,Zhang Y,Ma G,Tan P,Li Z,Zang S,Wu X,Jing J,Fang S,Zhou L,Wang Y,Huang Y,Hogan PG,Han G,Zhou Y
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2015
TLR2 engagement on CD4(+) T cells enhances effector functions and protective responses to Mycobacterium tuberculosis.
European journal of immunology
Reba SM,Li Q,Onwuzulike S,Ding X,Karim AF,Hernandez Y,Fulton SA,Harding CV,Lancioni CL,Nagy N,Rodriguez ME,Wearsch PA,Rojas RE
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu May 01 00:00:00 EDT 2014
Leukocyte population dynamics and detection of IL-9 as a major cytokine at the mouse fetal-maternal interface.
PloS one
Habbeddine M,Verbeke P,Karaz S,Bobé P,Kanellopoulos-Langevin C
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Jun 10 00:00:00 EDT 2015
The efficacy of tumor debulking surgery is improved by adjuvant immunotherapy using imiquimod and anti-CD40.
BMC cancer
Khong A,Cleaver AL,Fahmi Alatas M,Wylie BC,Connor T,Fisher SA,Broomfield S,Lesterhuis WJ,Currie AJ,Lake RA,Robinson BW
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Wed Dec 17 00:00:00 EST 2014
P2X7 receptor modulates inflammatory and functional pulmonary changes induced by silica.
PloS one
Monção-Ribeiro LC,Faffe DS,Santana PT,Vieira FS,da Graça CL,Marques-da-Silva C,Machado MN,Caruso-Neves C,Zin WA,Borojevic R,Takiya CM,Coutinho-Silva R
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2015
Genetic deletion of chemokine receptor Ccr7 exacerbates atherogenesis in ApoE-deficient mice.
Cardiovascular research
Wan W,Lionakis MS,Liu Q,Roffê E,Murphy PM
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
Characterization of two distinct lymphoproliferative diseases caused by ectopic expression of the Notch ligand DLL4 on T cells.
PloS one
Xiong H,Maraver A,Latkowski JA,Henderson T,Schlessinger K,Ding Y,Shen J,Tadokoro CE,Lafaille JJ
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Feb 23 00:00:00 EST 2015
Characterization of two distinct lymphoproliferative diseases caused by ectopic expression of the Notch ligand DLL4 on T cells.
PloS one
Xiong H,Maraver A,Latkowski JA,Henderson T,Schlessinger K,Ding Y,Shen J,Tadokoro CE,Lafaille JJ
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Mon Feb 23 00:00:00 EST 2015
Nod2 activates NF-kB in CD4+ T cells but its expression is dispensable for T cell-induced colitis.
PloS one
Zanello G,Goethel A,Forster K,Geddes K,Philpott DJ,Croitoru K
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Thu Jul 31 00:00:00 EDT 2014
Evaluation of Lassa virus vaccine immunogenicity in a CBA/J-ML29 mouse model.
Vaccine
Goicochea MA,Zapata JC,Bryant J,Davis H,Salvato MS,Lukashevich IS
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Feb 14 00:00:00 EST 2012
The stalk domain and the glycosylation status of the activating natural killer cell receptor NKp30 are important for ligand binding.
The Journal of biological chemistry
Hartmann J,Tran TV,Kaudeer J,Oberle K,Herrmann J,Quagliano I,Abel T,Cohnen A,Gatterdam V,Jacobs A,Wollscheid B,Tampé R,Watzl C,Diefenbach A,Koch J
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Sep 07 00:00:00 EDT 2012
SH2D2A modulates T cell mediated protection to a B cell derived tumor in transgenic mice.
PloS one
Berge T,Grønningsæter IH,Lorvik KB,Abrahamsen G,Granum S,Sundvold-Gjerstad V,Corthay A,Bogen B,Spurkland A
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Tue Aug 26 00:00:00 EDT 2014
Nuclear export of the NF-κB inhibitor IκBα is required for proper B cell and secondary lymphoid tissue formation.
Immunity
Wuerzberger-Davis SM,Chen Y,Yang DT,Kearns JD,Bates PW,Lynch C,Ladell NC,Yu M,Podd A,Zeng H,Huang TT,Wen R,Hoffmann A,Wang D,Miyamoto S
Published figure using CD4 monoclonal antibody (Product # 58-0042-82) in Flow Cytometry
Fri Feb 25 00:00:00 EST 2011