Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD41 Antibodies

          Invitrogen

          CD41a Monoclonal Antibody (eBioMWReg30 (MWReg30)), FITC, eBioscience™

          Advanced Verification
          68 Published Figures
          76 References
          View all (103) CD41 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD41a Monoclonal Antibody (eBioMWReg30 (MWReg30)), FITC, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (68)
          • Advanced Verification (1)
          CD41a Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD41a Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 70

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD41a Antibody (11-0411-82) in Flow

          Staining of C57BL/6 platelets with Anti-Mouse/Rat CD61 (Integrin beta 3) PE (Product # 12-0611-82) and 0.06 µg of Rat IgG1 K Isotype Control FITC (Product # 11-4301-82) (left) or 0.06 µg of Anti-Mouse CD41 FITC (right). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry, Peptide array (Flow, ARRAY)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD41a Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Flow Cytometry (Flow)
          CD41a Antibody in Immunocytochemistry (ICC/IF)
          CD41a Antibody in Immunocytochemistry (ICC/IF)
          CD41a Antibody in Immunocytochemistry (ICC/IF)
          CD41a Antibody in Immunocytochemistry (ICC/IF)
          CD41a Antibody in Immunohistochemistry (IHC)
          CD41a Antibody in Immunohistochemistry (IHC)
          CD41a Antibody in Immunohistochemistry (IHC)
          CD41a Antibody in Western Blot (WB)
          CD41a Antibody in Western Blot (WB)
          CD41a Antibody

          Product Details

          11-0411-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 7 publications 7 publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 58 publications 58 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Peptide Array (Array)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Hamster, Human, Mouse

          Host/Isotype

          Rat / IgG1, kappa

          Recommended Isotype Control

          Rat IgG1 kappa Isotype Control (eBRG1), FITC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioMWReg30 (MWReg30)

          Conjugate

          FITC FITC FITC
          • Unconjugated
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • eFluor 450
          • Functional Grade
          • NovaFluor Blue 510
          • NovaFluor Blue 555
          • NovaFluor Blue 660-120S
          • NovaFluor Yellow 570
          • NovaFluor Yellow 660
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_763481

          Product Specific Information

          Description: The eBioMWReg30 monoclonal antibody reacts with mouse CD41 (fibrinogen receptor, gpIIb, integrin alpha Iib). While initially thought to be expressed exclusively on the surface of platelets and megakaryocytes, it has been demonstrated that CD41 is also expressed on hematopoietic progenitors in the embryo, fetus and adult. CD41 associates with CD61 (gpIIIa, integrin beta III) to form a receptor which plays a major role in platelet function, including binding of several adhesion molecules such as fibrinogen, fibronectin and vitronectin.

          Recently, the SLAM-family markers, CD48 and CD150 have been used to reliably identify hematopoietic stem cells (HSC). Specifically, it was found that CD150+CD48- bone marrow cells were highly efficient in their ability to confer long-term multi-lineage reconstitution in irradiated mice. Furthermore, the efficiency of reconstitution was enhanced when HSCs were further enriched through the exclusion of CD41+ cells. Thus, the use of CD150+CD48-CD41- as an expression profile efficiently identifies hematopoietic stem cells.

          Applications Reported: This eBioMWReg30 (MWReg30) antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBioMWReg30 (MWReg30) antibody has been tested by flow cytometric analysis of mouse platelets. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD41, also known as platelet glycoprotein IIb or ITGA2B, is a protein composed of two subunits: a 120 kDa alpha subunit and a 23 kDa beta subunit. It interacts with CD61 (gpIIIa, integrin beta III) in the presence of calcium to form a functional adhesive protein receptor. Initially thought to be expressed exclusively on platelets and megakaryocytes, CD41 is also found on hematopoietic progenitors in the embryo, fetus, and adult, indicating its role in early stages of hematopoietic differentiation. CD41 plays a crucial role in platelet function and blood coagulation by mediating platelet aggregation. Upon blood vessel damage, CD41 binds to various adhesion molecules, including von Willebrand factor, fibrinogen, fibronectin, and vitronectin, facilitating platelet adhesion and aggregation. This receptor is essential for platelet function, contributing to the binding of several adhesion molecules and playing a significant role in hemostasis. Diseases associated with CD41 dysfunction include Glanzmann Thrombasthenia and Platelet type-16 bleeding disorder, highlighting its importance in normal platelet function and blood coagulation processes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: alpha IIb; CD41; form 1; form 2; GP IIb; GPalpha IIb; GPIIb; Integrin alpha-IIb; platelet glycoprotein IIb of IIb/II Ia complex; Platelet membrane glycoprotein IIb

          View more View less

          Gene Aliases: AI172977; alphaIIb; CD41; CD41B; GpIIb; Itga2b

          View more View less

          UniProt ID: (Mouse) Q9QUM0

          View more View less

          Entrez Gene ID: (Mouse) 16399

          View more View less

          Function(s)
          identical protein binding metal ion binding extracellular matrix binding fibrinogen binding integrin
          Process(es)
          positive regulation of leukocyte migration cell adhesion cell-matrix adhesion integrin-mediated signaling pathway platelet aggregation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-vvlg5:80/100.66.78.195:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline