Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
0.1 µg/test | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Host/Isotype |
Rat / IgG2a, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
RA3-6B2 |
Conjugate |
NovaFluor™ Yellow 570
|
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Concentration |
0.1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.2 |
Contains |
0.09% sodium azide |
Storage conditions |
4° C, store in dark, DO NOT FREEZE! |
Description: The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220 kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen.
Applications Reported: This RA3-6B2 antibody has been reported for use in flow cytometric analysis.
Our internal testing shows that NovaFluor Yellow 570 non-specifically stains B cells in SJL mice. Non-specific staining has not been observed in BALB/c or C57BL/6 mice. Other strains have not been tested. See the Antibody Testing Data for an example of this strain-dependent difference.
Applications Tested: This RA3-6B2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Each NovaFluor conjugate or kit is shipped with CellBlox Blocking Buffer. Use this buffer whenever staining with NovaFluor conjugates, including single color compensation controls using cells [link to CellBlox PDP or protocol showing use]. Use 5 µL of CellBlox Blocking Buffer per stained cell sample containing 10^3 to 10^8 cells.
Excitation: 568 nm; Emission: 568 nm; Laser: 561 nm (Yellow) Laser
NovaFluor conjugates are based on Phiton™ technology utilizing novel nucleic acid dye structures that allow for engineered fluorescent signatures with consideration for spillover and spread impacts. Learn more
CD45R (PTPRC) is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45 contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. CD45 is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists in multiple isoforms as a result of alternative splicing, and these isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. Further, these CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. In addition to its role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. Diseases associated with CD45 dysfunction include Severe Combined Immunodeficiency (T-cell negative, B-Cell Negative) and Hepatitis C.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: CD45; CD45 antigen; L-CA; Leukocyte common antigen; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen
Gene Aliases: B220; CD45; CD45R; GP180; L-CA; LCA; loc; Ly-5; LY5; Lyt-4; PTPRC; T200
UniProt ID: (Human) P08575, (Mouse) Q78EF1
Entrez Gene ID: (Human) 5788, (Mouse) 19264
Molecular Function:
protein phosphatase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support