Product References
The matrix microenvironment influences but does not dominate tissue-specific stem cell lineage differentiation.
Materials today. Bio
Pei YA,Mikaeiliagah E,Wang B,Zhang X,Pei M
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2023
Exosomal CD73 from serum of patients with melanoma suppresses lymphocyte functions and is associated with therapy resistance to anti-PD-1 agents.
Journal for immunotherapy of cancer
Turiello R,Capone M,Morretta E,Monti MC,Madonna G,Azzaro R,Del Gaudio P,Simeone E,Sorrentino A,Ascierto PA,Morello S
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2022
Induced Neurodifferentiation of hBM-MSCs through Activation of the ERK/CREB Pathway via Pulsed Electromagnetic Fields and Physical Stimulation Promotes Neurogenesis in Cerebral Ischemic Models.
International journal of molecular sciences
Park HJ,Choi JH,Nam MH,Seo YK
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Fri Jan 21 00:00:00 EST 2022
Reprogramming bone progenitor identity and potency through control of collagen density and oxygen tension.
iScience
Al Hosni R,Bozec L,Roberts SJ,Cheema U
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Fri Apr 15 00:00:00 EDT 2022
Human placental mesenchymal stem cells regulate inflammation via the NF‑κB signaling pathway.
Experimental and therapeutic medicine
Liu Y,Zhang X,Hu Y,Kang M,Wu Y,Wang Y,Deng C
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
hUCMSCs carrying exenatide prevent T1DM by improving intestinal microflora composition and islet tissue damage repair.
Molecular medicine (Cambridge, Mass.)
Wang W,Wang Y,Chi J,Tan X,Hu J,Ma X,Sun X,Che K,Lv W,Wang Y
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Tue Dec 13 00:00:00 EST 2022
Mechanical Compression by Simulating Orthodontic Tooth Movement in an In Vitro Model Modulates Phosphorylation of AKT and MAPKs via TLR4 in Human Periodontal Ligament Cells.
International journal of molecular sciences
Roth CE,Craveiro RB,Niederau C,Malyaran H,Neuss S,Jankowski J,Wolf M
17-0739-42 was used in Flow cytometry/Cell sorting to better understand the cell signaling downstream of key molecules involved in the process of sterile inflammation via TLR4.
Fri Jul 22 00:00:00 EDT 2022
Mesenchymal stem cells transfer mitochondria to allogeneic Tregs in an HLA-dependent manner improving their immunosuppressive activity.
Nature communications
Piekarska K,Urban-Wójciuk Z,Kurkowiak M,Pelikant-Małecka I,Schumacher A,Sakowska J,Spodnik JH,Arcimowicz Ł,Zielińska H,Tymoniuk B,Renkielska A,Siebert J,Słomińska E,Trzonkowski P,Hupp T,Marek-Trzonkowska NM
17-0739-42 was used in Flow Cytometry to demonstrate that both direct and indirect contact with allogeneic ASCs improves therapeutic potential of Tregs (Regulatory T cells).
Mon Feb 14 00:00:00 EST 2022
Single-cell RNA landscape of the osteoimmunology microenvironment in periodontitis.
Theranostics
Chen Y,Wang H,Yang Q,Zhao W,Chen Y,Ni Q,Li W,Shi J,Zhang W,Li L,Xu Y,Zhang H,Miao D,Xing L,Sun W
17-0739-42 was used in Flow Cytometry to uncover striking spatiotemporal dynamics in gene expression, population composition, and cell-cell interactions during periodontitis progression.
Thu Apr 21 00:00:00 EDT 2022
The Composition of Adipose-Derived Regenerative Cells Isolated from Lipoaspirate Using a Point of Care System Does Not Depend on the Subject's Individual Age, Sex, Body Mass Index and Ethnicity.
Cells
Schmitz C,Alt C,Azares AR,Pearce DA,Facile TR,Furia JP,Maffulli N,Huang C,Alt EU
17-0739-42 was used in Flow cytometry/Cell sorting to show that key characteristics of unmodified, autologous, adipose-derived regenerative cells (UA-ADRCs) can be independent of the subject's age, sex, BMI and ethnicity.
Wed Dec 21 00:00:00 EST 2022
Matrix reverses immortalization-mediated stem cell fate determination.
Biomaterials
Wang Y,Hu G,Hill RC,Dzieciatkowska M,Hansen KC,Zhang XB,Yan Z,Pei M
17-0739-42 was used in Flow Cytometry to provide a promising model for in-depth investigation of the matrix protein influence on surrounding stem cell differentiation.
Fri Jan 01 00:00:00 EST 2021
VAP-PLGA microspheres (VAP-PLGA) promote adipose-derived stem cells (ADSCs)-induced wound healing in chronic skin ulcers in mice via PI3K/Akt/HIF-1α pathway.
Bioengineered
Jiang W,Zhang J,Zhang X,Fan C,Huang J
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Wed Dec 01 00:00:00 EST 2021
Peptide 11R‑VIVIT promotes fracture healing in osteoporotic rats.
International journal of molecular medicine
Hou C,Wang X,Jiang W,Bian Z,Zhu L,Li M
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
Bcl-xL mutant promotes cartilage differentiation of BMSCs by upregulating TGF-β/BMP expression levels.
Experimental and therapeutic medicine
Xiao K,Yang L,Xie W,Gao X,Huang R,Xie M
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Tue Aug 31 00:00:00 EDT 2021
In Vitro Anti-cancer Activity of Adipose-Derived Mesenchymal Stem Cells Increased after Infection with Oncolytic Reovirus.
Advanced pharmaceutical bulletin
Babaei A,Bannazadeh Baghi H,Nezhadi A,Jamalpoor Z
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
In vitro differentiation of human embryonic stem cells to hemogenic endothelium and blood progenitors via embryoid body formation.
STAR protocols
Garcia-Alegria E,Potts B,Menegatti S,Kouskoff V
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Fri Mar 19 00:00:00 EDT 2021
Therapeutic potential of human umbilical cord mesenchymal stem cells on aortic atherosclerotic plaque in a high-fat diet rabbit model.
Stem cell research & therapy
Li Y,Shi G,Han Y,Shang H,Li H,Liang W,Zhao W,Bai L,Qin C
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu Jul 15 00:00:00 EDT 2021
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Tue Oct 06 00:00:00 EDT 2020
Vasculogenesis from Human Dental Pulp Stem Cells Grown in Matrigel with Fully Defined Serum-Free Culture Media.
Biomedicines
Luzuriaga J,Irurzun J,Irastorza I,Unda F,Ibarretxe G,Pineda JR
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Mon Nov 09 00:00:00 EST 2020
Vasculogenesis from Human Dental Pulp Stem Cells Grown in Matrigel with Fully Defined Serum-Free Culture Media.
Biomedicines
Luzuriaga J,Irurzun J,Irastorza I,Unda F,Ibarretxe G,Pineda JR
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Mon Nov 09 00:00:00 EST 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Fri Apr 03 00:00:00 EDT 2020
IGF-1 enhances BMSC viability, migration, and anti-apoptosis in myocardial infarction via secreted frizzled-related protein 2 pathway.
Stem cell research & therapy
Lin M,Liu X,Zheng H,Huang X,Wu Y,Huang A,Zhu H,Hu Y,Mai W,Huang Y
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu Jan 09 00:00:00 EST 2020
Chondrogenic Differentiation from Induced Pluripotent Stem Cells Using Non-Viral Minicircle Vectors.
Cells
Rim YA,Nam Y,Park N,Jung H,Lee K,Lee J,Ju JH
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Prospective isolation of human fibroadipogenic progenitors with CD73.
Heliyon
Goloviznina NA,Xie N,Dandapat A,Iaizzo PA,Kyba M
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Prospective isolation of human fibroadipogenic progenitors with CD73.
Heliyon
Goloviznina NA,Xie N,Dandapat A,Iaizzo PA,Kyba M
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Circulating Exosomes Inhibit B Cell Proliferation and Activity.
Cancers
Schroeder JC,Puntigam L,Hofmann L,Jeske SS,Beccard IJ,Doescher J,Laban S,Hoffmann TK,Brunner C,Theodoraki MN,Schuler PJ
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Wed Jul 29 00:00:00 EDT 2020
Impact of Fibronectin Knockout on Proliferation and Differentiation of Human Infrapatellar Fat Pad-Derived Stem Cells.
Frontiers in bioengineering and biotechnology
Wang Y,Fu Y,Yan Z,Zhang XB,Pei M
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Wed Dec 14 00:00:00 EST 2022
Modulation of proliferation and differentiation of gingiva‑derived mesenchymal stem cells by concentrated growth factors: Potential implications in tissue engineering for dental regeneration and repair.
International journal of molecular medicine
Chen X,Chen Y,Hou Y,Song P,Zhou M,Nie M,Liu X
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
Therapeutic Effects of Human Urine-Derived Stem Cells in a Rat Model of Cisplatin-Induced Acute Kidney Injury In Vivo and In Vitro.
Stem cells international
Sun B,Luo X,Yang C,Liu P,Yang Y,Dong X,Yang Z,Xu J,Zhang Y,Li L
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu May 13 00:00:00 EDT 2021
Sodium Selenite Improves The Therapeutic Effect Of BMSCs Via Promoting The Proliferation And Differentiation, Thereby Promoting The Hematopoietic Factors.
OncoTargets and therapy
Yan D,Tang B,Yan L,Zhang L,Miao M,Chen X,Sui G,Zhang Q,Liu D,Wang H
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Tue Apr 12 00:00:00 EDT 2022
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu May 02 00:00:00 EDT 2019
Isolation and Culture of Human Stem Cells from Apical Papilla under Low Oxygen Concentration Highlight Original Properties.
Cells
Rémy M,Ferraro F,Le Salver P,Rey S,Genot E,Djavaheri-Mergny M,Thébaud N,Boiziau C,Boeuf H
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu Nov 21 00:00:00 EST 2019
Better therapeutic potential of bone marrow-derived mesenchymal stem cells compared with chorionic villi-derived mesenchymal stem cells in airway injury model.
Regenerative medicine
Ji S,Wu C,Tong L,Wang L,Zhou J,Chen C,Song Y
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
BMP14 induces tenogenic differentiation of bone marrow mesenchymal stem cells in vitro.
Experimental and therapeutic medicine
Wang D,Jiang X,Lu A,Tu M,Huang W,Huang P
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
The influence of chemotherapy on adenosine-producing B cells in patients with head and neck squamous cell carcinoma.
Oncotarget
Ziebart A,Huber U,Jeske S,Laban S,Doescher J,Hoffmann TK,Brunner C,Jackson EK,Schuler PJ
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Fri Jan 19 00:00:00 EST 2018
Low-dose interleukin-2 promotes STAT-5 phosphorylation, Treg survival and CTLA-4-dependent function in autoimmune liver diseases.
Clinical and experimental immunology
Jeffery HC,Jeffery LE,Lutz P,Corrigan M,Webb GJ,Hirschfield GM,Adams DH,Oo YH
17-0739 was used in Flow cytometry/Cell sorting to establish the potential of interleukin-2 to enhance regulatory T cell therapy in autoimmune liver disease.
Thu Jun 01 00:00:00 EDT 2017
A high-yield isolation and enrichment strategy for human lung microvascular endothelial cells.
Pulmonary circulation
Gaskill C,Majka SM
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
A high-yield isolation and enrichment strategy for human lung microvascular endothelial cells.
Pulmonary circulation
Gaskill C,Majka SM
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu Mar 09 00:00:00 EST 2017
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Tue Jan 19 00:00:00 EST 2016
NR2F2 regulates bone marrow-derived mesenchymal stem cell-promoted proliferation of Reh cells.
Molecular medicine reports
Zhu N,Wang H,Wei J,Wang B,Shan W,Lai X,Zhao Y,Yu J,Huang H
17-0739 was used in Flow cytometry/Cell sorting to demonstrate NR2F2 mediates bone marrow-derived mesenchymal stem cell induced proliferation of Reh cells via regulation of vascular endothelial growth factor A.
Mon Aug 01 00:00:00 EDT 2016
Transendothelial migration of human umbilical mesenchymal stem cells across uterine endothelial monolayers: Junctional dynamics and putative mechanisms.
Placenta
Ebrahim NA,Leach L
17-0739 was used in Flow cytometry/Cell sorting to study the transmigration of foetal stem cells across maternal endothelial barriers, and investigate whether they can subsequently influence maternal endothelial integrity.
Thu Dec 01 00:00:00 EST 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Spontaneous Differentiation of Human Mesenchymal Stem Cells on Poly-Lactic-Co-Glycolic Acid Nano-Fiber Scaffold.
PloS one
Sonomoto K,Yamaoka K,Kaneko H,Yamagata K,Sakata K,Zhang X,Kondo M,Zenke Y,Sabanai K,Nakayamada S,Sakai A,Tanaka Y
17-0739 was used in Flow cytometry/Cell sorting to investigate the combination of MSCs and NFs as a therapeutic technique for the repair of joints affected by bone and cartilage destruction.
Thu Sep 01 00:00:00 EDT 2016
Human Gingival Integration-Free iPSCs; a Source for MSC-Like Cells.
International journal of molecular sciences
Umezaki Y,Hashimoto Y,Nishishita N,Kawamata S,Baba S
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Mon Jun 15 00:00:00 EDT 2015
Epithelial cell differentiation of human mesenchymal stromal cells in decellularized lung scaffolds.
Tissue engineering. Part A
Mendez JJ,Ghaedi M,Steinbacher D,Niklason LE
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Sun Jun 01 00:00:00 EDT 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu Dec 11 00:00:00 EST 2014
FOXF1 mediates mesenchymal stem cell fusion-induced reprogramming of lung cancer cells.
Oncotarget
Wei HJ,Nickoloff JA,Chen WH,Liu HY,Lo WC,Chang YT,Yang PC,Wu CW,Williams DF,Gelovani JG,Deng WP
17-0739 was used in Flow cytometry/Cell sorting to study the effect of fusion of mesenchymal stem cells with lung cancer cells, showing that fusion is mediated by FOXF1 and leads to a more benign state due to complementation of tumorigenic defects such as the restoration of p21 function.
Wed Oct 15 00:00:00 EDT 2014
Myoblasts derived from normal hESCs and dystrophic hiPSCs efficiently fuse with existing muscle fibers following transplantation.
Molecular therapy : the journal of the American Society of Gene Therapy
Goudenege S,Lebel C,Huot NB,Dufour C,Fujii I,Gekas J,Rousseau J,Tremblay JP
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu Nov 01 00:00:00 EDT 2012
Myoblasts derived from normal hESCs and dystrophic hiPSCs efficiently fuse with existing muscle fibers following transplantation.
Molecular therapy : the journal of the American Society of Gene Therapy
Goudenege S,Lebel C,Huot NB,Dufour C,Fujii I,Gekas J,Rousseau J,Tremblay JP
Published figure using CD73 monoclonal antibody (Product # 17-0739-42) in Flow Cytometry
Thu Nov 01 00:00:00 EDT 2012