Product References
Spontaneous immunomodulation and regulation of angiogenesis and osteogenesis by Sr/Cu-borosilicate glass (BSG) bone cement to repair critical bone defects.
Bioactive materials
Li S,Zhang L,Liu C,Kim J,Su K,Chen T,Zhao L,Lu X,Zhang H,Cui Y,Cui X,Yuan F,Pan H
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon May 01 00:00:00 EDT 2023
A platinum@polymer-catechol nanobraker enables radio-immunotherapy for crippling melanoma tumorigenesis, angiogenesis, and radioresistance.
Bioactive materials
Li W,Yan J,Tian H,Li B,Wang G,Sang W,Zhang Z,Zhang X,Dai Y
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
CD8α+ dendritic cells potentiate antitumor and immune activities against murine ovarian cancers.
Scientific reports
Lee SW,Lee H,Lee KW,Kim MJ,Kang SW,Lee YJ,Kim H,Kim YM
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Tue Jan 03 00:00:00 EST 2023
Acinetobacter baumannii outer membrane protein A induces autophagy in bone marrow-derived dendritic cells involving the PI3K/mTOR pathway.
Immunity, inflammation and disease
Tan H,Cao L
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
Calcaratarin D, a labdane diterpenoid, attenuates mouse asthma via modulating alveolar macrophage function.
British journal of pharmacology
Liao W,Foo HYC,Tran TNQ,Chai CLL,Wong WSF
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
Oxygen-carrying nanoplatform to reprogram tumor immunosuppressive microenvironment and enhance photothermal-immunotherapy.
Materials today. Bio
Huang J,Leng X,Jiang T,Xu L,Zheng J,Fang M,Wang J,Wang Z,Zhang L
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
Oxygen-carrying nanoplatform to reprogram tumor immunosuppressive microenvironment and enhance photothermal-immunotherapy.
Materials today. Bio
Huang J,Leng X,Jiang T,Xu L,Zheng J,Fang M,Wang J,Wang Z,Zhang L
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
Enhanced uptake and anti-maturation effect of celastrol-loaded mannosylated liposomes on dendritic cells for psoriasis treatment.
Acta pharmaceutica Sinica. B
Xi L,Lin Z,Qiu F,Chen S,Li P,Chen X,Wang Z,Zheng Y
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sat Jan 01 00:00:00 EST 2022
CD4+ CTLs Act as a Key Effector Population for Allograft Rejection of MSCs in a Donor MHC-II Dependent Manner in Injured Liver.
Aging and disease
Shen S,Li Y,Jin M,Fan D,Pan R,Lin A,Chen Y,Xiang L,Zhao RC,Shao J
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2022
In situ generation of micrometer-sized tumor cell-derived vesicles as autologous cancer vaccines for boosting systemic immune responses.
Nature communications
Guo Y,Wang SZ,Zhang X,Jia HR,Zhu YX,Zhang X,Gao G,Jiang YW,Li C,Chen X,Wu SY,Liu Y,Wu FG
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
TcpC Inhibits M1 but Promotes M2 Macrophage Polarization via Regulation of the MAPK/NF-κB and Akt/STAT6 Pathways in Urinary Tract Infection.
Cells
Fang J,Ou Q,Wu B,Li S,Wu M,Qiu J,Cen N,Hu K,Che Y,Ma Y,Pan J
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sun Aug 28 00:00:00 EDT 2022
Self-assembled traditional Chinese nanomedicine modulating tumor immunosuppressive microenvironment for colorectal cancer immunotherapy.
Theranostics
Mao Q,Min J,Zeng R,Liu H,Li H,Zhang C,Zheng A,Lin J,Liu X,Wu M
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Thu Sep 29 00:00:00 EDT 2022
Dendritic cell Piezo1 directs the differentiation of TH1 and Treg cells in cancer.
eLife
Wang Y,Yang H,Jia A,Wang Y,Yang Q,Dong Y,Hou Y,Cao Y,Dong L,Bi Y,Liu G
17-0801-82 was used in Flow cytometry/Cell sorting to demonstrate the role of the dendritic cells-based mechanical regulation of immunopathology in directing T cell lineage commitment in tumor microenvironments.
Mon Aug 22 00:00:00 EDT 2022
SYK coordinates neuroprotective microglial responses in neurodegenerative disease.
Cell
Ennerfelt H,Frost EL,Shapiro DA,Holliday C,Zengeler KE,Voithofer G,Bolte AC,Lammert CR,Kulas JA,Ulland TK,Lukens JR
17-0801-82 was used in flow cytometry to study the critical intracellular molecules that orchestrate neuroprotective functions of microglia.
Thu Oct 27 00:00:00 EDT 2022
A bilirubin-derived nanomedicine attenuates the pathological cascade of pulmonary fibrosis.
Biomaterials
Keum H,Kim D,Kim J,Kim TW,Whang CH,Jung W,Jon S
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
Prostaglandin E2 serves a dual role in regulating the migration of dendritic cells.
International journal of molecular medicine
Diao G,Huang J,Zheng X,Sun X,Tian M,Han J,Guo J
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Myeloid and plasmacytoid dendritic cell combined vaccines loaded with heat-treated tumor cell lysates enhance antitumor activity in murine lung cancer.
Oncology letters
Chen H,Tan J,Li X,Li H,Wu W,Wu Y,Zhang J,Gu L
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Cell-permeable transgelin-2 as a potent therapeutic for dendritic cell-based cancer immunotherapy.
Journal of hematology & oncology
Kim HR,Park JS,Park JH,Yasmin F,Kim CH,Oh SK,Chung IJ,Jun CD
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Wed Mar 17 00:00:00 EDT 2021
Polarization of Type 1 Macrophages Is Associated with the Severity of Viral Encephalitis Caused by Japanese Encephalitis Virus and Dengue Virus.
Cells
Jhan MK,Chen CL,Shen TJ,Tseng PC,Wang YT,Satria RD,Yu CY,Lin CF
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Nov 15 00:00:00 EST 2021
Theranostic near-infrared-IIb emitting nanoprobes for promoting immunogenic radiotherapy and abscopal effects against cancer metastasis.
Nature communications
Li H,Wang M,Huang B,Zhu SW,Zhou JJ,Chen DR,Cui R,Zhang M,Sun ZJ
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Thu Dec 09 00:00:00 EST 2021
Azithromycin alleviates systemic lupus erythematosus via the promotion of M2 polarisation in lupus mice.
Cell death discovery
Wang J,Chen Q,Zhang Z,Wang S,Wang Y,Xiang M,Liang J,Xu J
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Fri Apr 16 00:00:00 EDT 2021
Resolving macrophage polarization through distinct Ca2+ entry channel that maintains intracellular signaling and mitochondrial bioenergetics.
iScience
Nascimento Da Conceicao V,Sun Y,Ramachandran K,Chauhan A,Raveendran A,Venkatesan M,DeKumar B,Maity S,Vishnu N,Kotsakis GA,Worley PF,Gill DL,Mishra BB,Madesh M,Singh BB
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Fri Nov 19 00:00:00 EST 2021
Urolithin A ameliorates experimental autoimmune encephalomyelitis by targeting aryl hydrocarbon receptor.
EBioMedicine
Shen PX,Li X,Deng SY,Zhao L,Zhang YY,Deng X,Han B,Yu J,Li Y,Wang ZZ,Zhang Y
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
PP2Cδ Controls the Differentiation and Function of Dendritic Cells Through Regulating the NSD2/mTORC2/ACLY Pathway.
Frontiers in immunology
Lv N,Jin S,Liang Z,Wu X,Kang Y,Su L,Dong Y,Wang B,Ma T,Shi L
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Feb 28 00:00:00 EST 2022
PP2Cδ Controls the Differentiation and Function of Dendritic Cells Through Regulating the NSD2/mTORC2/ACLY Pathway.
Frontiers in immunology
Lv N,Jin S,Liang Z,Wu X,Kang Y,Su L,Dong Y,Wang B,Ma T,Shi L
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Feb 28 00:00:00 EST 2022
Requirement of direct contact between chondrocytes and macrophages for the maturation of regenerative cartilage.
Scientific reports
Kanda K,Asawa Y,Inaki R,Fujihara Y,Hoshi K,Hikita A
17-0801-82 was used in Flow Cytometry to provide insights for establishing a method of in vitro cartilage regeneration.
Thu Nov 18 00:00:00 EST 2021
The Toxoplasma gondii virulence factor ROP16 acts in cis and trans, and suppresses T cell responses.
The Journal of experimental medicine
Chen L,Christian DA,Kochanowsky JA,Phan AT,Clark JT,Wang S,Berry C,Oh J,Chen X,Roos DS,Beiting DP,Koshy AA,Hunter CA
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Mar 02 00:00:00 EST 2020
E6‑regulated overproduction of prostaglandin E2 may inhibit migration of dendritic cells in human papillomavirus 16‑positive cervical lesions.
International journal of oncology
Huang J,Diao G,Zhang Q,Chen Y,Han J,Guo J
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2020
Lung Carcinoma Cells Secrete Exosomal MALAT1 to Inhibit Dendritic Cell Phagocytosis, Inflammatory Response, Costimulatory Molecule Expression and Promote Dendritic Cell Autophagy via AKT/mTOR Pathway.
OncoTargets and therapy
Liu Y,Yin Z,Lu P,Ma Y,Luo B,Xiang L,Zhang W,He Y,Liang X
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sun Apr 17 00:00:00 EDT 2022
Interleukin-9 Inhibits Lung Metastasis of Melanoma through Stimulating Anti-Tumor M1 Macrophages.
Molecules and cells
Park SM,Do-Thi VA,Lee JO,Lee H,Kim YS
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sun May 31 00:00:00 EDT 2020
Effects of combined treatment with PD‑L1 Ig and CD40L mAb on immune tolerance in the CBA/J x DBA/2 mouse model.
Molecular medicine reports
Li G,Yang L,Li D,Zhang J,Du L,Xia L,Liu Y,Hu W
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2020
Vaccination with early ferroptotic cancer cells induces efficient antitumor immunity.
Journal for immunotherapy of cancer
Efimova I,Catanzaro E,Van der Meeren L,Turubanova VD,Hammad H,Mishchenko TA,Vedunova MV,Fimognari C,Bachert C,Coppieters F,Lefever S,Skirtach AG,Krysko O,Krysko DV
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2020
Carrot Pomace Polysaccharide (CPP) Improves Influenza Vaccine Efficacy in Immunosuppressed Mice via Dendritic Cell Activation.
Nutrients
Sun P,Kim Y,Lee H,Kim J,Han BK,Go E,Kwon S,Kang JG,You S,Kwon J
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Wed Sep 09 00:00:00 EDT 2020
Photodynamic Therapy-Based Dendritic Cell Vaccination Suited to Treat Peritoneal Mesothelioma.
Cancers
Trempolec N,Doix B,Degavre C,Brusa D,Bouzin C,Riant O,Feron O
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Thu Feb 27 00:00:00 EST 2020
Lactobacillus reuteri 5454 and Bifidobacterium animalis ssp. lactis 5764 improve colitis while differentially impacting dendritic cells maturation and antimicrobial responses.
Scientific reports
Hrdý J,Alard J,Couturier-Maillard A,Boulard O,Boutillier D,Delacre M,Lapadatescu C,Cesaro A,Blanc P,Pot B,Ryffel B,Chamaillard M,Grangette C
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Tue Mar 24 00:00:00 EDT 2020
B7-CD28 co-stimulation modulates central tolerance via thymic clonal deletion and Treg generation through distinct mechanisms.
Nature communications
Watanabe M,Lu Y,Breen M,Hodes RJ
17-0801 was used in Flow cytometry/Cell sorting to reveal a function of B7-CD28 co-stimulation in shaping the T cell repertoire and limiting autoimmunity through both thymic clonal deletion and Treg cell generation.
Tue Dec 08 00:00:00 EST 2020
The ROP16III-dependent early immune response determines the subacute CNS immune response and type III Toxoplasma gondii survival.
PLoS pathogens
Tuladhar S,Kochanowsky JA,Bhaskara A,Ghotmi Y,Chandrasekaran S,Koshy AA
17-0801-82 was used in Flow Cytometry to show ROP16III in type III strain Toxoplasma gondii can influence the subacute CNS immune response and mediate parasitic survival and persistence.
Tue Oct 01 00:00:00 EDT 2019
Classical dendritic cells regulate acute lung inflammation and injury in mice with lipopolysaccharide‑induced acute respiratory distress syndrome.
International journal of molecular medicine
Li L,Dong L,Zhao D,Gao F,Yan J
17-0801-82 was used in Flow Cytometry to explore the regulatory effects of pulmonary cDCs on acute lung inflammation and injury in lipopolysaccharide (LPS)‚induced ARDS.
Thu Aug 01 00:00:00 EDT 2019
cGAS/STING/TBK1/IRF3 Signaling Pathway Activates BMDCs Maturation Following Mycobacterium bovis Infection.
International journal of molecular sciences
Li Q,Liu C,Yue R,El-Ashram S,Wang J,He X,Zhao D,Zhou X,Xu L
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Tue Feb 19 00:00:00 EST 2019
A β2-Integrin/MRTF-A/SRF Pathway Regulates Dendritic Cell Gene Expression, Adhesion, and Traction Force Generation.
Frontiers in immunology
Guenther C,Faisal I,Uotila LM,Asens ML,Harjunpää H,Savinko T,Öhman T,Yao S,Moser M,Morris SW,Tojkander S,Fagerholm SC
17-0801 was used in Flow cytometry/Cell sorting to demonstrate that mutation of the kindlin-3 binding site in the β2-integrin abolishes activation of the actin-regulated myocardin related transcription factor A/serum response factor signalling pathway in dendritic cells and MRTF-A/SRF-dependent gene expression.
Mon Aug 17 00:00:00 EDT 2020
Mitophagy in Intestinal Epithelial Cells Triggers Adaptive Immunity during Tumorigenesis.
Cell
Ziegler PK,Bollrath J,Pallangyo CK,Matsutani T,Canli Ö,De Oliveira T,Diamanti MA,Müller N,Gamrekelashvili J,Putoczki T,Horst D,Mankan AK,Öner MG,Müller S,Müller-Höcker J,Kirchner T,Slotta-Huspenina J,Taketo MM,Reinheckel T,Dröse S,Larner AC,Wels WS,Ernst M,Greten TF,Arkan MC,Korn T,Wirth D,Greten FR
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Thu Jun 28 00:00:00 EDT 2018
CNS lymphatic drainage and neuroinflammation are regulated by meningeal lymphatic vasculature.
Nature neuroscience
Louveau A,Herz J,Alme MN,Salvador AF,Dong MQ,Viar KE,Herod SG,Knopp J,Setliff JC,Lupi AL,Da Mesquita S,Frost EL,Gaultier A,Harris TH,Cao R,Hu S,Lukens JR,Smirnov I,Overall CC,Oliver G,Kipnis J
17-0801 was used in Flow cytometry/Cell sorting to demonstrate that meningeal lymphatics fulfill two critical criteria: they assist in the drainage of cerebrospinal fluid components and enable immune cells to enter draining lymph nodes in a CCR7-dependent manner.
Mon Oct 01 00:00:00 EDT 2018
Murine CMV induces type 1 IFN that impairs differentiation of MDSCs critical for transplantation tolerance.
Blood advances
Dangi A,Zhang L,Zhang X,Luo X
17-0801 was used in Flow cytometry/Cell sorting to provide a solid foundation for seeking effective therapies for promoting transplantation tolerance in settings of CMV infection.
Tue Mar 27 00:00:00 EDT 2018
Effect of immune tolerance induced by immature dendritic cells and CTLA4-Ig on systemic lupus erythematosus: An in vivo study.
Experimental and therapeutic medicine
Huang C,Zhang L,Ling F,Wen S,Luo Y,Liu H,Liu J,Zheng W,Liang M,Sun J,Lin YK
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Thu Mar 01 00:00:00 EST 2018
T cell microvilli constitute immunological synaptosomes that carry messages to antigen-presenting cells.
Nature communications
Kim HR,Mun Y,Lee KS,Park YJ,Park JS,Park JH,Jeon BN,Kim CH,Jun Y,Hyun YM,Kim M,Lee SM,Park CS,Im SH,Jun CD
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Fri Sep 07 00:00:00 EDT 2018
miR-21 Protects Against Ischemia/Reperfusion-Induced Acute Kidney Injury by Preventing Epithelial Cell Apoptosis and Inhibiting Dendritic Cell Maturation.
Frontiers in physiology
Song N,Zhang T,Xu X,Lu Z,Yu X,Fang Y,Hu J,Jia P,Teng J,Ding X
Fri Apr 08 00:00:00 EDT 2022
Lin-CCR2+ hematopoietic stem and progenitor cells overcome resistance to PD-1 blockade.
Nature communications
Flores CT,Wildes TJ,Drake JA,Moore GL,Dean BD,Abraham RS,Mitchell DA
Wed Oct 17 00:00:00 EDT 2018
Culling of APCs by inflammatory cell death pathways restricts TIM3 and PD-1 expression and promotes the survival of primed CD8 T cells.
Cell death and differentiation
Patel R,Kim K,Shutinoski B,Wachholz K,Krishnan L,Sad S
17-0801 was used in Flow cytometry/Cell sorting to evaluate the impact of premature cell death of antigen-presenting cells by Caspase-1- and RipK3-signalling pathways on CD8+ T-cell priming during infection of mice with Salmonella typhimurium.
Wed Nov 01 00:00:00 EDT 2017
FcεRI γ-Chain Negatively Modulates Dectin-1 Responses in Dendritic Cells.
Frontiers in immunology
Pan YG,Yu YL,Lin CC,Lanier LL,Chu CL
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
Discrimination of the heterogeneity of bone marrow‑derived dendritic cells.
Molecular medicine reports
Wang J,Dai X,Hsu C,Ming C,He Y,Zhang J,Wei L,Zhou P,Wang CY,Yang J,Gong N
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Wed Nov 01 00:00:00 EDT 2017
Calreticulin is an effective immunologic adjuvant to tumor-associated antigens.
Experimental and therapeutic medicine
Wang J,Gao ZP,Qin S,Liu CB,Zou LL
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sun Oct 01 00:00:00 EDT 2017
Batf3-dependent CD8α+ Dendritic Cells Aggravates Atherosclerosis via Th1 Cell Induction and Enhanced CCL5 Expression in Plaque Macrophages.
EBioMedicine
Li Y,Liu X,Duan W,Tian H,Zhu G,He H,Yao S,Yi S,Song W,Tang H
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Paeoniflorin Ameliorates Experimental Autoimmune Encephalomyelitis via Inhibition of Dendritic Cell Function and Th17 Cell Differentiation.
Scientific reports
Zhang H,Qi Y,Yuan Y,Cai L,Xu H,Zhang L,Su B,Nie H
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Feb 06 00:00:00 EST 2017
Pplase of Dermatophagoides farinae promotes ovalbumin-induced airway allergy by modulating the functions of dendritic cells in a mouse model.
Scientific reports
Wang H,Mo L,Xiao X,An S,Liu X,Ba J,Wu W,Ran P,Yang P,Liu Z
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Feb 27 00:00:00 EST 2017
Tumor-derived exosomes educate dendritic cells to promote tumor metastasis via HSP72/HSP105-TLR2/TLR4 pathway.
Oncoimmunology
Shen Y,Guo D,Weng L,Wang S,Ma Z,Yang Y,Wang P,Wang J,Cai Z
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Der f 31, a novel allergen from Dermatophagoides farinae, activates epithelial cells and enhances lung-resident group 2 innate lymphoid cells.
Scientific reports
Wang H,Lin J,Zeng L,Ouyang C,Ran P,Yang P,Liu Z
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Thu Aug 17 00:00:00 EDT 2017
Reprogramming MHC specificity by CRISPR-Cas9-assisted cassette exchange.
Scientific reports
Kelton W,Waindok AC,Pesch T,Pogson M,Ford K,Parola C,Reddy ST
17-0801 was used in Flow Cytometry to demonstrate a proof-of-concept for reprogramming MHC-specificity by performing CRISPR-Cas9-assisted cassette exchange.
Tue Apr 04 00:00:00 EDT 2017
Identification of 14-dehydroergosterol as a novel anti-inflammatory compound inducing tolerogenic dendritic cells.
Scientific reports
Ano Y,Ikado K,Shindo K,Koizumi H,Fujiwara D
17-0801 was used in Flow cytometry/Cell sorting to investigate if 14-dehydroergosterol induces tolerogenic dendritic cells (DCs) to convert to a tolerogenic type.
Tue Oct 24 00:00:00 EDT 2017
A temperature sensitive Mycobacterium paragordonae induces enhanced protective immune responses against mycobacterial infections in the mouse model.
Scientific reports
Kim BJ,Kim BR,Kook YH,Kim BJ
17-0801 was used in Flow cytometry/Cell sorting to evaluate the use of temperature sensitive Mycobacterium spp. for live vaccination against Mycobacterium tuberculosis and Mycobacterium abscessus.
Thu Nov 09 00:00:00 EST 2017
Opposite Effects of Coinjection and Distant Injection of Mesenchymal Stem Cells on Breast Tumor Cell Growth.
Stem cells translational medicine
Zheng H,Zou W,Shen J,Xu L,Wang S,Fu YX,Fan W
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2016
Repulsive guidance molecule a blockade exerts the immunoregulatory function in DCs stimulated with ABP and LPS.
Human vaccines & immunotherapeutics
Xu X,Gao Y,Zhai Z,Zhang S,Shan F,Feng J
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Tue Aug 02 00:00:00 EDT 2016
CD80 and CD86 knockdown in dendritic cells regulates Th1/Th2 cytokine production in asthmatic mice.
Experimental and therapeutic medicine
Li JG,DU YM,Yan ZD,Yan J,Zhuansun YX,Chen R,Zhang W,Feng SL,Ran PX
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
Ablating the aryl hydrocarbon receptor (AhR) in CD11c+ cells perturbs intestinal epithelium development and intestinal immunity.
Scientific reports
Chng SH,Kundu P,Dominguez-Brauer C,Teo WL,Kawajiri K,Fujii-Kuriyama Y,Mak TW,Pettersson S
17-0801 was used in Flow cytometry/Cell sorting to explore the contribution of the Aryl hydrocarbon Receptor signalling pathway to the regulation of innate immunity.
Tue Apr 12 00:00:00 EDT 2016
Monomeric annexin A2 is an oxygen-regulated toll-like receptor 2 ligand and adjuvant.
Journal for immunotherapy of cancer
Andersen BM,Xia J,Epstein AL,Ohlfest JR,Chen W,Blazar BR,Pennell CA,Olin MR
17-0801 was used in Flow cytometry/Cell sorting to study the effect of ANXA2m on CD80 and CD86 expression, antigen cross-presentation, and the secretion of various cytokines.
Wed Feb 17 00:00:00 EST 2016
B-cell very late antigen-4 deficiency reduces leukocyte recruitment and susceptibility to central nervous system autoimmunity.
Annals of neurology
Lehmann-Horn K,Sagan SA,Bernard CC,Sobel RA,Zamvil SS
17-0801 was used in Flow cytometry/Cell sorting to report that the selective inhibition of VLA-4 expression on B cells impedes CNS accumulation of B cells, and recruitment of Th17 cells and macrophages.
Fri May 01 00:00:00 EDT 2015
Systemically delivered adipose stromal vascular fraction cells disseminate to peripheral artery walls and reduce vasomotor tone through a CD11b+ cell-dependent mechanism.
Stem cells translational medicine
Morris ME,Beare JE,Reed RM,Dale JR,LeBlanc AJ,Kaufman CL,Zheng H,Ng CK,Williams SK,Hoying JB
17-0801 was used in Flow cytometry/Cell sorting to investigate the potential of SVF cells to therapeutically modulate small artery vasoactivity.
Wed Apr 01 00:00:00 EDT 2015
Adjuvants may reduce in vivo transfection levels for DNA vaccination in mice leading to reduced antigen-specific CD8+ T cell responses.
Human vaccines & immunotherapeutics
Rose AH,Hoffmann FW,Hara JH,Urschitz J,Moisyadi S,Hoffmann PR,Bertino P
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon May 16 00:00:00 EDT 2016
ROCK-isoform-specific polarization of macrophages associated with age-related macular degeneration.
Cell reports
Zandi S,Nakao S,Chun KH,Fiorina P,Sun D,Arita R,Zhao M,Kim E,Schueller O,Campbell S,Taher M,Melhorn MI,Schering A,Gatti F,Tezza S,Xie F,Vergani A,Yoshida S,Ishikawa K,Yamaguchi M,Sasaki F,Schmidt-Ullrich R,Hata Y,Enaida H,Yuzawa M,Yokomizo T,Kim YB,Sweetnam P,Ishibashi T,Hafezi-Moghadam A
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Tue Feb 24 00:00:00 EST 2015
Lignin-rich enzyme lignin (LREL), a cellulase-treated lignin-carbohydrate derived from plants, activates myeloid dendritic cells via Toll-like receptor 4 (TLR4).
The Journal of biological chemistry
Tsuji R,Koizumi H,Aoki D,Watanabe Y,Sugihara Y,Matsushita Y,Fukushima K,Fujiwara D
17-0801 was used in Flow cytometry/Cell sorting to investigate the role of lignin-rich enzyme lignin in dendritic cells.
Fri Feb 13 00:00:00 EST 2015
Apoptotic cell clearance of Leishmania major-infected neutrophils by dendritic cells inhibits CD8⁺ T-cell priming in vitro by Mer tyrosine kinase-dependent signaling.
Cell death & disease
Ribeiro-Gomes FL,Romano A,Lee S,Roffê E,Peters NC,Debrabant A,Sacks D
17-0801 was used in Flow cytometry/Cell sorting to define the clearance of infected neutrophils by dendritic cells as central to the early immune evasion strategies of L. major.
Thu Dec 10 00:00:00 EST 2015
CCDC88B is a novel regulator of maturation and effector functions of T cells during pathological inflammation.
The Journal of experimental medicine
Kennedy JM,Fodil N,Torre S,Bongfen SE,Olivier JF,Leung V,Langlais D,Meunier C,Berghout J,Langat P,Schwartzentruber J,Majewski J,Lathrop M,Vidal SM,Gros P
17-0801 was used in Flow cytometry/Cell sorting to suggest that CCDC88B is the morbid gene underlying the pleiotropic effect of the 11q13 locus on inflammation.
Mon Dec 15 00:00:00 EST 2014
Eosinophils regulate peripheral B cell numbers in both mice and humans.
Journal of immunology (Baltimore, Md. : 1950)
Wong TW,Doyle AD,Lee JJ,Jelinek DF
17-0801 was used in Flow cytometry/Cell sorting to assess the effect of eosinophils on B cell survival, proliferation, and Ig secretion.
Tue Apr 15 00:00:00 EDT 2014
Oxidized lipids block antigen cross-presentation by dendritic cells in cancer.
Journal of immunology (Baltimore, Md. : 1950)
Ramakrishnan R,Tyurin VA,Veglia F,Condamine T,Amoscato A,Mohammadyani D,Johnson JJ,Zhang LM,Klein-Seetharaman J,Celis E,Kagan VE,Gabrilovich DI
17-0801 was used in Flow cytometry/Cell sorting to reveal a novel role for oxidised lipids in the regulation of cross-presentation.
Sat Mar 15 00:00:00 EDT 2014
Nizatidine, a small molecular compound, enhances killed H5N1 vaccine cell-mediated responses and protects mice from lethal viral challenge.
Human vaccines & immunotherapeutics
Wang S,Wu B,Xue J,Wang M,Chen R,Wang B
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Jan 05 00:00:00 EST 2015
BCG vaccine mediated reduction in the MHC-II expression of macrophages and dendritic cells is reversed by activation of Toll-like receptors 7 and 9.
Cellular immunology
Bakhru P,Sirisaengtaksin N,Soudani E,Mukherjee S,Khan A,Jagannath C
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2014
Lung dendritic cells undergo maturation and polarization towards a T helper type 2-stimulating phenotype in a mouse model of asthma: Role of nerve growth factor.
Experimental and therapeutic medicine
Qin Q,Wang Z,Pan P,Cao Z,Xia Q,Tan H,Hu C
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Sat Nov 01 00:00:00 EDT 2014
The mucosal adjuvant cyclic di-AMP exerts immune stimulatory effects on dendritic cells and macrophages.
PloS one
Škrnjug I,Rueckert C,Libanova R,Lienenklaus S,Weiss S,Guzmán CA
17-0801 was used in Flow cytometry/Cell sorting to assess changes in cell surface marker expression of dendritic cells and macrophages in response to c-di-AMP.
Wed Jan 14 00:00:00 EST 2015
Intercellular communication through contacts between continuous pseudopodial extensions in a macrophage-like cell line.
Cell communication & adhesion
Arrevillaga-Boni G,Hernández-Ruiz M,Castillo EC,Ortiz-Navarrete V
17-0801 was used in Flow cytometry/Cell sorting to investigate direct intercellular communication in a macrophage-like cell line, showing communication through contacts between continuous pseudopodial extensions.
Fri Aug 01 00:00:00 EDT 2014
Multifaceted therapeutic targeting of ovarian peritoneal carcinomatosis through virus-induced immunomodulation.
Molecular therapy : the journal of the American Society of Gene Therapy
Gujar S,Dielschneider R,Clements D,Helson E,Shmulevitz M,Marcato P,Pan D,Pan LZ,Ahn DG,Alawadhi A,Lee PW
17-0801 was used in Flow cytometry/Cell sorting to develop a virus-based therapy that suppresses immunomodulation and targets ovarian peritoneal carcinomatosis.
Fri Feb 01 00:00:00 EST 2013
Neutralization of TSLP inhibits airway remodeling in a murine model of allergic asthma induced by chronic exposure to house dust mite.
PloS one
Chen ZG,Zhang TT,Li HT,Chen FH,Zou XL,Ji JZ,Chen H
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Tue Aug 20 00:00:00 EDT 2013
Resistance to malaria by enhanced phagocytosis of erythrocytes in LMP7-deficient mice.
PloS one
Duan X,Imai T,Chou B,Tu L,Himeno K,Suzue K,Hirai M,Taniguchi T,Okada H,Shimokawa C,Hisaeda H
17-0801 was used in Flow cytometry/Cell sorting to study how RBCs of LMP7-deficient mice were more likely to deform in response to infection with malaria parasites.
Tue Sep 17 00:00:00 EDT 2013
Orientia tsutsugamushi subverts dendritic cell functions by escaping from autophagy and impairing their migration.
PLoS neglected tropical diseases
Choi JH,Cheong TC,Ha NY,Ko Y,Cho CH,Jeon JH,So I,Kim IK,Choi MS,Kim IS,Cho NH
17-0801 was used in Flow cytometry/Cell sorting to investigate how Orientia tsutsugamushi can exploit dendritic cells as replication reservoirs and impair functional maturation of these cells during infection in mammals.
Wed Jun 19 00:00:00 EDT 2013
Nonviral delivery of small interfering RNA into pancreas-associated immune cells prevents autoimmune diabetes.
Molecular therapy : the journal of the American Society of Gene Therapy
Leconet W,Petit P,Peraldi-Roux S,Bresson D
17-0801 was used in Flow cytometry/Cell sorting to provide new insights into the development of siRNA-based therapeutics for the treatment of T1D.
Sat Dec 01 00:00:00 EST 2012
Enhancement of CTLs induced by DCs loaded with ubiquitinated hepatitis B virus core antigen.
World journal of gastroenterology
Chen JH,Yu YS,Chen XH,Liu HH,Zang GQ,Tang ZH
17-0801 was used in Flow cytometry/Cell sorting to demonstrate a method for the HBV-induced augmentation of the CTL response.
Wed Mar 28 00:00:00 EDT 2012
Losartan inhibits conventional dendritic cell maturation and Th1 and Th17 polarization responses: Νovel mechanisms of preventive effects on lipopolysaccharide-induced acute lung injury.
International journal of molecular medicine
Liu J,Zhang PS,Yu Q,Liu L,Yang Y,Guo FM,Qiu HB
17-0801 was used in Flow cytometry/Cell sorting to demonstrate that losartan suppresses conventional dendritic cell maturation and Th1 and Th17 polarization responses.
Wed Feb 01 00:00:00 EST 2012
Immunogenicity and protective capacity of a virosomal respiratory syncytial virus vaccine adjuvanted with monophosphoryl lipid A in mice.
PloS one
Kamphuis T,Meijerhof T,Stegmann T,Lederhofer J,Wilschut J,de Haan A
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Wed Sep 19 00:00:00 EDT 2012
Flt3L combined with rapamycin promotes cardiac allograft tolerance by inducing regulatory dendritic cells and allograft autophagy in mice.
PloS one
Xiong A,Duan L,Chen J,Fan Z,Zheng F,Tan Z,Gong F,Fang M
17-0801 was used in Flow cytometry/Cell sorting to investigate the role of Flt3 and Flt3L in immune tolerance, showing that Flt3L combined with rapamycin promotes cardiac allograft tolerance via dendritic cell induction.
Tue Jun 11 00:00:00 EDT 2013
A critical role for SOCS3 in innate resistance to Toxoplasma gondii.
Cell host & microbe
Whitmarsh RJ,Gray CM,Gregg B,Christian DA,May MJ,Murray PJ,Hunter CA
17-0801 was used in Flow cytometry/Cell sorting to study the role of SOCS3 in suppressing IL-6 signalling during immune responses to T. gondii infection.
Thu Sep 15 00:00:00 EDT 2011
Orally administered eicosapentaenoic acid induces rapid regression of atherosclerosis via modulating the phenotype of dendritic cells in LDL receptor-deficient mice.
Arteriosclerosis, thrombosis, and vascular biology
Nakajima K,Yamashita T,Kita T,Takeda M,Sasaki N,Kasahara K,Shinohara M,Rikitake Y,Ishida T,Yokoyama M,Hirata K
17-0801 was used in Flow cytometry/Cell sorting to investigate the effect of the administration of EPA on the regression of atherosclerosis.
Thu Sep 01 00:00:00 EDT 2011
Excess circulating alternatively activated myeloid (M2) cells accelerate ALS progression while inhibiting experimental autoimmune encephalomyelitis.
PloS one
Vaknin I,Kunis G,Miller O,Butovsky O,Bukshpan S,Beers DR,Henkel JS,Yoles E,Appel SH,Schwartz M
17-0801 was used in Flow cytometry/Cell sorting to establish circulating MDSCs as a novel target for therapeutic intervention in ALS, at least at the early stage.
Mon Mar 26 00:00:00 EDT 2012
Dendritic cells transduced with lentiviral vectors expressing VIP differentiate into VIP-secreting tolerogenic-like DCs.
Molecular therapy : the journal of the American Society of Gene Therapy
Toscano MG,Delgado M,Kong W,Martin F,Skarica M,Ganea D
17-0801 was used in Flow cytometry/Cell sorting to characterise the phenotype and therapeutic effects of transduced LentiVIP-dendritic cells in models of EAE and CLP, showing that they differentiate into VIP-secreting tolerogenic-like DCs.
Sat May 01 00:00:00 EDT 2010
Oral administration of an active form of vitamin D3 (calcitriol) decreases atherosclerosis in mice by inducing regulatory T cells and immature dendritic cells with tolerogenic functions.
Arteriosclerosis, thrombosis, and vascular biology
Takeda M,Yamashita T,Sasaki N,Nakajima K,Kita T,Shinohara M,Ishida T,Hirata K
17-0801 was used in Flow cytometry/Cell sorting to demonstrate that immune modulation by calcitriol may be a potentially valuable therapeutic approach against atherosclerosis.
Wed Dec 01 00:00:00 EST 2010
Dendritic cells continue to capture and present antigens after maturation in vivo.
Journal of immunology (Baltimore, Md. : 1950)
Drutman SB,Trombetta ES
17-0801 was used in Flow cytometry/Cell sorting to study the dynamics of Ag capture and presentation in situ, showing that dendritic cells continue to capture and present antigens after maturation.
Sun Aug 15 00:00:00 EDT 2010
Dynamic equilibrium of B7-1 dimers and monomers differentially affects immunological synapse formation and T cell activation in response to TCR/CD28 stimulation.
Journal of immunology (Baltimore, Md. : 1950)
Bhatia S,Sun K,Almo SC,Nathenson SG,Hodes RJ
17-0801 was used in Flow cytometry/Cell sorting to suggest a novel model in which optimal T-cell costimulatory function of B7-1 requires high-avidity CD28 engagement by dimeric B7-1.
Mon Feb 15 00:00:00 EST 2010
Antigen-specific immunity and cross-priming by epithelial ovarian carcinoma-induced CD11b(+)Gr-1(+) cells.
Journal of immunology (Baltimore, Md. : 1950)
Tomihara K,Guo M,Shin T,Sun X,Ludwig SM,Brumlik MJ,Zhang B,Curiel TJ,Shin T
17-0801 was used in Flow cytometry/Cell sorting to investigate CD11b(+)Gr-1(+) cells found in epithelial ovarian cancer-bearing mice, which are induced by ovarian cancer cells and undergo antigen-specific immunity and cross-priming.
Tue Jun 01 00:00:00 EDT 2010
Magnetic nanoparticles for imaging dendritic cells.
Magnetic resonance in medicine
Kobukai S,Baheza R,Cobb JG,Virostko J,Xie J,Gillman A,Koktysh D,Kerns D,Does M,Gore JC,Pham W
17-0801 was used in Flow cytometry/Cell sorting to develop magnetic nanoparticles for imaging dendritic cells.
Sat May 01 00:00:00 EDT 2010
CCR7 deficiency leads to leukocyte activation and increased clearance in response to pulmonary Pseudomonas aeruginosa infection.
Infection and immunity
Eppert BL,Motz GT,Wortham BW,Flury JL,Borchers MT
17-0801 was used in Flow cytometry/Cell sorting to investigate the roles of CCR7 in the host defense against pulmonary infection, showing that CCR7 deficiency leads to leukocyte activation and increased infection clearance.
Sat May 01 00:00:00 EDT 2010
Host-derived interleukin-1alpha is important in determining the immunogenicity of 3-methylcholantrene tumor cells.
Journal of immunology (Baltimore, Md. : 1950)
Elkabets M,Krelin Y,Dotan S,Cerwenka A,Porgador A,Lichtenstein RG,White MR,Zoller M,Iwakura Y,Dinarello CA,Voronov E,Apte RN
17-0801 was used in Flow cytometry/Cell sorting to elucidate the involvement of IL-1 molecules in 3-MCA-induced carcinogenesis.
Wed Apr 15 00:00:00 EDT 2009
Dynamics of lung macrophage activation in response to helminth infection.
Journal of leukocyte biology
Siracusa MC,Reece JJ,Urban JF,Scott AL
Published figure using CD80 (B7-1) monoclonal antibody (Product # 17-0801-82) in Flow Cytometry
Mon Dec 01 00:00:00 EST 2008
T cell-dendritic cell immunological synapses contain TCR-dependent CD28-CD80 clusters that recruit protein kinase C theta.
Journal of immunology (Baltimore, Md. : 1950)
Tseng SY,Waite JC,Liu M,Vardhana S,Dustin ML
17-0801 was used in Flow cytometry/Cell sorting to demonstrate that the T cell-DC interface contains dynamic costimulatory foci that share characteristics of microclusters and cSMACs.
Wed Oct 01 00:00:00 EDT 2008
Recruitment and expansion of dendritic cells in vivo potentiate the immunogenicity of plasmid DNA vaccines.
The Journal of clinical investigation
Sumida SM,McKay PF,Truitt DM,Kishko MG,Arthur JC,Seaman MS,Jackson SS,Gorgone DA,Lifton MA,Letvin NL,Barouch DH
17-0801 was used in Flow cytometry/Cell sorting to suggest that the availability of mature DCs at the site of inoculation is a critical rate-limiting factor for DNA vaccine immunogenicity.
Mon Nov 01 00:00:00 EST 2004