Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene encodes a protein that has been reported to function in signaling pathways governing transcriptional regulation and cell cycle progression. It may play a role in tumorigenesis and metastasis. A pseudogene of this gene is located on the long arm of chromosome 20. Alternative splicing results in multiple transcript variants that encode different isoforms. [provided by RefSeq, May 2013].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDK5 activator-binding protein C53; CDK5 regulatory subunit associated protein IC53-2; IC53-2; ischemic heart CDK5 activator-binding protein C53; LXXLL/leucine-zipper-containing ARF-binding protein; LXXLL/leucine-zipper-containing ARFbinding protein; OK/SW-cl.114; unnamed protein product
Gene Aliases: 1810007E24Rik; C53; HSF-27; IC53; LZAP; MST016; OK/SW-cl.114; PP1553
UniProt ID: (Human) Q96JB5, (Mouse) Q99LM2
Entrez Gene ID: (Human) 80279, (Rat) 80278, (Mouse) 80280
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support