Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CDKN2B Antibodies

          Invitrogen

          CDKN2B Polyclonal Antibody

          View all (26) CDKN2B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CDKN2B Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          CDKN2B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CDKN2B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CDKN2B Antibody (PA5-116445) in ICC/IF

          Immunocytochemical analysis of CDKN2B in hela cells using a CDKN2B polyclonal antibody (Product #PA5-116445). Formalin fixed cells, as seen in green, were permeabilized with 0.1% Triton X-100 in TBS for 10 minutes at room temperature and blocked with 1% Blocker BSA for 15 minutes at room temperature. The cells were probed with the primary antibody at a dilution of 1:200 for 1 hour at room temperature, washed with PBS. Alexa Fluor™ 488 Goat anti-Rabbit IgG was used as the secondary antibody at 1:100 dilution. The nuclear counter stain is DA... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CDKN2B Antibody in Immunocytochemistry (ICC/IF)
          CDKN2B Antibody in Immunocytochemistry (ICC/IF)
          CDKN2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CDKN2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CDKN2B Antibody in Western Blot (WB)
          CDKN2B Antibody in Flow Cytometry (Flow)
          CDKN2B Polyclonal Antibody

          Product Details

          PA5-116445

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein within Human p15 INK4b aa 1-138.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 0.2% BSA, 50% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2901077

          Product Specific Information

          Positive controls: Hela, SiHa, rat kidney tissue, human esophagus cancer tissue, human small intestine tissue, mouse liver tissue.

          Target Information

          The four known 15-to 19 kDa INK4 proteins (p15INK4b, p16INK4a, p18INK4c andp19INK4d) bind and inhibit CDK4 and CDK6 but not other CDKs, including cyclin E-Cdk2, cyclin A-Cdk2 and cyclin B-Cdk1. Like the three D-type cyclins, the INK4 genes are expressed in distinct tissue-specific patterns, suggesting that they are not strictly redundant. 5 p18INK4c is expressed during G1 to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S-phase.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CDK inhibitory protein; CDK4B inhibitor; CDKN2B; Cyclin dependent kinase inhibitor 2B (p15, inhibits CDK4); Cyclin-dependent kinase 4 inhibitor B; cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4); cyclin-dependent kinase inhibitor p15; cyclin-dependent kinase inhibitor p15INK4b; cyclin-dependent kinase inhibitor protein; cyclin-dependent kinases 4 and 6 binding protein; MTS-2; Multiple tumor suppressor 2; p10; p14-INK4b; p14_CDK inhibitor; p14_INK4B; p15; p15 CDK inhibitor; p15-INK4b; p15_INK4B; p15INK4B

          View more View less

          Gene Aliases: AV083695; CDK4I; CDKN2B; Ink4; INK4B; MTS2; P15; p15(INK4b); p15INK4b; TP15

          View more View less

          UniProt ID: (Human) P42772, (Mouse) P55271, (Rat) P55272

          View more View less

          Entrez Gene ID: (Human) 1030, (Mouse) 12579, (Rat) 25164

          View more View less

          Function(s)
          cyclin-dependent protein serine/threonine kinase inhibitor activity protein binding protein kinase binding molecular_function
          Process(es)
          transforming growth factor beta receptor signaling pathway negative regulation of cell proliferation megakaryocyte differentiation positive regulation of transforming growth factor beta receptor signaling pathway cellular response to nutrient spleen development negative regulation of epithelial cell proliferation regulation of G0 to G1 transition cellular response to cell-matrix adhesion cellular senescence regulation of G1/S transition of mitotic cell cycle negative regulation of G1/S transition of mitotic cell cycle regulation of cyclin-dependent protein serine/threonine kinase activity G2/M transition of mitotic cell cycle cell cycle cell cycle arrest mitotic cell cycle checkpoint cellular response to extracellular stimulus negative regulation of phosphorylation negative regulation of protein serine/threonine kinase activity liver development aging response to organic cyclic compound positive regulation of epithelial cell differentiation response to cytokine
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline