    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • COX4NB Antibodies
          Proteintech
          COX4NB Monoclonal Antibody (2B8D3), CoraLite® Plus 488
          View all (23) COX4NB antibodies
          Questions & Answers
          Cite COX4NB Monoclonal Antibody (2B8D3), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          COX4NB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          COX4NB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          COX4NB Antibody (CL488-66547) in ICC/IF
          Immunofluorescent analysis of HeLa cells using CoraLite® Plus 488-conjugated COX4NB Monoclonal antibody (CL488-66547, Clone: 2B8D3). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          COX4NB Antibody in Immunocytochemistry (ICC/IF)
          COX4NB Antibody in Flow Cytometry (Flow)
          COX4NB Monoclonal Antibody (2B8D3), CoraLite® Plus 488
          Product Details
          CL488-66547
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.4 µg/1x10^6 cell suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2B8D3
          Immunogen
          Recombinant protein
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Purification
          Protein A
          Storage buffer
          PBS with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MPGVKLTTQA YCKMVLHGAK YPHCAVNGLL VAEKQKPRKE HLPLGGPGAH HTLFVDCIPL FHGTLALAPM LEVALTLIDS WCKDHSYVIA GYYQANERVK DASPNQVAEK VASRIAEGFS DTALIMVDNT KFTMDCVAPT IHVYEHHENR WRCRDPHHDY CEDWPEAQRI SASLLDSRSY ETLVDFDNHL DDIRNDWTNP EINKAVLHLC
          Target Information
          EMC8 (ER Membrane Protein Complex Subunit 8) is a Protein Coding gene. An important paralog of this gene is EMC9.[GeneCards]
          Bioinformatics
          Protein Aliases: COX4 neighbor; ER membrane protein complex subunit 8; family with sequence similarity 158, member B; Neighbor of COX4; Protein FAM158B; unnamed protein product
          View more View less
          Gene Aliases: C16orf2; C16orf4; COX4AL; COX4NB; EMC8; FAM158B; NOC4
          View more View less
          UniProt ID: (Human) O43402, (Mouse) O70378, (Rat) Q5FVL2
          View more View less
          Entrez Gene ID: (Human) 10328, (Mouse) 18117, (Rat) 361425
          View more View less
          Function(s)
          protein binding membrane insertase activity molecular_function
          Process(es)
          protein insertion into ER membrane by stop-transfer membrane-anchor sequence tail-anchored membrane protein insertion into ER membrane biological_process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0