    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SLC25A1 Antibodies
          Proteintech
          SLC25A1 Monoclonal Antibody (1F11A3), CoraLite® Plus 488
          View all (9) SLC25A1 antibodies
          Questions & Answers
          Cite SLC25A1 Monoclonal Antibody (1F11A3), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          SLC25A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SLC25A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SLC25A1 Antibody (CL488-66771) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL488-66802 (Cytokeratin 16 antibody) at dilution of 1:8,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SLC25A1 Antibody in Western Blot (WB)
          SLC25A1 Monoclonal Antibody (1F11A3), CoraLite® Plus 488
          Product Details
          CL488-66771
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:4,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG3
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1F11A3
          Immunogen
          SLC25A1 fusion protein Ag7520
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MPAPRAPRAL AAAAPASGKA KLTHPGKAIL AGGLAGGIEI CITFPTEYVK TQLQLDERSH PPRYRGIGDC VRQTVRSHGV LGLYRGLSSL LYGSIPKAAV RFGMFEFLSN HMRDAQGRLD STRGLLCGLG AGVAEAVVVV CPMETIKVKF IHDQTSPNPK YRGFFHGVRE IVREQGLKGT YQGLTATVLK QGSNQAIRFF VMTSLRNWYR GDNPNKPMNP LITGVFGAIA GAASVFGNTP LDVIKTRMQG LEAHKYRNTW DCGLQILKKE GLKAFYKGTV PRLGRVCLDV AIVFVIYDEV VKLLNKVWKT D
          Target Information
          The mitochondrial tricarboxylate transporter (also called citrate transport protein, or CTP) is responsible for the movement of citrate across the mitochondrial inner membrane.
          Bioinformatics
          Protein Aliases: CIC; Citrate carrier; citrate isocitrate carrier; Citrate transport protein; citrate transporter) member 1; CTP; DiGeorge syndrome gene j; Mitochondrial citrate carrier; mitochondrial tricarboxylate carrier; pmCiC; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; Solute carrier family 25 member 1; Tricarboxylate carrier protein; Tricarboxylate transport protein, mitochondrial; txtp; unnamed protein product
          View more View less
          Gene Aliases: 1300019P08Rik; 2610100G11Rik; AI194714; CIC; CMS23; CTP; D2L2AD; Dgsj; SEA; SLC20A3; SLC25A1
          View more View less
          UniProt ID: (Human) P53007, (Rat) P32089, (Mouse) Q8JZU2
          View more View less
          Entrez Gene ID: (Human) 6576, (Rat) 29743, (Mouse) 13358
          View more View less
          Function(s)
          citrate transmembrane transporter activity tricarboxylic acid transmembrane transporter activity antiporter activity citrate secondary active transmembrane transporter activity molecular_function transporter
          Process(es)
          ion transport mitochondrial citrate transport transmembrane transport negative regulation of ferroptosis transport
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline