    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cytokeratin 16 Antibodies
          Proteintech
          Cytokeratin 16 Monoclonal Antibody (2H4D8), CoraLite® Plus 488
          View all (46) Cytokeratin 16 antibodies
          Questions & Answers
          Cite Cytokeratin 16 Monoclonal Antibody (2H4D8), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Cytokeratin 16 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Cytokeratin 16 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Cytokeratin 16 Antibody (CL488-66802) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL488-66823 (OGT antibody) at dilution of 1:2,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Cytokeratin 16 Antibody in Western Blot (WB)
          Cytokeratin 16 Monoclonal Antibody (2H4D8), CoraLite® Plus 488
          Product Details
          CL488-66802
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:16,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2H4D8
          Immunogen
          Cytokeratin 16 fusion protein Ag11240
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MQNLNDRLAS YLDKVRALEE ANADLEVKIR DWYQRQRPSE IKDYSPYFKT IEDLRNKIIA ATIENAQPIL QIDNARLAAD DFRTKYEHEL ALRQTVEADV NGLRRVLDEL TLARTDLEMQ IEGLKEELAY LRKNHEEEML ALRGQTGGDV NVEMDAAPGV DLSRILNEMR DQYEQMAEKN RRDAETWFLS KTEELNKEVA SNSELVQSSR SEVTELRRVL QGLEIELQSQ LSMKASLENS LEETKGRYCM QLSQIQGLIG SVEEQLAQLR CEMEQQSQEY QILLDVKTRL EQEIATYRRL LEGEDAHLSS QQASGQSYSS REVFTSSSSS SSRQTRPILK EQSSSSFSQG QSS
          Target Information
          KRT16 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region of chromosome 17q12-q21. This keratin has been coexpressed with keratin 14 in a number of epithelial tissues, including esophagus, tongue, and hair follicles. Mutations in this gene are associated with type 1 pachyonychia congenita, non-epidermolytic palmoplantar keratoderma and unilateral palmoplantar verrucous nevus.
          Bioinformatics
          Protein Aliases: CK-16; cytokeratin 16; Cytokeratin-16; focal non-epidermolytic palmoplantar keratoderma; intermediate filament protein K16; K16; keratin 16 homolog; keratin 16, type I; Keratin, type I cytoskeletal 16; Keratin-16; unnamed protein product
          View more View less
          Gene Aliases: CK16; FNEPPK; K16; K1CP; KRT16; KRT16A; NEPPK; PC1
          View more View less
          UniProt ID: (Human) P08779
          View more View less
          Entrez Gene ID: (Human) 3868
          View more View less
          Function(s)
          structural molecule activity structural constituent of cytoskeleton protein binding structural constituent of epidermis
          Process(es)
          morphogenesis of an epithelium inflammatory response cytoskeleton organization keratinocyte differentiation negative regulation of cell migration keratinization hair cycle innate immune response intermediate filament cytoskeleton organization intermediate filament organization keratinocyte migration establishment of skin barrier
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline