    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BCAT1 Antibodies
          Proteintech
          BCAT1 Monoclonal Antibody (1G3A10), CoraLite® Plus 488
          View all (16) BCAT1 antibodies
          Questions & Answers
          Cite BCAT1 Monoclonal Antibody (1G3A10), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          BCAT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          BCAT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          BCAT1 Antibody (CL488-67084) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL488-68004 (ASNS antibody) at dilution of 1:2,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          BCAT1 Antibody in Western Blot (WB)
          Product Details
          CL488-67084
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:16,000
          -
          Product Specifications
          Species Reactivity
          Human, Pig
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1G3A10
          Immunogen
          BCAT1 fusion protein Ag28328
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MDCSNGCSAE CTGEGGSKEV VGTFKAKDLI VTPATILKEK PDPNNLVFGT VFTDHMLTVE WSSEFGWEKP HIKPLQNLSL HPGSSALHYA VELFEGLKAF RGVDNKIRLF QPNLNMDRMY RSAVRATLPV FDKEELLECI QQLVKLDQEW VPYSTSASLY IRPTFIGTEP SLGVKKPTKA LLFVLLSPVG PYFSSGTFNP VSLWANPKYV RAWKGGTGDC KMGGNYGSSL FAQCEAVDNG CQQVLWLYGE DHQITEVGTM NLFLYWINED GEEELATPPL DGIILPGVTR RCILDLAHQW DTELSLFSIN LPDFLQFIYF
          Target Information
          This gene encodes the cytosolic form of the enzyme branched-chain amino acid transaminase. This enzyme catalyzes the reversible transamination of branched-chain alpha-keto acids to branched-chain L-amino acids essential for cell growth. Two different clinical disorders have been attributed to a defect of branched-chain amino acid transamination: hypervalinemia and hyperleucine-isoleucinemia. As there is also a gene encoding a mitochondrial form of this enzyme, mutations in either gene may contribute to these disorders. Alternatively spliced transcript variants have been described.
          Bioinformatics
          Protein Aliases: BCAT(c); branched chain amino-acid transaminase 1, cytosolic; branched chain aminotransferase 1, cytosolic; Branched-chain-amino-acid aminotransferase, cytosolic; ECA39; placental protein 18; unnamed protein product
          View more View less
          Gene Aliases: BCAT1; BCATC; BCT1; ECA39; MECA39; PNAS121; PP18
          View more View less
          UniProt ID: (Human) P54687
          View more View less
          Entrez Gene ID: (Human) 586, (Pig) 100518784
          View more View less
          Function(s)
          catalytic activity branched-chain-amino-acid transaminase activity protein binding transaminase activity transferase activity L-leucine transaminase activity L-valine transaminase activity L-isoleucine transaminase activity transaminase transferase metabolite interconversion enzyme
          Process(es)
          G1/S transition of mitotic cell cycle lipid metabolic process cellular amino acid biosynthetic process branched-chain amino acid metabolic process branched-chain amino acid biosynthetic process branched-chain amino acid catabolic process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-7c59b9b899-4kcjf:80/100.66.78.36:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline