    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GNA13 Antibodies
          Proteintech
          GNA13 Monoclonal Antibody (2G8F10), CoraLite® Plus 488
          View all (15) GNA13 antibodies
          Questions & Answers
          Cite GNA13 Monoclonal Antibody (2G8F10), CoraLite® Plus 488
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          GNA13 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GNA13 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          GNA13 Antibody (CL488-67188) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed SH-SY5Y cells using CoraLite® Plus 488 GNA13 antibody (CL488-67188, Clone: 2G8F10) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GNA13 Antibody in Immunocytochemistry (ICC/IF)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          CL488-67188
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2G8F10
          Immunogen
          GNA13 fusion protein Ag7316
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MADFLPSRS VLSVCFPGCL LTSGEAEQQR KSKEIDKCLS REKTYVKRLV KILLLGAGES GKSTFLKQMR IIHGQDFDQR AREEFRPTIY SNVIKGMRVL VDAREKLHIP WGDNSNQQHG DKMMSFDTRA PMAAQGMVET RVFLQYLPAI RALWADSGIQ NAYDRRREFQ LGESVKYFLD NLDKLGEPDY IPSQQDILLA RRPTKGIHEY DFEIKNVPFK MVDVGGQRSE RKRWFECFDS VTSILFLVSS SEFDQVLMED RLTNRLTESL NIFETIVNNR VFSNVSIILF LNKTDLLEEK VQIVSIKDYF LEFEGDPHCL RDVQKFLVEC FRNKRRDQQQ KPLYHHFTTA INTENIRLVF RDVKDTILHD NLKQLMLQ
          Target Information
          Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems (PubMed:15240885, PubMed:16787920, PubMed:16705036, PubMed:27084452). Activates effector molecule RhoA by binding and activating RhoGEFs (ARHGEF1/p115RhoGEF, ARHGEF11/PDZ-RhoGEF and ARHGEF12/LARG) (PubMed:15240885, PubMed:12515866). GNA13-dependent Rho signaling subsequently regulates transcription factor AP-1 (activating protein-1). Promotes tumor cell invasion and metastasis by activating RhoA/ROCK signaling pathway (PubMed:16787920, PubMed:16705036, PubMed:27084452). Inhibits CDH1-mediated cell adhesion in process independent from Rho activation (PubMed:11976333). [UniProt]
          Bioinformatics
          Protein Aliases: G alpha 13; G alpha-13; G protein; G protein family member; G-protein subunit alpha-13; Galpha 13; guanine nucleotide binding protein (G protein), alpha 13; guanine nucleotide binding protein, alpha 13; heterotrimeric guanine nucleotide-binding protein 1N; unnamed protein product
          View more View less
          Gene Aliases: G13; Galpha13; HG1N
          View more View less
          UniProt ID: (Human) Q14344, (Rat) Q6Q7Y5, (Mouse) P27601
          View more View less
          Entrez Gene ID: (Human) 10672, (Pig) 100522824, (Rat) 303634, (Mouse) 14674
          View more View less
          Function(s)
          G-protein coupled receptor binding GTPase activity obsolete small monomeric GTPase activity guanyl-nucleotide exchange factor activity protein binding guanyl nucleotide binding G-protein beta/gamma-subunit complex binding D5 dopamine receptor binding heterotrimeric G-protein
          Process(es)
          signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway Rho protein signal transduction regulation of blood pressure regulation of small GTPase mediated signal transduction regulation of postsynapse assembly Rho-activating G protein-coupled receptor signaling pathway angiogenesis patterning of blood vessels in utero embryonic development positive regulation of cytosolic calcium ion concentration regulation of cell shape positive regulation of phospholipase activity cell differentiation platelet activation regulation of cell migration intracellular signal transduction negative regulation of vascular smooth muscle cell proliferation negative regulation of vascular associated smooth muscle cell migration
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0