    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SOX9 Antibodies
          Proteintech
          SOX9 Monoclonal Antibody (1H9A6), CoraLite® Plus 488
          View all (66) SOX9 antibodies
          Questions & Answers
          Cite SOX9 Monoclonal Antibody (1H9A6), CoraLite® Plus 488
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          SOX9 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          SOX9 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SOX9 Antibody (CL488-67439) in Flow
          1x10^6 HeLa cells were intracellularly stained with 0.8 µg CoraLite® Plus 488 Anti-Human SOX9 (CL488-67439, Clone: 1H9A6) (red), or 0.8 µg CoraLite® Plus 488 Mouse IgG1 Isotype Control (MOPC-21) (CL488-65124, Clone: MOPC-21) (blue). Cells were fixed and permeabilized with Transcription Factor Staining Buffer Kit (PF00011). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SOX9 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          CL488-67439
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1H9A6
          Immunogen
          SOX9 fusion protein Ag28620
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • CoraLite Plus 647
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          0.94 mg/mL
          Amount
          94 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: ADSPHSSSG MSEVHSPGEH SGQSQGPPTP PTTPKTDVQP GKADLKREGR PLPEGGRQPP IDFRDVDIGE LSSDVISNIE TFDVNEFDQY LPPNGHPGVP ATHGQVTYTG SYGISSTAAT PASAGHVWMS KQQAPPPPPQ QPPQAPPAPQ APPQPQAAPP QQPAAPPQQP QAHTLTTLSS EPGQSQRTHI KTEQLSPSHY SEQQQHSPQQ IAYSPFNLPH YSPSYPPITR SQYDYTDHQN SSSYYSHAAG QGTGLYSTFT YMNPAQRPMY TPIADTSGVP SIPQTHSPQH WEQPVYTQLT RP
          Target Information
          SOX9 has a role in sex determination and differentiation of Sertoli cells. It is involved in chondrogenesis and regulates the expression of other genes involved in chondrogenesis by acting as a transcription factor for these genes. Translocation of this gene can cause campomelic dysplasia. SOX9 is involved in the formation of testes from the indifferent fetal gonads. It is a major molecular component of the neuron-glia switch in developing spinal cord. During normal development, SOX9 allows the prostate epithelium to outgrow into the mesenchyme and then provides basal cell support for development and maintenance of the luminal epithelium. These functions of SOX9 are subverted in prostate cancer to support tumor growth and invasion. SOX9 may direct the formation of neural crest precursors and the development of a range of neural crest derivative. It has a transcriptional regulation in melanin production in cells. Northern blot analysis shows its expression on adult testis, adult heart, and fetal brain.
          Bioinformatics
          Protein Aliases: SOX 9; SOX9; SRY (sex determining region Y)-box9; SRY (sex-determining region Y)-box 9 protein; SRY-box 9; SRY-Box 9 ; SRY-box containing gene 9; SRY-related HMG-box, gene 9; unnamed protein product
          View more View less
          Gene Aliases: 2010306G03Rik; CMD1; CMPD1; ENH13; mKIAA4243; mSox9; SRA1; SRXX2; SRXY10; TESCO
          View more View less
          UniProt ID: (Human) P48436, (Mouse) Q04887
          View more View less
          Entrez Gene ID: (Human) 6662, (Mouse) 20682
          View more View less
          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding beta-catenin binding protein kinase A catalytic subunit binding bHLH transcription factor binding sequence-specific DNA binding pre-mRNA intronic binding sequence-specific double-stranded DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter skeletal system development cartilage condensation ossification branching involved in ureteric bud morphogenesis cell fate specification epithelial to mesenchymal transition tissue homeostasis hair follicle development morphogenesis of an epithelium positive regulation of mesenchymal cell proliferation chondrocyte differentiation negative regulation of immune system process heart valve development heart valve morphogenesis aortic valve morphogenesis endocardial cushion morphogenesis chondrocyte differentiation involved in endochondral bone morphogenesis chondrocyte hypertrophy growth plate cartilage chondrocyte growth nucleosome assembly chromatin remodeling signal transduction epidermal growth factor receptor signaling pathway spermatogenesis central nervous system development heart development positive regulation of cell proliferation male gonad development gene expression regulation of cell cycle process positive regulation of gene expression positive regulation of epithelial cell migration neural crest cell fate specification male germ-line sex determination negative regulation of ossification prostate gland development positive regulation of epithelial cell differentiation mammary gland development notochord development otic vesicle formation negative regulation of chondrocyte differentiation positive regulation of chondrocyte differentiation somatic stem cell population maintenance regulation of cell proliferation regulation of apoptotic process negative regulation of apoptotic process negative regulation of myoblast differentiation negative regulation of osteoblast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of fatty acid oxidation negative regulation of photoreceptor cell differentiation oligodendrocyte differentiation positive regulation of epithelial cell proliferation negative regulation of epithelial cell proliferation cartilage development Sertoli cell differentiation Sertoli cell development retina development in camera-type eye limb bud formation retinal rod cell differentiation epithelial cell proliferation involved in prostatic bud elongation intestinal epithelial structure maintenance regulation of cell proliferation involved in tissue homeostasis positive regulation of cartilage development morphogenesis of a branching epithelium protein-containing complex assembly negative regulation of biomineral tissue development ERK1 and ERK2 cascade response to fatty acid cellular response to mechanical stimulus cellular response to retinoic acid cellular response to interleukin-1 cellular response to epidermal growth factor stimulus cellular response to heparin cellular response to transforming growth factor beta stimulus otic vesicle development cellular response to BMP stimulus renal vesicle induction metanephric tubule development ureter urothelium development metanephric nephron tubule formation negative regulation of canonical Wnt signaling pathway cochlea morphogenesis positive regulation of kidney development positive regulation of branching involved in ureteric bud morphogenesis positive regulation of chondrocyte proliferation negative regulation of pri-miRNA transcription from RNA polymerase II promoter negative regulation of beta-catenin-TCF complex assembly positive regulation of male gonad development positive regulation of mesenchymal stem cell differentiation positive regulation of protein phosphorylation chondrocyte development heart valve formation regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter cytoskeleton organization Notch signaling pathway negative regulation of cell proliferation regulation of gene expression negative regulation of gene expression neural crest cell development cell differentiation regulation of cell adhesion extracellular matrix organization male sex determination negative regulation of bone mineralization negative regulation of epithelial cell differentiation endocrine pancreas development lacrimal gland development protein localization to nucleus intrahepatic bile duct development cell fate commitment regulation of cell differentiation positive regulation of protein catabolic process homeostasis of number of cells within a tissue astrocyte fate commitment endochondral bone morphogenesis epithelial tube branching involved in lung morphogenesis lung epithelial cell differentiation prostate gland morphogenesis bronchus cartilage development trachea cartilage development regulation of branching involved in lung morphogenesis lung smooth muscle development Harderian gland development ureter development ureter smooth muscle cell differentiation ureter morphogenesis anterior head development cell-cell adhesion positive regulation of extracellular matrix assembly positive regulation of cell proliferation involved in heart morphogenesis regulation of epithelial cell proliferation involved in lung morphogenesis negative regulation of mesenchymal cell apoptotic process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0