    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CNOT4 Antibodies
          Proteintech
          CNOT4 Recombinant Rabbit Monoclonal Antibody (240559E1), CoraLite® Plus 488
          View all (40) CNOT4 antibodies
          Questions & Answers
          Cite CNOT4 Recombinant Rabbit Monoclonal Antibody (240559E1), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CNOT4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CNOT4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CNOT4 Antibody (CL488-83615-4) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 488 CNOT4 antibody (CL488-83615-4, Clone: 240559E1) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CNOT4 Antibody in Immunocytochemistry (ICC/IF)
          CNOT4 Recombinant Rabbit Monoclonal Antibody (240559E1), CoraLite® Plus 488
          Product Details
          CL488-83615-4
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240559E1
          Immunogen
          CNOT4 fusion protein Ag3163
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: IDKPSDSLSI GNGDNSQQIS NSDTPSPPPG LSKSNPVIPI SSSNHSARSP FEGAVTESQS LFSDIFRHPN PIPSGLPPFP SSPQTSSDWP TAPEPQSLFT SETIPVSSST DWQAAFGFGS SKQPEDDLGF DPFDVTRKAL ADLIEKELSV QDQPSLSPTS LQNSSSHTTT AKGPGSGFLH PAAATNANSL NSTFSVLPQR FPQFQQHRAV YNSFSFPGQA ARYPWMAFPR NSIMHLNHTA NPTSNSNFLD LNLPPQHNTG LGGIPVAGEE EVKVSTMPLS TSSHSLQQGQ QPTSLHTTVA
          Target Information
          CNOT4 is a 575 amino acid protein with E3 ubiquitin ligase activity, mainly involved in protein modification and protein ubiquitination. It contains a ring type zinc finger domain of the C4C4 type, an RNA recognition motif and a bipartite nuclear localization signal.Studies show that CNOT4 is a component of the CCR4-NOT (CAF1) core complex that functions as general transcription regulation complex and has both a positive as well as a negative effect on the transcription of multiple functionally unrelated genes, thus controls gene expression. CNOT4 interacts with CNOT1 via C-terminus and with E2 ubiquitin ligases via C4C4 RING domain. Reports suggest that CNOT4 gets phosphorylated in response to DNA damage on consensus sites recognized by ATM and ATR.
          Bioinformatics
          Protein Aliases: CCR4-associated factor 4; CCR4-NOT transcription complex subunit 4; E3 ubiquitin-protein ligase CNOT4; general negative regulator of transcription subunit 4; NLS-containing variant; negative regulator of transcription 4; NOT4 (negative regulator of transcription 4, yeast) homolog; Potential transcriptional repressor NOT4Hp; RING-type E3 ubiquitin transferase CNOT4; transcriptional repressor NOT4Hp; unnamed protein product
          View more View less
          Gene Aliases: CLONE243; CNOT4; NOT4; NOT4H; Not4hp
          View more View less
          UniProt ID: (Human) O95628, (Mouse) Q8BT14
          View more View less
          Entrez Gene ID: (Human) 4850, (Mouse) 53621
          View more View less
          Function(s)
          nucleic acid binding RNA binding ubiquitin-protein transferase activity protein binding zinc ion binding transferase activity metal ion binding ubiquitin protein ligase activity nucleotide binding UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity ligase activity ribosomal S6-glutamic acid ligase activity coenzyme F420-0 gamma-glutamyl ligase activity coenzyme F420-2 alpha-glutamyl ligase activity poly(A) RNA binding protein-glycine ligase activity protein-glycine ligase activity, initiating protein-glycine ligase activity, elongating tubulin-glycine ligase activity protein-glutamic acid ligase activity tubulin-glutamic acid ligase activity ubiquitin-protein ligase
          Process(es)
          nuclear-transcribed mRNA poly(A) tail shortening ubiquitin-dependent protein catabolic process protein ubiquitination regulation of megakaryocyte differentiation protein autoubiquitination transcription, DNA-templated regulation of transcription, DNA-templated
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline