    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FBXO32 Antibodies
          Proteintech
          FBXO32 Monoclonal Antibody (3C2G3), CoraLite® 594
          View all (23) FBXO32 antibodies
          Questions & Answers
          Cite FBXO32 Monoclonal Antibody (3C2G3), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          FBXO32 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          FBXO32 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          FBXO32 Antibody (CL594-67172) in IHC (PFA)
          Immunofluorescent analysis of (4% PFA) fixed mouse skeletal muscle tissue using CoraLite®594 FBXO32 antibody (CL594-67172, Clone: 3C2G3) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FBXO32 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          FBXO32 Monoclonal Antibody (3C2G3), CoraLite® 594
          Product Details
          CL594-67172
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1:200
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3C2G3
          Immunogen
          FBXO32 fusion protein Ag25247
          3D Epitope / Immunogen
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: IAQKNFMNIL EKVVLKVLED QQNIRLIREL LQTLYTSLCT LVQRVGKSVL VGNINMWVYR METILHWQQQ LNNIQITRPA FKGLTFTDLP LCLQLNIMQR LSDGRDLVSL GQAAPDLHVL SEDRLLWKKL CQYHFSERQI RKRLILSDKG QLDWKKMYFK LVRCYPRKEQ YGDTLQLRKH CHILSWKGTD HPCTANNPES CSVSLSPQDF INLFKF
          Target Information
          This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs, which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and contains an F-box domain. This protein is highly expressed during muscle atrophy, whereas mice deficient in this gene were found to be resistant to atrophy. This protein is thus a potential drug target for the treatment of muscle atrophy. Alternative splicing of this gene results in two transcript variants encoding two isoforms of different sizes.
          Bioinformatics
          Protein Aliases: atrogin 1; Atrogin-1; F-box only protein 32; fbxo25; FLJ32424; MAFbx; MGC108443; MGC137646; MGC33610; Muscle atrophy F-box protein; SCF ubiquitin ligase; unnamed protein product
          View more View less
          Gene Aliases: 4833442G10Rik; AI430017; ATROGIN1; Fbx32; FBXO32; Gm20361; MAFbx
          View more View less
          UniProt ID: (Human) Q969P5, (Mouse) Q9CPU7
          View more View less
          Entrez Gene ID: (Human) 114907, (Mouse) 67731
          View more View less
          Function(s)
          protein binding
          Process(es)
          response to denervation involved in regulation of muscle adaptation protein ubiquitination cellular response to dexamethasone stimulus response to electrical stimulus involved in regulation of muscle adaptation muscle atrophy
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline