    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MTA3 Antibodies
          Proteintech
          MTA3 Recombinant Rabbit Monoclonal Antibody (230350D12), CoraLite® Plus 750
          View all (16) MTA3 antibodies
          Questions & Answers
          Cite MTA3 Recombinant Rabbit Monoclonal Antibody (230350D12), CoraLite® Plus 750
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          MTA3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MTA3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          MTA3 Antibody (CL750-83042-3) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL750-83042-3 (MTA3 antibody) at dilution of 1:4,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MTA3 Antibody in Western Blot (WB)
          MTA3 Antibody in Flow Cytometry (Flow)
          MTA3 Recombinant Rabbit Monoclonal Antibody (230350D12), CoraLite® Plus 750
          Product Details
          CL750-83042-3
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:8,000
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230350D12
          Immunogen
          MTA3 fusion protein Ag6400
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAANMYRVGD YVYFENSSSN PYLIRRIEEL NKTASGNVEA KVVCFYRRRD ISNTLIMLAD KHAKEIEEES ETTVEADLTD KQKHQLKHRE LFLSRQYESL PATHIRGKCS VALLNETESV LSYLDKEDTF FYSLVYDPSL KTLLADKGEI RVGPRYQADI PEMLLEGESD EREQSKLEVK VWDPNSPLTD RQIDQFLVVA RAVGTFARAL DCSSSVRQPS LHMSAAAASR DITLFHAMDT LYRHSYDLSS AISVLVPLGG PVLCRDEMEE WSASEASLFE EALEKYGKDF NDIRQDFLPW KSLTSIIEYY YMWKTTDRYV QQKRLKAAEA ESKLKQVYIP TYS
          Target Information
          Plays a role in maintenance of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels. Contributes to transcriptional repression by BCL6. [UniProt]
          Bioinformatics
          Protein Aliases: metastasis associated gene family, member 3; Metastasis-associated protein MTA3; unnamed protein product
          View more View less
          Gene Aliases: 1110002J22Rik; KIAA1266; mKIAA1266; MTA3
          View more View less
          UniProt ID: (Human) Q9BTC8, (Mouse) Q924K8
          View more View less
          Entrez Gene ID: (Human) 57504, (Mouse) 116871
          View more View less
          Function(s)
          DNA binding chromatin binding transcription coactivator activity transcription corepressor activity protein binding zinc ion binding histone deacetylase binding sequence-specific DNA binding macromolecular complex binding metal ion binding transcription factor activity, sequence-specific DNA binding homeodomain transcription factor
          Process(es)
          G2/M transition of mitotic cell cycle negative regulation of transcription from RNA polymerase II promoter chromatin remodeling regulation of transcription, DNA-templated cell proliferation positive regulation of G2/M transition of mitotic cell cycle regulation of cell fate specification negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of granulosa cell proliferation granulosa cell proliferation regulation of stem cell differentiation positive regulation of cell proliferation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-7c59b9b899-crt6c:80/100.66.78.36:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline