    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SAR1A Antibodies
          Proteintech
          SAR1A Recombinant Rabbit Monoclonal Antibody (230471A11), CoraLite® Plus 750
          View all (14) SAR1A antibodies
          Questions & Answers
          Cite SAR1A Recombinant Rabbit Monoclonal Antibody (230471A11), CoraLite® Plus 750
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          SAR1A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SAR1A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SAR1A Antibody (CL750-83168-6) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL750-83168-6 (SAR1A antibody) at dilution of 1:500 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SAR1A Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          CL750-83168-6
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230471A11
          Immunogen
          Peptide
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          Involved in transport from the endoplasmic reticulum to the Golgi apparatus. Required to maintain SEC16A localization at discrete locations on the ER membrane perhaps by preventing its dissociation. SAR1A-GTP-dependent assembly of SEC16A on the ER membrane forms an organized scaffold defining endoplasmic reticulum exit sites (ERES). [UniProt]
          Bioinformatics
          Protein Aliases: COPII-associated small GTPase; GTP-binding protein SAR1a; GTPBP2; SAR1 gene homolog A; SAR1 homolog A; SAR1a gene homolog 1; SARA1; unnamed protein product
          View more View less
          Gene Aliases: 1600019H17Rik; masra2; SAR1; Sara; SARA1
          View more View less
          UniProt ID: (Human) Q9NR31
          View more View less
          Entrez Gene ID: (Human) 56681, (Mouse) 20224
          View more View less
          Function(s)
          GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding amino acid sensor activity
          Process(es)
          intracellular protein transport ER to Golgi vesicle-mediated transport vesicle organization vesicle-mediated transport COPII vesicle coating membrane organization cargo loading into COPII-coated vesicle regulation of cargo loading into COPII-coated vesicle negative regulation of cargo loading into COPII-coated vesicle regulation of TORC1 signaling negative regulation of TORC1 signaling cellular response to leucine starvation lipoprotein transport positive regulation of protein exit from endoplasmic reticulum positive regulation of ER to Golgi vesicle-mediated transport
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch