This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
CLASRP is probably functions as an alternative splicing regulator. CLASRP may regulate the mRNA splicing of genes such as CLK1. This may act by regulating members of the CLK kinase family.