Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CLIP2 Antibodies

          Abnova

          CLIP2 Polyclonal Antibody, MaxPab™

          View all (9) CLIP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CLIP2 Polyclonal Antibody, MaxPab™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          CLIP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CLIP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CLIP2 Antibody (H00007461-B01P) in WB

          Western Blot analysis of CLIP2 expression in transfected 293T cell line by CLIP2 MaxPab polyclonal antibody. Lane 1: CLIP2 transfected lysate(115.06 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CLIP2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          H00007461-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CLIP2 (AAI48703.1, 1 a.a. approximately 1046 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MQKPSGLKPP GRGGKHSSPM GRTSTGSASS SAAVAASSKE GSPLHKQSSG PSSSPAAAAA PEKPGPKAAE VGDDFLGDFV VGERVWVNGV KPGVVQYLGE TQFAPGQWAG VVLDDPVGKN DGAVGGVRYF ECPALQGIFT RPSKLTRQPT AEGSGSDAHS VESLTAQNLS LHSGTATPPL TSRVIPLRES VLNSSVKTGN ESGSNLSDSG SVKRGEKDLR LGDRVLVGGT KTGVVRYVGE TDFAKGEWCG VELDEPLGKN DGAVAGTRYF QCPPKFGLFA PIHKVIRIGF PSTSPAKAKK TKRMAMGVSA LTHSPSSSSI SSVSSVASSV GGRPSRSGLL TETSSRYARK ISGTTALQEA LKEKQQHIEQ LLAERDLERA EVAKATSHIC EVEKEIALLK AQHEQYVAEA EEKLQRARLL VESVRKEKVD LSNQLEEERR KVEDLQFRVE EESITKGDLE TQTQLEHARI GELEQSLLLE KAQAERLLRE LADNRLTTVA EKSRVLQLEE ELTLRRGEIE ELQQCLLHSG PPPPDHPDAA EILRLRERLL SASKEHQRES GVLRDKYEKA LKAYQAEVDK LRAANEKYAQ EVAGLKDKVQ QATSENMGLM DNWKSKLDSL ASDHQKSLED LKATLNSGPG AQQKEIGELK AVMEGIKMEH QLELGNLQAK HDLETAMHVK EKEALREKLQ EAQEELAGLQ RHWRAQLEVQ ASQHRLELQE AQDQRRDAEL RVHELEKLDV EYRGQAQAIE FLKEQISLAE KKMLDYERLQ RAEAQGKQEV ESLREKLLVA ENRLQAVEAL CSSQHTHMIE SNDISEETIR TKETVEGLQD KLNKRDKEVT ALTSQTEMLR AQVSALESKC KSGEKKVDAL LKEKRRLEAE LETVSRKTHD ASGQLVLISQ ELLRKERSLN ELRVLLLEAN RHSPGPERDL SREVHKAEWR IKEQKLKDDI RGLREKLTGL DKEKSLSDQR RYSLIDPSSA PELLRLQHQL MSTEDALRDA LDQAQQVEKL MEAMRSCPDK AQTIGNSGSA NGIHQQDKAQ KQEDKH

          Target Information

          The protein encoded by this gene belongs to the family of cytoplasmic linker proteins, which have been proposed to mediate the interaction between specific membranous organelles and microtubules. This protein was found to associate with both microtubules and an organelle called the dendritic lamellar body. This gene is hemizygously deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cytoplasmic linker 2; cytoplasmic linker protein 115; cytoplasmic linker protein 2; KIAA0291; testicular tissue protein Li 40; williams-Beuren syndrome chromosomal region 3 protein; williams-Beuren syndrome chromosomal region 4 protein; Williams-Beuren syndrome chromosome region 3; Williams-Beuren syndrome chromosome region 4

          View more View less

          Gene Aliases: CLIP; CLIP-115; CYLN2; WBSCR3; WBSCR4; WSCR3; WSCR4

          View more View less

          UniProt ID: (Human) Q9UDT6

          View more View less

          Entrez Gene ID: (Human) 7461, (Rat) 29264

          View more View less

          Function(s)
          microtubule binding microtubule plus-end binding protein binding
          Process(es)
          negative regulation of microtubule depolymerization cytoplasmic microtubule organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch