Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MHHCKRYRSP EPDPYLSYRW KRRRSYSREH EGRLRYPSRR EPPPRRSRSR SHDRLPYQRR YRERRDSDTY RCEERSPSFG EDYYGPSRSR HRRRSRERGP YRTRKHAHHC HKRRTRSCSS ASSRSQQSSK RSSRS
CLK3 is a dual specificity kinase that can interact with and cause the nuclear redistribution of SR proteins. Two transcript variants encoding different isoforms have been found for CLK3. Related pseudogenes are located on chromosomes 1 and 9.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cdc2/CDC28-like protein kinase 3; clk3-152; putative; clk3-490; putative; Protein kinase CLK3; SR protein kinase; unnamed protein product
Gene Aliases: PHCLK3; PHCLK3/152
UniProt ID: (Human) P49761, (Mouse) O35492
Entrez Gene ID: (Human) 1198, (Mouse) 102414
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support