Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: AAIRRKENG WYDEEHPLVF LFLGSSGIGK TELAKQTAKY MHKDAKKGFI RLDMSEFQER HEVAKFIGSP PGYVGHEEGG QLTKKLKQCP NAVVLFDEVD KAHPDVLTIM LQLFDEGRLT DGKGKTIDCK DAIFIMTSNV ASDEIAQHAL QLRQEALEMS RNRIAENLGD VQISDKITIS KNFKENVIRP ILKAHFRRDE FLGRINEIVY FLPFCHSELI QLVNKELNFW AKRAKQRHNI TLLWDREVAD VLVDGYNVHY GARSIKHEVE RRVVNQLAAA YEQDLLPGGC TLRITVEDSD KQLLKSPELP SPQAEKRLPK LRLEIIDKDS KTRRLDIRAP LHPEKVCNTI (359-707 aa encoded by BC006404)
CLPB may function as a regulatory ATPase and be related to secretion/protein trafficking process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin-repeat containing bacterial clp fusion; caseinolytic mitochondrial matrix peptidase chaperone subunit B; caseinolytic peptidase B protein homolog; ClpB caseinolytic peptidase B homolog; ClpB homolog, mitochondrial AAA ATPase chaperonin; SKD3; suppressor of K+ transport defect 3; suppressor of potassium transport defect 3; testicular secretory protein Li 11; unnamed protein product
Gene Aliases: ANKCLB; ANKCLP; HSP78; MEGCANN; MGCA7; MGCA7A; SCN9; SKD3
UniProt ID: (Human) Q9H078, (Mouse) Q60649, (Rat) Q9WTT2
Entrez Gene ID: (Human) 81570, (Mouse) 20480, (Rat) 65041
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support