Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Coronin 3 Antibodies

          Proteintech

          CORO1C Polyclonal Antibody

          16 References
          View all (7) Coronin 3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CORO1C Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          CORO1C Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CORO1C Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CORO1C Antibody (14749-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using CORO1C antibody 14749-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CORO1C Antibody in Immunocytochemistry (ICC/IF)
          CORO1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CORO1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CORO1C Antibody in Western Blot (WB)
          CORO1C Antibody in Western Blot (WB)
          CORO1C Antibody in Western Blot (WB)
          CORO1C Antibody in Western Blot (WB)
          CORO1C Antibody in Western Blot (WB)
          CORO1C Antibody in Western Blot (WB)
          CORO1C Antibody in Immunoprecipitation (IP)
          CORO1C Polyclonal Antibody

          Product Details

          14749-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 11 publications 11 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Monkey, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CORO1C Fusion Protein Ag6488 (1-306 aa encoded by BC002342)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.22 mg/mL

          Amount

          33 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MRRVVRQSK FRHVFGQAVK NDQCYDDIRV SRVTWDSSFC AVNPRFVAII IEASGGGAFL VLPLHKTGRI DKSYPTVCGH TGPVLDIDWC PHNDQVIASG SEDCTVMVWQ IPENGLTLSL TEPVVILEGH SKRVGIVAWH PTARNVLLSA GCDNAIIIWN VGTGEALINL DDMHSDMIYN VSWNRNGSLI CTASKDKKVR VIDPRKQEIV AEKEKAHEGA RPMRAIFLAD GNVFTTGFSR MSERQLALWN PKNMQEPIAL HEMDTSNGVL LPFYDPDTSI IYLCGKGDSS IRYFEITDES PYVHYLN (1-306 aa encoded by BC002342)

          Target Information

          Coronin 3 is associated with the aggression and metastasis of hepatocellular carcinoma. Coronins are a family of highly evolutionary conserved proteins reportedly involved in the regulation of actin cytoskeletal dynamics, although only coronin 3 has been shown to be related to cancer cell migration.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: coronin 1c; coronin 3; coronin, actin binding protein, 1C; Coronin-1C; Coronin-3; hCRNN4; unnamed protein product

          View more View less

          Gene Aliases: AL022675; AW455561; AW548837; CORO1C; CRN2; CRNN4; HCRNN4; RGD1564490

          View more View less

          UniProt ID: (Human) Q9ULV4, (Mouse) Q9WUM4

          View more View less

          Entrez Gene ID: (Human) 23603, (Rat) 501841, (Mouse) 23790

          View more View less

          Function(s)
          actin binding protein binding small GTPase binding actin filament binding Rac GTPase binding non-motor actin binding protein
          Process(es)
          neural crest cell migration regulation of protein phosphorylation negative regulation of protein phosphorylation phagocytosis actin filament organization signal transduction regulation of epithelial cell migration negative regulation of epithelial cell migration regulation of fibroblast migration endosomal transport ventricular system development corpus callosum development negative regulation of protein kinase activity by regulation of protein phosphorylation establishment of protein localization positive regulation of osteoclast differentiation macrophage chemotaxis negative regulation of cellular component organization regulation of focal adhesion assembly negative regulation of focal adhesion assembly membrane fission activation of GTPase activity endosome membrane tubulation endosome fission regulation of substrate adhesion-dependent cell spreading negative regulation of substrate adhesion-dependent cell spreading regulation of ruffle assembly positive regulation of lamellipodium morphogenesis actin cytoskeleton organization actin cortical patch assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0