Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: SAHKTKHDL MMKGEGSRKG SFFKQAKKSY PMFPAPEERI KWDEYGEIIK PEDFLVPELQ ATEEEKSKLE SGLTNGDEPM DQDLSDVPTK CISTTESIEI KARVTYIDYE GRSDGDSIKK IINQMKPRQL IIVHGPPEAS QDLAECCRAF GGKDIKVYMP KLHETVDATS ETHIYQVRLK DSLVSSLQFC KAKDAELAWI DGVLDMRVSK VDTGVILEEG ELRDDGEDSE MQVEAPSDSS VIAQQKAMKS LFGDDEKETG EESEIIPTLE PLPPHEVPGH QSVFMNEPRL SDFKQVLLRE GIQAEFVGGV LVCNNQVAVR RTETGRIGLE GCLCQDFYRI RDLLYEQYAI V (433-782 aa encoded by BC070095)
CPSF2 gene ontology annotations related to this gene include mRNA cleavage; mRNA polyadenylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cleavage and polyadenylation specific factor 2, 100kD subunit; cleavage and polyadenylation specific factor 2, 100kDa; cleavage and polyadenylation specificity factor 100 kDa subunit; CPSF 100 kDa subunit; CPSF 100kDa subunit; unnamed protein product
Gene Aliases: 100kDa; 2610024B04Rik; Cpsf; CPSF100; MCPSF; mKIAA1367
UniProt ID: (Human) Q9P2I0, (Mouse) O35218
Entrez Gene ID: (Human) 53981, (Rat) 299256, (Mouse) 51786
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support