    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Collagen V Antibodies
          Invitrogen
          Collagen V (soluble) Monoclonal Antibody (1E2-E4/Col5)
          View all (5) Collagen V antibodies
          Questions & Answers
          Cite Collagen V (soluble) Monoclonal Antibody (1E2-E4/Col5)
          Additional Information:
          {{banner.description}}
          Product Details
          CSI 006-01-02
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          ELISA (ELISA)
          1:50
          -
          Product Specifications
          Species Reactivity
          Bovine, Dog, Human, Marsupial, Sheep, Pig, Rabbit
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1E2-E4/Col5
          Immunogen
          Acid-digested pepsin soluble dog type V collagen.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.05 mg/mL
          Amount
          420 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1073787
          Product Specific Information
          CSI 006-01-02 is highly specific for type V collagen. It has been shown to have no cross-reactivity with type I, III and VI collagens by ELISA and immunoblotting. There is no evidence for cross-reactivity with other connective tissue proteins (laminin, fibronectin, elastin). CSI 006-01-02 shows reactivity with human, dog, calf, sheep, pig and kangaroo in type V collagens by ELISA and immunoblotting. CSI 006-01-02 does not show reactivity with mouse, rat, guinea pig and chicken type V collagens by ELISA and immunoblotting.
          CSI 006-01-02 has been successfully used in Western blotting, ELISA and Immunohistochemistry procedures. CSI 006-01 has been used succesfully for immunohistology on frozen unfixed sections of human, bovine and dog skin, of foetal bovine cornea, and of new dog tissue associated with a biomaterial implant. If fixation of tissue is required, acetone or ethanol is recommended. CSI 006-01 binds poorly to Collagen V when tested in ELISA with Collagen V coated directly onto the microtiter well.
          NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
          Target Information
          COL5A1 encodes an alpha chain for one of the lowabundance fibrillar collagens. Fibrillar collagen molecules aretrimers that can be composed of one or more types of alpha chains.Type V collagen is found in tissues containing type I collagen andappears to regulate the assembly of heterotypic fibers composed ofboth type I and type V collagen. COL5A1 product is closelyrelated to type XI collagen and it is possible that the collagenchains of types V and XI constitute a single collagen type withtissue-specific chain combinations. Mutations in this gene areassociated with Ehlers-Danlos syndrome, types I and II.
          Bioinformatics
          Protein Aliases: AB collagen; alpha-1 type V collagen; COL5; Collagen 5; Collagen alpha-1(V) chain; collagen type 5 alpha 1; collagen, type V, alpha 1; procollagen alpha 1(V); RP11-263F14.1; Type V Collagen
          View more View less
          Gene Aliases: COL5A1; EDSC; EDSCL1; FMDMF
          View more View less
          UniProt ID: (Human) P20908
          View more View less
          Entrez Gene ID: (Human) 1289, (Pig) 397533, (Sheep) 101107777, (Dog) 480684, (Rabbit) 100327268
          View more View less
          Function(s)
          integrin binding extracellular matrix structural constituent protein binding heparin binding extracellular matrix structural constituent conferring tensile strength proteoglycan binding metal ion binding platelet-derived growth factor binding extracellular matrix structural protein
          Process(es)
          blood vessel development heart morphogenesis cell adhesion cell migration collagen fibril organization collagen biosynthetic process wound healing, spreading of epidermal cells tendon development skin development integrin biosynthetic process eye morphogenesis regulation of cellular component organization fibril organization negative regulation of endodermal cell differentiation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline