Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Cullin 4A Antibodies

          Abnova

          CUL4A Polyclonal Antibody, MaxPab™

          View all (22) Cullin 4A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CUL4A Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CUL4A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CUL4A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CUL4A Antibody (H00008451-D01P) in WB

          CUL4A MaxPab rabbit polyclonal antibody. Western Blot analysis of CUL4A expression in HeLa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CUL4A Antibody in Western Blot (WB)
          CUL4A Polyclonal Antibody, MaxPab™

          Product Details

          H00008451-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CUL4A (NP_001008895.1, 1 a.a. approximately 759 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MADEAPRKGS FSALVGRTNG LTKPAALAAA PAKPGGAGGS KKLVIKNFRD RPRLPDNYTQ DTWRKLHEAV RAVQSSTSIR YNLEELYQAV ENLCSHKVSP MLYKQLRQAC EDHVQAQILP FREDSLDSVL FLKKINTCWQ DHCRQMIMIR SIFLFLDRTY VLQNSTLPSI WDMGLELFRT HIISDKMVQS KTIDGILLLI ERERSGEAVD RSLLRSLLGM LSDLQVYKDS FELKFLEETN CLYAAEGQRL MQEREVPEYL NHVSKRLEEE GDRVITYLDH STQKPLIACV EKQLLGEHLT AILQKGLDHL LDENRVPDLA QMYQLFSRVR GGQQALLQHW SEYIKTFGTA IVINPEKDKD MVQDLLDFKD KVDHVIEVCF QKNERFVNLM KESFETFINK RPNKPAELIA KHVDSKLRAG NKEATDEELE RTLDKIMILF RFIHGKDVFE AFYKKDLAKR LLVGKSASVD AEKSMLSKLK HECGAAFTSK LEGMFKDMEL SKDIMVHFKQ HMQNQSDSGP IDLTVNILTM GYWPTYTPME VHLTPEMIKL QEVFKAFYLG KHSGRKLQWQ TTLGHAVLKA EFKEGKKEFQ VSLFQTLVLL MFNEGDGFSF EEIKMATGIE DSELRRTLQS LACGKARVLI KSPKGKEVED GDKFIFNGEF KHKLFRIKIN QIQMKETVEE QVSTTERVFQ DRQYQIDAAI VRIMKMRKTL GHNLLVSELY NQLKFPVKPG DLKKRIESLI DRDYMERDKD NPNQYHYVA

          Target Information

          CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CUL 4A; CUL-4A; Cul4a protein; Cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; ubiquitin E3 ligase; unnamed protein product

          View more View less

          Gene Aliases: 2810470J21Rik; AW495282; CUL4A

          View more View less

          UniProt ID: (Human) Q13619, (Mouse) Q3TCH7

          View more View less

          Entrez Gene ID: (Human) 8451, (Mouse) 99375

          View more View less

          Function(s)
          protein binding ubiquitin protein ligase binding ubiquitin protein ligase activity ubiquitin ligase complex scaffold activity ubiquitin-protein ligase
          Process(es)
          G1/S transition of mitotic cell cycle in utero embryonic development DNA repair ubiquitin-dependent protein catabolic process cellular response to DNA damage stimulus spermatogenesis cell proliferation positive regulation of cell proliferation protein ubiquitination hemopoiesis negative regulation of granulocyte differentiation developmental process cellular response to UV somatic stem cell population maintenance mRNA cleavage involved in gene silencing by miRNA T cell activation ribosome biogenesis proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of protein catabolic process rhythmic process regulation of protein metabolic process type I interferon signaling pathway intrinsic apoptotic signaling pathway base-excision repair, AP site formation via deaminated base removal ubiquitin-dependent protein catabolic process via the C-end degron rule pathway positive regulation of G1/S transition of mitotic cell cycle regulation of DNA damage checkpoint regulation of nucleotide-excision repair viral process protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline