Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CXCL6 Antibodies

          Invitrogen

          CXCL6 Polyclonal Antibody, Biotin, PeproTech®

          View all (11) CXCL6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CXCL6 Polyclonal Antibody, Biotin, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          CXCL6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CXCL6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CXCL6 Antibody (500-P120BT-50UG) in WB

          Western Blot: To detect hGCP-2 by Western Blot analysis CXCL6 Polyclonal Antibody, Biotin (Product # 500-P120BT-500UG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hGCP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CXCL6 Antibody in Western Blot (WB)
          CXCL6 Antibody in Western Blot (WB)
          CXCL6 Antibody in ELISA (ELISA)
          CXCL6 Polyclonal Antibody, Biotin, PeproTech®

          Product Details

          500-P120BT-50UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          ELISA (ELISA)

          0.25-1.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived Recombinant Human GCP-2 (CXCL6)
          3D Epitope / Immunogen

          Conjugate

          Biotin Biotin Biotin

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2929320

          Product Specific Information

          AA Sequence of recombinant protein: VLTELRCTCL RVTLRVNPKT IGKLQVFPAG PQCSKVEVVA SLKNGKQVCL DPEAPFLKKV IQKILDSGNK KN.

          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GCP-2 (CXCL6). Anti-Human GCP-2 (CXCL6)-specific antibody was purified by affinity chromatography and then biotinylated.

          Sandwich ELISA: To detect Human GCP-2 (CXCL6) by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human GCP-2 (CXCL6) (500-P120) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human GCP-2 (CXCL6).

          Western Blot: To detect hGCP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hGCP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          500-P120BT-1MG will be provided as 2 x 500 µg

          Target Information

          CXCL6 is a chemotactic for neutrophil granulocytes. It signals through binding and activation of its receptors (CXCR1 and CXCR2). In addition to its chemotactic and angiogenic properties, it has strong antibacterial activity against Gram-positive and Gram-negative bacteria (90-fold-higher when compared to CXCL5 and CXCL7).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 6; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); Chemokine alpha 3; CKA-3; CKA3; CXC; CXC motif chemokine; CXCL; GCP-2; Granulocyte chemotactic protein 2; small inducible cytokine subfamily B (Cys-X-Cys), member 6 (granulocyte chemotactic protein 2); small inducible cytokine subfamily B (Cys-X-Cys), member b; Small-inducible cytokine B6; unnamed protein product

          View more View less

          Gene Aliases: CKA-3; CXCL6; GCP-2; GCP2; SCYB6

          View more View less

          UniProt ID: (Human) P80162

          View more View less

          Entrez Gene ID: (Human) 6372

          View more View less

          Function(s)
          cytokine activity protein binding chemokine activity heparin binding CXCR chemokine receptor binding
          Process(es)
          leukocyte homeostasis chemotaxis defense response inflammatory response immune response signal transduction cell-cell signaling neutrophil chemotaxis response to lipopolysaccharide regulation of chemokine production neutrophil activation defense response to bacterium antimicrobial humoral immune response mediated by antimicrobial peptide chemokine-mediated signaling pathway regulation of neutrophil mediated killing of gram-negative bacterium cellular response to lipopolysaccharide antibacterial innate immune response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline