Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MWGLKVLLL PVVSFALYPE EILDTHWELW KKTHRKQYNN KVDEISRRLI WEKNLKYISI HNLEASLGVH TYELAMNHLG DMTSEEVVQK MTGLKVPLSH SRSNDTLYIP EWEGRAPDSV DYRKKGYVTP VKNQGQCGSC WAFSSVGALE GQLKKKTGKL LNLSPQNLVD CVSENDGCGG GYMTNAFQYV QKNRGIDSED AYPYVGQEES CMYNPTGKAA KCRGYREIPE GNEKALKRAV ARVGPVSVAI DASLTSFQFY SKGVYYDESC NSDNLNHAVL AVGYGIQKGN KHWIIKNSWG ENWGNKGYIL MARNKNNACG IANLASFPKM (1-329 aa encoded by BC016058)
The protein encoded by this gene is a lysosomal cysteine proteinase involved in bone remodeling and resorption. This protein, which is a member of the peptidase C1 protein family, is predominantly expressed in osteoclasts. However, the encoded protein is also expressed in a significant fraction of human breast cancers, where it could contribute to tumor invasiveness. Mutations in this gene are the cause of pycnodysostosis, an autosomal recessive disease characterized by osteosclerosis and short stature. This gene may be subject to RNA editing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cat K; cathepsin O; cathepsin O1; cathepsin O2; cathepsin X; CTSK; MGC23107; unnamed protein product
Gene Aliases: catK; CTS02; CTSO; CTSO1; CTSO2; MMS10-Q; Ms10q; PKND; PYCD
UniProt ID: (Human) P43235, (Rat) O35186, (Mouse) P55097
Entrez Gene ID: (Human) 1513, (Rat) 29175, (Mouse) 13038
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support