Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: EPRKVLLRL YGAILQVGAE AMVLESVMFA ILAERSLGPK LYGIFPQGRL EQFIPSRRLD TEELGLPDIS AEIAEKMATF HGMKMPFNKE PKWLFGTMEK YLKEVLRIKF TEESRIKKLH KLLSYNLPLE LENLRSLLES TPSPVVFCHN DCQEGNILLL EGRENSEKQK LMLIDFEYSS YNYRGFDIGN HFCEWMYDYS YEKYPFFRAN IRKYPTKKQQ LHFISSYLPA FQNDFENLST EEKSIIKEEM LLEVNRFALA SHFLWGQWSI VQAKISSIEF GYMDYAQARF DAYFHQKRKL GV (139-439 aa encoded by BC036471)
The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. Choline kinase alpha is the initial enzyme in the sequence and may play a regulatory role. This protein also catalyzes the phosphorylation of ethanolamine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHETK-alpha; choline kinase R; choline/ethanolamine kinase alpha; CK; EK; ethanolamine kinase; unnamed protein product
Gene Aliases: Chetk-alpha; CHK; ChoK; CK; CK-R; CK/EK-alpha; CKI; EK; EtnK-alpha; NEDMIMS
UniProt ID: (Human) P35790, (Rat) Q01134, (Mouse) O54804
Entrez Gene ID: (Human) 1119, (Rat) 29194, (Mouse) 12660
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support