Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The complement factor C3 consists of an alpha and a beta chain. C3 is a central factor in the complement cascade. It is central to the alternative pathway that leads to the C3 convertase C3bBb. The classical mannose binding lectin activation pathway leads to the C3 convertase C4b2a. These convertases cleave C3 resulting in C3a and C3b. Further degradation leads to the formation of the alpha chain products C3d, C3g and C3c. C3 is an acute phase protein that is produced by a wide range of tissues, including renal epithelial cells and hepatocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acylation stimulating protein; acylation-stimulating protein cleavage product; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3 convertase cleavage product; C3a anaphylatoxin; complement 3; complement component 3; complement component 3d; complement component C3; complement component C3 precursor; complement component C3a; complement component C3b; complement factor 3; CRK SH3-binding GNRP; epididymis secretory sperm binding protein Li 62p; guanine nucleotide releasing factor 2; guanine nucleotide-releasing factor 2 (specific for crk proto-oncogene); prepro-C3; preprocomplement component C3; Rap guanine nucleotide exchange factor (GEF) 1; signal P; unnamed protein product
Gene Aliases: 4932418O06Rik; AHUS5; ARMD9; ASP; C3a; C3b; C3G; C3G-1; C3G-2; CPAMD1; GRF2; HEL-S-62p; HSE-MSF; Plp
UniProt ID: (Mouse) P01027, (Rat) P01026, (Human) P01024, (Human) Q13905
Entrez Gene ID: (Mouse) 12266, (Rat) 24232, (Human) 718, (Human) 2889, (Rat) 63881, (Mouse) 107746
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support