MA1-20029 detects D-Dimer in human samples.
MA1-20029 has been successfully used in ELISA and Western blot procedures.
The MA1-20029 immunogen is homogenized fibrin clot.
Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.