Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene encodes a protein with a domain similar to one found in dendritic cells (PMID:11169400) which play a key role in antigen processing and display for immune responses. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2011].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DC-STAMP domain-containing protein 1; E3 ubiquitin-protein ligase DCST1; MLL5; RING-type E3 ubiquitin transferase; unnamed protein product
Gene Aliases: A330106H01Rik; RGD1307756; SNKY; SPE49
UniProt ID: (Human) Q5T197, (Mouse) Q059Y8
Entrez Gene ID: (Human) 149095, (Rat) 295246, (Mouse) 77772
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support