Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DIXDC1 Antibodies

          Proteintech

          DIXDC1 Polyclonal Antibody

          1 Reference
          View all (18) DIXDC1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite DIXDC1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (14)
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DIXDC1 Antibody (13816-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human heart tissue slide using 13816-1-AP ( DIXDC1 antibody) at dilution of 1:200 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIXDC1 Antibody in Western Blot (WB)
          DIXDC1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          DIXDC1 Polyclonal Antibody

          Product Details

          13816-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DIXDC1 Fusion Protein Ag4904 (330-683 aa encoded by BC033034)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: VNPEEQLII IQSRLDQSME ENQDLKKELL KCKQEARNLQ GIKDALQQRL TQQDTSVLQL KQELLRANMD KDELHNQNVD LQRKLDERNR LLGEYKKELG QKDRLLQQHQ AKLEEALRKL SDVSYHQVDL ERELEHKDVL LAHCMKREAD EATNYNSHNS QSNGFLLPTA GKGATSVSNR GTSDLQLVRD ALRSLRNSFS GHDPQHHTID SLEQGISSLM ERLHVMETQK KQERKVRVKS PRTQVGSEYR ESWPPNSKLP HSQSSPTVSS TCTKVLYFTD RSLTPFMVNI PKRLEEVTLK DFKAAIDREG NHRYHFKALD PEFGTVKEEI FHDDDAIPGW EGKIVAWVEE DHGEN (330-683 aa encoded by BC033034)

          Target Information

          This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: actin-binding protein; DIXDC1L; coiled-coil protein DIX1; coiled-coil-DIX1; DIX domain-containing protein 1; unnamed protein product

          View more View less

          Gene Aliases: 4930563F16Rik; CCD1; Ccd1Aalpha1L; Ccd1Abeta1L; Ccd1BalphaL; Ccd1BbetaL; Ccd1CL; mKIAA1735

          View more View less

          UniProt ID: (Human) Q155Q3, (Mouse) Q80Y83, (Rat) Q2VUH7

          View more View less

          Entrez Gene ID: (Human) 85458, (Mouse) 330938, (Rat) 363062

          View more View less

          Function(s)
          protein binding protein domain specific binding gamma-tubulin binding mitogen-activated protein kinase kinase kinase binding
          Process(es)
          Wnt signaling pathway cerebellar cortex development cerebral cortex cell migration cerebral cortex radially oriented cell migration cell proliferation in forebrain forebrain ventricular zone progenitor cell division positive regulation of Wnt signaling pathway regulation of actin cytoskeleton organization negative regulation of neuron differentiation positive regulation of JNK cascade positive regulation of axonogenesis canonical Wnt signaling pathway regulation of microtubule cytoskeleton organization modification of postsynaptic actin cytoskeleton
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch