Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DLL4 Antibodies

          Proteintech

          DLL4 Polyclonal Antibody

          16 References
          View all (38) DLL4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite DLL4 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          DLL4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DLL4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DLL4 Antibody (21584-1-AP) in WB

          Transfected HEK-293 cells were subjected to SDS PAGE followed by western blot with 21584-1-AP ( DLL4 Antibody) at dilution of 1:1000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DLL4 Antibody in Western Blot (WB)
          DLL4 Antibody in Western Blot (WB)
          DLL4 Antibody in Western Blot (WB)
          DLL4 Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          DLL4 Polyclonal Antibody

          Product Details

          21584-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 13 publications 13 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DLL4 Fusion Protein Ag16140 (317-530 aa encoded by BC106950)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.22 mg/mL

          Amount

          33 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: TGVDCELEL SECDSNPCRN GGSCKDQEDG YHCLCPPGYY GLHCEHSTLS CADSPCFNGG SCRERNQGAN YACECPPNFT GSNCEKKVDR CTSNPCANGG QCLNRGPSRM CRCRPGFTGT YCELHVSDCA RNPCAHGGTC HDLENGLMCT CPAGFSGRRC EVRTSIDACA SSPCFNRATC YTDLSTDTFV CNCPYGFVGS RCEFPVGLPP SFPWV (317-530 aa encoded by BC106950)

          Target Information

          Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: delta 4; delta ligand 4; delta-like 4; delta-like 4 homolog; delta-like 4 protein; delta-like ligand 4; drosophila Delta homolog 4; MGC126344; notch ligand delta-2; notch ligand DLL4; unnamed protein product

          View more View less

          Gene Aliases: AOS6; Delta4; hdelta2

          View more View less

          UniProt ID: (Human) Q9NR61, (Mouse) Q9JI71

          View more View less

          Entrez Gene ID: (Human) 54567, (Mouse) 54485

          View more View less

          Function(s)
          Notch binding calcium ion binding protein binding receptor agonist activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter angiogenesis patterning of blood vessels blood vessel remodeling aortic valve morphogenesis cardiac ventricle morphogenesis cardiac atrium morphogenesis ventricular trabecula myocardium morphogenesis pericardium morphogenesis cell communication signal transduction Notch signaling pathway negative regulation of cell proliferation negative regulation of endothelial cell migration positive regulation of gene expression negative regulation of gene expression cell differentiation T cell differentiation dorsal aorta morphogenesis cellular response to vascular endothelial growth factor stimulus cellular response to fibroblast growth factor stimulus negative regulation of Notch signaling pathway positive regulation of Notch signaling pathway animal organ development system development ventral spinal cord interneuron fate commitment regulation of neural retina development blood vessel lumenization negative regulation of cell migration involved in sprouting angiogenesis negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis positive regulation of neural precursor cell proliferation morphogenesis of an epithelium endothelium development negative regulation of blood vessel endothelial cell migration regulation of neurogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-kp6hx:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0