Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LVREPLSRR RQLLRENFVE TEGEFVFATS LDTKDIEQIA EFLEQSVKDS CEGLMVKTLD VDATYEIAKR SHNWLKLKKD YLDGVGDTLD LVVIGAYLGR GKRAGRYGGF LLASYDEDSE ELQAICKLGT GFSDEELEEH HQSLKALVLP SPRPYVRIDG AVIPDHWLDP SAVWEVKCAD LSLSPIYPAA RGLVDSDKGI SLRFPRFIRV REDKQPEQAT TSAQVACLYR KQSQIQNQQG EDSGSDPEDT Y (670-919 aa encoded by BC108318)
The protein encoded by this gene is involved in the synthesis of asparagine. This gene complements a mutation in the temperature-sensitive hamster mutant ts11, which blocks progression through the G1 phase of the cell cycle at nonpermissive temperature. There are three alternatively spliced transcript variants encoding the same protein described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA ligase (ATP) 1; DNA ligase I; ligase 1; ligase I, DNA, ATP-dependent; MGC117397; MGC130025; polydeoxyribonucleotide synthase [ATP] 1; unnamed protein product
Gene Aliases: hLig1; IMD96; LIGI
UniProt ID: (Human) P18858, (Mouse) P37913
Entrez Gene ID: (Human) 3978, (Mouse) 16881, (Rat) 81513
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support