Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DNAJC2 Antibodies

          Proteintech

          DNAJC2/MPP11 Polyclonal Antibody

          2 References
          View all (7) DNAJC2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DNAJC2/MPP11 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (28)
          DNAJC2/MPP11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DNAJC2/MPP11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 28

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DNAJC2/MPP11 Antibody (11971-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using DNAJC2 antibody 11971-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DNAJC2/MPP11 Antibody in Immunocytochemistry (ICC/IF)
          DNAJC2/MPP11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DNAJC2/MPP11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Western Blot (WB)
          DNAJC2/MPP11 Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          DNAJC2/MPP11 Polyclonal Antibody

          Product Details

          11971-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DNAJC2/MPP11 Fusion Protein Ag2594 (1-167 aa encoded by BC056682)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MLLLPSAAD GRGTAITHAL TSASTLCQVE PVGRWFEAFV KRRNRNASAS FQELEDKKEL SEESEDEELQ LEEFPMLKTL DPKDWKNQDH YAVLGLGHVR YKATQRQIKA AHKAMVLKHH PDKRKAAGEP IKEGDNDYFT CITKGSPGDS YQKLILQLIH LMKCYLIQ (1-167 aa encoded by BC056682)

          Target Information

          EIF4EBP1 is one member of a family of translation repressor proteins. It directly interacts with eukaryotic translation initiation factor 4E (eIF4E), which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: (rat homolog) mouse id-associated protein 1; DnaJ (Hsp40) homolog, subfamily C, member 2; gliosarcoma-related antigen MIDA1; Id associate 1; M-phase phosphoprotein 11; unnamed protein product; zuotin related factor 2; zuotin-related factor 1

          View more View less

          Gene Aliases: MIDA1; MPHOSPH11; MPP11; ZRF1; Zrf2; ZUO1

          View more View less

          UniProt ID: (Human) Q99543, (Mouse) P54103, (Rat) Q7TQ20

          View more View less

          Entrez Gene ID: (Human) 27000, (Mouse) 22791, (Rat) 116456

          View more View less

          Function(s)
          ATPase activator activity chromatin binding RNA binding protein binding Hsp70 protein binding histone binding ribosome binding ubiquitinated histone binding
          Process(es)
          DNA replication chromatin organization regulation of translational fidelity negative regulation of cell growth positive regulation of transcription, DNA-templated 'de novo' cotranslational protein folding negative regulation of DNA biosynthetic process protein folding regulation of cellular response to heat
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch