Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MDASLSMLA NCEKLSLSTN CIEKIANLNG LKNLRILSLG RNNIKNLNGL EAVGDTLEEL WISYNFIEKL KGIHIMKKLK ILYMSNNLVK DWAEFVKLAE LPCLEDLVFV GNPLEEKHSA ENNWIEEATK RVPKLKKLDG TPVIKGDEEE DN (1-151 aa encoded by BC005343)
DNAL1 was identified as a potential candidate gene for primary ciliary dyskinesia (PCD), a genetically heterologous disorder characterized by chronic infections of the upper and lower airways that often leads to permanent lung damage, randomization of left/right body symmetry, and reduced fertility. DNAL1 is reported to be expressed solely in tissues carrying motile cilia for flagella and interacts with DNAH5, a protein that when mutated has been shown to result in PCD. It has been suggested that DNAL1 serves a regulatory function for DNAH5 activity in outer dynein arms of sperm flagella, respiratory cilia, and ependymal cilia. DNAL1 has also been recently identified as an HIV dependency factor (HDF), suggesting that DNAL1 may be an important drug target in HIV treatment. At least two isoforms of DNAL1 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: axonemal dynein light chain 1; DNAL1; dynein light chain 1, axonemal; unnamed protein product
Gene Aliases: 1700010H15Rik; C14orf168; CILD16; Dnalc1; E330027P08Rik; LC1
UniProt ID: (Human) Q4LDG9, (Mouse) Q05A62, (Rat) A0A096MJZ0
Entrez Gene ID: (Human) 83544, (Mouse) 105000, (Rat) 685664
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support