The PA1-73030 antibody reacts with E. coli. And crossreacts with related enterobacteriaceae.
PA1-73030 has been successfully used in ELISA applications.
The PA1-73030 antibody was raised against O and K antigenic serotypes (All types) of Escherichia coli.
Recommended short term storage of less than six months, keep at 2-8°C. For long term storage, store at -20°C.
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.