Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ERK1/2 Antibodies

          Invitrogen

          ERK1/ERK2 Polyclonal Antibody

          Advanced Verification
          5 Published Figures
          4 References
          View all (108) ERK1/2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ERK1/ERK2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (5)
          • Advanced Verification (4)
          ERK1/ERK2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ERK1/ERK2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ERK1/ERK2 Antibody (82380) in ICC/IF

          Immunofluorescence analysis of ERK1/ERK2 was performed using 70% confluent log phase RSC96 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with ERK1/ERK2 Rabbit Polyclonal Antibody (Product # 82380) at 1:100 dilution in 0.1% BSA, incubated at 4 degree Cels... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ERK1/ERK2 Antibody in Immunocytochemistry (ICC/IF)
          ERK1/ERK2 Antibody in Western Blot (WB)
          ERK1/ERK2 Antibody in Western Blot (WB)
          ERK1/ERK2 Antibody in Western Blot (WB)
          ERK1/ERK2 Antibody in Western Blot (WB)
          ERK1/ERK2 Antibody in Western Blot (WB)
          ERK1/ERK2 Antibody in Western Blot (WB)
          ERK1/ERK2 Antibody in Western Blot (WB)
          ERK1/ERK2 Antibody in Western Blot (WB)
          ERK1/ERK2 Antibody
          ERK1/ERK2 Antibody
          ERK1/ERK2 Antibody
          ERK1/ERK2 Antibody
          ERK1/ERK2 Polyclonal Antibody

          Product Details

          82380

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Immunoprecipitation (IP)

          1:50
          -
          Product Specifications

          Species Reactivity

          Bovine, Hamster, Human, Mink, Mouse, Non-human primate, Pig, Rat, Yeast, Zebrafish

          Published species

          Human, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to a sequence in the C-terminus of rat p44 MAP Kinase
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          50 µg/mL

          Amount

          10 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1185592

          Product Specific Information

          It is not recommended to aliquot this antibody.

          Target Information

          ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ERK-1; ERK-2; erk1 erk2; Erk1; cancer susceptibility protein; ERK1b; ERK1c; ERK2; Erk2; cancer susceptibility protein; ERT1; ERT2; Extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; extracellular-signal-regulated kinase 2; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MAPK 1; MAPK 2; MAPK 3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase 1; mitogen-activated 3; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; MNK1; p42; p42 MAP Kinase; p42-MAPK; p44 MAP kinase; p44-ERK1; p44-MAPK; pp42/MAP kinase; protein kinase; protein tyrosine kinase ERK2; unnamed protein product

          View more View less

          Gene Aliases: 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK-1; ERK-2; ERK1; ERK2; ERT1; ERT2; Esrk1; fi06b09; HS44KDAP; HUMKER1A; I79_009500; I79_018350; Mapk; MAPK1; MAPK2; MAPK3; MNK1; Mtap2k; NS13; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; p44; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; PRKM1; PRKM2; PRKM3; wu:fi06b09; zERK1; zERK2

          View more View less

          UniProt ID: (Bovine) P46196, (Human) P28482, (Human) P27361, (Rat) P63086, (Mouse) P63085, (Mouse) Q63844, (Rat) P21708

          View more View less

          Entrez Gene ID: (Bovine) 327672, (Pig) 100153927, (Human) 5594, (Bovine) 531391, (Human) 5595, (Pig) 445013, (Chinese hamster) 100759514, (Rat) 116590, (Mouse) 26413, (Mouse) 26417, (Rat) 50689, (Chinese hamster) 100753149, (Zebrafish) 360144, (Zebrafish) 399480

          View more View less

          Function(s)
          phosphotyrosine binding protein serine/threonine kinase activity MAP kinase kinase activity ATP binding RNA polymerase II carboxy-terminal domain kinase activity phosphatase binding identical protein binding nucleotide binding DNA binding protein kinase activity MAP kinase activity protein binding kinase activity transferase activity protein serine kinase activity DNA-binding transcription factor binding transcription factor binding protein kinase binding mitogen-activated protein kinase kinase kinase binding transferase activity, transferring phosphorus-containing groups scaffold protein binding non-receptor serine/threonine protein kinase
          Process(es)
          activation of MAPK activity protein phosphorylation apoptotic process cellular response to DNA damage stimulus cell cycle positive regulation of gene expression positive regulation of peptidyl-threonine phosphorylation peptidyl-serine phosphorylation peptidyl-threonine phosphorylation cytosine metabolic process regulation of ossification regulation of cellular pH thyroid gland development regulation of protein stability lipopolysaccharide-mediated signaling pathway positive regulation of telomere maintenance via telomerase mammary gland epithelial cell proliferation cellular response to amino acid starvation cellular response to reactive oxygen species response to nicotine ERBB signaling pathway outer ear morphogenesis response to exogenous dsRNA negative regulation of cell differentiation thymus development T cell receptor signaling pathway B cell receptor signaling pathway stress-activated MAPK cascade positive regulation of telomerase activity Bergmann glial cell differentiation long-term synaptic potentiation face development lung morphogenesis trachea formation labyrinthine layer blood vessel development cardiac neural crest cell development involved in heart development ERK1 and ERK2 cascade response to epidermal growth factor cellular response to cadmium ion cellular response to tumor necrosis factor cellular response to granulocyte macrophage colony-stimulating factor stimulus cellular response to dopamine positive regulation of telomere capping MAPK cascade regulation of transcription from RNA polymerase II promoter chemotaxis signal transduction cell surface receptor signaling pathway epidermal growth factor receptor signaling pathway chemical synaptic transmission heart development learning or memory insulin receptor signaling pathway organ morphogenesis positive regulation of macrophage chemotaxis neural crest cell development Schwann cell development positive regulation of telomere maintenance response to lipopolysaccharide regulation of stress-activated MAPK cascade intracellular signal transduction ERBB2-ERBB3 signaling pathway myelination positive regulation of cholesterol biosynthetic process insulin-like growth factor receptor signaling pathway regulation of cytoskeleton organization interleukin-34-mediated signaling pathway chemokine-mediated signaling pathway caveolin-mediated endocytosis regulation of Golgi inheritance positive regulation of macrophage proliferation positive regulation of neuroinflammatory response regulation of early endosome to late endosome transport autophagosome assembly negative regulation of T cell mediated immune response to tumor cell transcription, DNA-templated regulation of gene expression negative regulation of autophagy positive regulation of autophagy phosphorylation sensory perception of pain BMP signaling pathway positive regulation of cyclase activity cellular response to nutrient levels negative regulation of proteasomal ubiquitin-dependent protein catabolic process peptidyl-tyrosine autophosphorylation TORC1 signaling glucose homeostasis signal transduction in response to DNA damage positive regulation of transcription from RNA polymerase II promoter modulation of synaptic transmission protein stabilization negative regulation of T cell activation cartilage development positive regulation of ERK1 and ERK2 cascade interleukin-1-mediated signaling pathway cellular response to mechanical stimulus negative regulation of cholesterol efflux xenophagy negative regulation of TORC1 signaling positive regulation of xenophagy MAPK import into nucleus positive regulation of cell proliferation response to toxic substance positive regulation of cell migration response to estrogen positive regulation of translation positive regulation of transcription, DNA-templated cellular response to organic substance regulation of sequence-specific DNA binding transcription factor activity positive regulation of protein phosphorylation protein complex assembly DNA damage induced protein phosphorylation positive regulation of histone phosphorylation positive regulation of histone acetylation negative regulation of apolipoprotein binding arachidonic acid metabolic process gastrulation eye development brain development central nervous system projection neuron axonogenesis chordate embryonic development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0