Search
Search
FIGURE: 1 / 6
Antibody detects endogenous levels of total FCER1G.
The high affinity IgE receptor is a key molecule involved in allergic reactions. It is a tetramer composed of 1 alpha, 1 beta, and 2 gamma chains. The gamma chains are also subunits of other Fc receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fc fragment of IgE receptor Ig; Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide; Fc receptor gamma-chain; Fc receptor, IgE, high affinity I, gamma polypeptide; Fc-epsilon RI-gamma; Fc-epsilon-receptor gamma-chain protein precursor; FcepsilonRI gamma chain; FcepsilonRIgamma; FceRI gamma; FcRgamma; gamma polypeptide; High affinity immunoglobulin epsilon receptor subunit gamma; IgE Fc receptor subunit gamma; immunoglobulin E receptor, high affinity, gamma chain
Gene Aliases: AI573376; CD23; Fce1g; FcepsilonRI; FCER1G; FcR-gamma; FcR[g]; FCRG; FcRgamma; Ly-50
UniProt ID: (Human) P30273, (Rat) P20411, (Mouse) P20491
Entrez Gene ID: (Human) 2207, (Rat) 25441, (Mouse) 14127
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support