Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
In direct ELISAs, no cross-reactivity with recombinant human FCRL1 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein has four extracellular C2-type immunoglobulin domains, a transmembrane domain and a cytoplasmic domain that contains three immune-receptor tyrosine-based inhibitory motifs. This protein may play a role in the function of memory B-cells in the epithelia. Aberrations in the chromosomal region encoding this gene are associated with non-Hodgkin lymphoma and multiple myeloma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD307d; Fc receptor homolog 4; Fc receptor-like 4; Fc receptor-like protein 4; FcR-like protein 4; FcRH4; hIFGP2; IFGP family protein 2; Immune receptor translocation-associated protein 1; immunoglobulin superfamily Fc receptor, gp42; immunoglobulin superfamily receptor translocation associated 1; MGC150522; MGC150523
Gene Aliases: CD307d; FCRH4; FCRL4; IFGP2; IGFP2; IRTA1
UniProt ID: (Human) Q96PJ5
Entrez Gene ID: (Human) 83417
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support