Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNCCRTLREL EIQVGEKVFK NIKTVMKAFE LIDVNKTGLV RPQELRRVLE TFCMKLRDEE YEKFSKHYNI HKDTAVDYNV FLKNLSINND LNLRYCMGNQ EVSLENQQAK NSKKERLLGS ASSEDIWRNY SLDEIERNFC LQLSKSYEKV EKALSAGDPC KGGYVSFNYL KIVLDTFVYQ IPRRIFIQLM KRFGLKATTK INWKQFLTSF HEPQGLQVSS KGPLTKRNSI NSRNESHKEN IITKLFRHTE DHSASLKKAL LIINTKPDGP ITREEFRYIL NCMAVKLSDS EFKELMQMLD PGDTGVVNTS MFIDLIEENC RMRKTSPCTD AKTPFLLAWD SVEEIVHDTI TRNLQAFYNM LRSYDLGDTG RIGRNNFKKI MHVFCPFLTN AHFIKLCSKI QDIGSGRILY KKLLACIGID GPPTVSPVLV PKDQLLSEHL QKDEQQQPDL SERTKLTEDK TTLTKKMTTE EVIEKFKKCI QQQDPAFKKR FLDFSKEPNG KINVHDFKKV LEDTGMPMDD DQYALLTTKI GFEKEGMSYL DFAAGFEDPP MRGPETTPPQ PPTPSKSYVN SHFITAEECL KLFPRRLKES FRDPYSAFFK TDADRDGIIN MHDLHRLLLH LLLNLKDDEF ERFLGLLGLR LSVTLNFREF QNLCEKRPWR TDEAPQRLIR PKQKVADSEL ACEQAHQYLV TKAKNRWSDL SKNFLETDNE GNGILRRRDI KNALYGFDIP LTPREFEKLW ARYDTEGKGH ITYQEFLQKL GINYSPAVHR PCAEDYFNFM GHFTKPQQLQ EEMKELQQST EKAVAARDKL MDRHQDISKA FTKTDQSKTN YISICKMQEV LEECGCSLTE GELTHLLNSW GVSRHDNAIN YLDFLRAVEN SKSTGAQPKE KEESMPINFA TLNPQEVVRK IQEVVESSQL ALSTAFSALG KEDTGFVKAT EFGQVLKDFC YKLTDNQYHY FLRKLRIHLT PYINWKYFLQ NFSCFLEETA DEWAEKMPKG PPPTSPKATA DRDILARLHK AVTSHYHAIT QEFENFDTMK TNTISREEFR AICNRRVQIL TDEQFGRLWN EMPVNAKGRL KYPDFLSRFS SETAATPMAT GDSAVAQRGS SVPDVSEGTR SALSLPTQEL RPGSKSQSHP CTPASTTVIP GTPPLQNCDP IESRLRKRIQ GCWRQLLKEC KEKDVARQGD INASDFLALV EKFNLDISKE ECQQLIIKYD LKSNGKFAYC DFIQSCVLLL KAKESSLMHR MKIQNAHKMK EAGAETPSFY SALLRIQPKI VHCWRPMRRT FKSYDEAGTG LLSVADFRTV LRQYSINLSE EEFFHILEYY DKTLSSKISY NDFLRAFLQ
This gene encodes a protein which directly binds the oncogene DJ-1 and androgen receptor to form a ternary complex in cells. This binding protein recruits histone-deacetylase complexes in order to repress transcription activity of androgen receptor. This protein may also play a role in spermatogenesis and fertilization. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAP-binding protein complex-interacting protein 1; DJ-1-binding protein; epididymis secretory sperm binding protein; unnamed protein product
Gene Aliases: dJ185D5.1; DJBP; HSCBCIP1
UniProt ID: (Human) Q5THR3
Entrez Gene ID: (Human) 64800
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support