PA5-27465 targets Folate Receptor alpha in IF, IHC (P), and WB applications and shows reactivity with Human samples.
The PA5-27465 immunogen is recombinant fragment corresponding to a region within amino acids 7 and 234 of Human Folate Receptor alpha.
The protein encoded by this gene is a member of the folate receptor family. Members of this gene family bind folic acid and its reduced derivatives, and transport 5-methyltetrahydrofolate into cells. This gene product is a secreted protein that either anchors to membranes via a glycosyl-phosphatidylinositol linkage or exists in a soluble form. Mutations in this gene have been associated with neurodegeneration due to cerebral folate transport deficiency. Due to the presence of two promoters, multiple transcription start sites, and alternative splicing, multiple transcript variants encoding the same protein have been found for this gene.